Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34653771-34654280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL11RA
Entrez Gene ID
3590
Gene Name
interleukin 11 receptor subunit alpha
Gene Aliases
CRSDA
UniGene
Hs.591088
Gene Chromosome Location
Chr.9: 34652185-34661902 on Build GRCh38
Transcripts
NM_001142784.2 NR_052010.1

SNP Detail

SNP ID Chromosome Location
rs1007063562 Chr.9: 34654007-34654007 on Build GRCh38
rs1056964651 Chr.9: 34654216-34654216 on Build GRCh38
rs915242575 Chr.9: 34654124-34654124 on Build GRCh38
rs1010025315 Chr.9: 34653837-34653837 on Build GRCh38
rs12551907 Chr.9: 34653872-34653872 on Build GRCh38
rs529050372 Chr.9: 34654205-34654205 on Build GRCh38
rs899580792 Chr.9: 34654255-34654255 on Build GRCh38
rs749770212 Chr.9: 34653941-34653941 on Build GRCh38
rs997689386 Chr.9: 34654253-34654253 on Build GRCh38
rs1048113026 Chr.9: 34653993-34653993 on Build GRCh38
rs1017550051 Chr.9: 34654242-34654242 on Build GRCh38
rs1018408100 Chr.9: 34654019-34654019 on Build GRCh38
rs941050040 Chr.9: 34654083-34654083 on Build GRCh38
rs765071414 Chr.9: 34654236-34654236 on Build GRCh38
rs3808868 Chr.9: 34654145-34654145 on Build GRCh38
rs11575586 Chr.9: 34654109-34654109 on Build GRCh38
rs769073167 Chr.9: 34654012-34654012 on Build GRCh38
rs13300653 Chr.9: 34653824-34653824 on Build GRCh38
rs1022300244 Chr.9: 34653873-34653873 on Build GRCh38
rs982911203 Chr.9: 34654052-34654052 on Build GRCh38
rs951171146 Chr.9: 34654015-34654015 on Build GRCh38
rs945492584 Chr.9: 34654179-34654179 on Build GRCh38
rs542721202 Chr.9: 34653796-34653796 on Build GRCh38
rs956675578 Chr.9: 34654102-34654102 on Build GRCh38
rs966252752 Chr.9: 34653988-34653988 on Build GRCh38
rs998433578 Chr.9: 34654069-34654069 on Build GRCh38
rs903821790 Chr.9: 34653914-34653914 on Build GRCh38
rs763614273 Chr.9: 34653828-34653828 on Build GRCh38
rs545083705 Chr.9: 34654133-34654133 on Build GRCh38
rs34606053 Chr.9: 34654132-34654132 on Build GRCh38
rs1048020156 Chr.9: 34654252-34654252 on Build GRCh38
rs929913294 Chr.9: 34654250-34654250 on Build GRCh38
rs1057016809 Chr.9: 34653934-34653934 on Build GRCh38
rs533099117 Chr.9: 34654027-34654027 on Build GRCh38
rs993822294 Chr.9: 34653964-34653964 on Build GRCh38
rs965929587 Chr.9: 34654057-34654057 on Build GRCh38
rs892589370 Chr.9: 34653829-34653829 on Build GRCh38
rs896818158 Chr.9: 34653935-34653935 on Build GRCh38
rs377087199 Chr.9: 34653800-34653800 on Build GRCh38
rs892258017 Chr.9: 34654150-34654150 on Build GRCh38
rs1026861447 Chr.9: 34654009-34654009 on Build GRCh38
rs1017835138 Chr.9: 34653961-34653961 on Build GRCh38
rs927146644 Chr.9: 34654182-34654182 on Build GRCh38
rs761546354 Chr.9: 34653828-34653828 on Build GRCh38
rs1031289812 Chr.9: 34654098-34654098 on Build GRCh38
rs11575585 Chr.9: 34653997-34653997 on Build GRCh38

View Full Product Details