Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34656944-34657438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL11RA
Entrez Gene ID
3590
Gene Name
interleukin 11 receptor subunit alpha
Gene Aliases
CRSDA
UniGene
Hs.591088
Gene Chromosome Location
Chr.9: 34652185-34661902 on Build GRCh38
Transcripts
NM_001142784.2 NR_052010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6837405 c.440C>G p.S147C Chr.9: 34657143-34657143 on Build GRCh38
4373965 c.344G>A p.R115H Chr.9: 34657047-34657047 on Build GRCh38
6269600 c.349G>T p.V117F Chr.9: 34657052-34657052 on Build GRCh38
5411029 c.400C>T p.P134S Chr.9: 34657103-34657103 on Build GRCh38
5547395 c.434_446+2delTCACCTCCTACAGGT p.? Chr.9: 34657137-34657151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780262490 Chr.9: 34656999-34656999 on Build GRCh38
rs147692403 Chr.9: 34657117-34657117 on Build GRCh38
rs201353728 Chr.9: 34657323-34657323 on Build GRCh38
rs760131430 Chr.9: 34657148-34657148 on Build GRCh38
rs951958006 Chr.9: 34657224-34657224 on Build GRCh38
rs113716899 Chr.9: 34657072-34657072 on Build GRCh38
rs535435563 Chr.9: 34657011-34657011 on Build GRCh38
rs533937850 Chr.9: 34656983-34656983 on Build GRCh38
rs560967238 Chr.9: 34657360-34657360 on Build GRCh38
rs755274967 Chr.9: 34657199-34657199 on Build GRCh38
rs750878767 Chr.9: 34657069-34657069 on Build GRCh38
rs137972337 Chr.9: 34657043-34657043 on Build GRCh38
rs748187476 Chr.9: 34657115-34657115 on Build GRCh38
rs143961331 Chr.9: 34657327-34657327 on Build GRCh38
rs767925757 Chr.9: 34657059-34657059 on Build GRCh38
rs920225288 Chr.9: 34657219-34657219 on Build GRCh38
rs754095914 Chr.9: 34657078-34657078 on Build GRCh38
rs971726885 Chr.9: 34657215-34657215 on Build GRCh38
rs11575591 Chr.9: 34657398-34657398 on Build GRCh38
rs371329032 Chr.9: 34657131-34657131 on Build GRCh38
rs376279953 Chr.9: 34657271-34657271 on Build GRCh38
rs879517554 Chr.9: 34657353-34657353 on Build GRCh38
rs181486891 Chr.9: 34657385-34657385 on Build GRCh38
rs755185119 Chr.9: 34657080-34657080 on Build GRCh38
rs775152788 Chr.9: 34657139-34657139 on Build GRCh38
rs376790435 Chr.9: 34657201-34657201 on Build GRCh38
rs377421299 Chr.9: 34657130-34657130 on Build GRCh38
rs777601555 Chr.9: 34657118-34657118 on Build GRCh38
rs771524578 Chr.9: 34657410-34657410 on Build GRCh38
rs767002833 Chr.9: 34657166-34657166 on Build GRCh38
rs374717464 Chr.9: 34657386-34657386 on Build GRCh38
rs781670516 Chr.9: 34656984-34656984 on Build GRCh38
rs544363313 Chr.9: 34657343-34657343 on Build GRCh38
rs767258365 Chr.9: 34657310-34657311 on Build GRCh38
rs751596237 Chr.9: 34657275-34657275 on Build GRCh38
rs754185847 Chr.9: 34657192-34657192 on Build GRCh38
rs961819228 Chr.9: 34657209-34657209 on Build GRCh38
rs764138471 Chr.9: 34657368-34657368 on Build GRCh38
rs758439988 Chr.9: 34657259-34657259 on Build GRCh38
rs752902570 Chr.9: 34657364-34657364 on Build GRCh38
rs370362629 Chr.9: 34657007-34657007 on Build GRCh38
rs766685036 Chr.9: 34657073-34657073 on Build GRCh38
rs761135908 Chr.9: 34657179-34657179 on Build GRCh38
rs934721431 Chr.9: 34657354-34657354 on Build GRCh38
rs35093960 Chr.9: 34657297-34657298 on Build GRCh38
rs776445508 Chr.9: 34657040-34657040 on Build GRCh38
rs950183315 Chr.9: 34656976-34656976 on Build GRCh38
rs553842580 Chr.9: 34657015-34657015 on Build GRCh38
rs773141225 Chr.9: 34657022-34657022 on Build GRCh38
rs746893925 Chr.9: 34657119-34657119 on Build GRCh38
rs761400983 Chr.9: 34656981-34656981 on Build GRCh38
rs766793151 Chr.9: 34657188-34657188 on Build GRCh38
rs544846004 Chr.9: 34657376-34657376 on Build GRCh38
rs763623246 Chr.9: 34657151-34657151 on Build GRCh38
rs200472583 Chr.9: 34657093-34657093 on Build GRCh38
rs201997285 Chr.9: 34657047-34657047 on Build GRCh38
rs749476915 Chr.9: 34657003-34657003 on Build GRCh38
rs747673752 Chr.9: 34657409-34657409 on Build GRCh38
rs368236740 Chr.9: 34657355-34657355 on Build GRCh38
rs762377674 Chr.9: 34657150-34657150 on Build GRCh38
rs781199286 Chr.9: 34657303-34657303 on Build GRCh38
rs201354346 Chr.9: 34657372-34657372 on Build GRCh38
rs757247176 Chr.9: 34657286-34657286 on Build GRCh38
rs999293166 Chr.9: 34657012-34657012 on Build GRCh38
rs769319734 Chr.9: 34657134-34657134 on Build GRCh38
rs200417443 Chr.9: 34657031-34657031 on Build GRCh38
rs779105192 Chr.9: 34657111-34657111 on Build GRCh38
rs756415535 Chr.9: 34656986-34656986 on Build GRCh38
rs376621888 Chr.9: 34656985-34656985 on Build GRCh38
rs11575590 Chr.9: 34657393-34657393 on Build GRCh38
rs771887245 Chr.9: 34657018-34657018 on Build GRCh38
rs1020296640 Chr.9: 34657228-34657228 on Build GRCh38
rs200580340 Chr.9: 34657046-34657046 on Build GRCh38
rs199757118 Chr.9: 34657336-34657336 on Build GRCh38
rs368126626 Chr.9: 34657356-34657356 on Build GRCh38
rs745660566 Chr.9: 34657311-34657311 on Build GRCh38
rs990584599 Chr.9: 34657357-34657357 on Build GRCh38
rs745584639 Chr.9: 34657133-34657133 on Build GRCh38
rs773955066 Chr.9: 34657010-34657010 on Build GRCh38
rs755888812 Chr.9: 34657378-34657378 on Build GRCh38
rs759241972 Chr.9: 34657041-34657041 on Build GRCh38
rs372556212 Chr.9: 34657371-34657371 on Build GRCh38
rs886110038 Chr.9: 34657008-34657008 on Build GRCh38
rs387906787 Chr.9: 34657331-34657331 on Build GRCh38
rs761240511 Chr.9: 34657342-34657342 on Build GRCh38
rs779864103 Chr.9: 34657381-34657381 on Build GRCh38
rs752917080 Chr.9: 34657244-34657244 on Build GRCh38
rs111658229 Chr.9: 34656995-34656995 on Build GRCh38
rs949949361 Chr.9: 34657328-34657328 on Build GRCh38
rs574264852 Chr.9: 34657202-34657202 on Build GRCh38
rs769572458 Chr.9: 34657312-34657312 on Build GRCh38
rs773734789 Chr.9: 34657152-34657152 on Build GRCh38
rs770808785 Chr.9: 34657035-34657035 on Build GRCh38
rs1009159318 Chr.9: 34657147-34657147 on Build GRCh38
rs996772251 Chr.9: 34657107-34657107 on Build GRCh38
rs924728040 Chr.9: 34657351-34657351 on Build GRCh38
rs768087208 Chr.9: 34657335-34657335 on Build GRCh38

View Full Product Details