Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35737660-35738158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA2
Entrez Gene ID
57704
Gene Name
glucosylceramidase beta 2
Gene Aliases
AD035, NLGase, SPG46
UniGene
Hs.443134
Gene Chromosome Location
Chr.9: 35736862-35749255 on Build GRCh38
Transcripts
NM_020944.2 XM_005251526.4 XM_006716809.3 XM_011517973.2 XM_017014937.1 XM_017014938.1 XM_017014939.1 XM_017014940.1 XM_017014941.1 XM_017014942.1 XM_017014943.1 XM_017014944.1 XM_017014945.1 XM_017014946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3848489 c.2495T>C p.M832T Chr.9: 35737758-35737758 on Build GRCh38
4872310 c.2408C>T p.A803V Chr.9: 35737863-35737863 on Build GRCh38
3848495 c.2469T>G p.G823G Chr.9: 35737784-35737784 on Build GRCh38
5001604 c.2263G>T p.A755S Chr.9: 35738087-35738087 on Build GRCh38
3664335 c.2371A>G p.I791V Chr.9: 35737900-35737900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748939597 Chr.9: 35737920-35737920 on Build GRCh38
rs773822816 Chr.9: 35737689-35737691 on Build GRCh38
rs770645171 Chr.9: 35738041-35738041 on Build GRCh38
rs762088879 Chr.9: 35737979-35737979 on Build GRCh38
rs750727245 Chr.9: 35737998-35737998 on Build GRCh38
rs143345363 Chr.9: 35737886-35737886 on Build GRCh38
rs755079907 Chr.9: 35737718-35737718 on Build GRCh38
rs760546007 Chr.9: 35737973-35737976 on Build GRCh38
rs757586951 Chr.9: 35737769-35737769 on Build GRCh38
rs374440648 Chr.9: 35737775-35737775 on Build GRCh38
rs200851781 Chr.9: 35738018-35738019 on Build GRCh38
rs947546049 Chr.9: 35738125-35738125 on Build GRCh38
rs769425692 Chr.9: 35737814-35737814 on Build GRCh38
rs375817378 Chr.9: 35738010-35738010 on Build GRCh38
rs201520272 Chr.9: 35738122-35738122 on Build GRCh38
rs774411642 Chr.9: 35737840-35737840 on Build GRCh38
rs751575696 Chr.9: 35737717-35737717 on Build GRCh38
rs148461769 Chr.9: 35738043-35738043 on Build GRCh38
rs62637647 Chr.9: 35738110-35738110 on Build GRCh38
rs746694187 Chr.9: 35737975-35737975 on Build GRCh38
rs778985234 Chr.9: 35737828-35737828 on Build GRCh38
rs898668285 Chr.9: 35737870-35737870 on Build GRCh38
rs766699931 Chr.9: 35738121-35738121 on Build GRCh38
rs546011240 Chr.9: 35737865-35737865 on Build GRCh38
rs757588425 Chr.9: 35737899-35737899 on Build GRCh38
rs747616868 Chr.9: 35737799-35737799 on Build GRCh38
rs768775425 Chr.9: 35738077-35738077 on Build GRCh38
rs200894732 Chr.9: 35738130-35738130 on Build GRCh38
rs781057774 Chr.9: 35738019-35738021 on Build GRCh38
rs78562202 Chr.9: 35738033-35738033 on Build GRCh38
rs570984524 Chr.9: 35738024-35738024 on Build GRCh38
rs73438752 Chr.9: 35737710-35737710 on Build GRCh38
rs753906293 Chr.9: 35737686-35737686 on Build GRCh38
rs772758704 Chr.9: 35737822-35737822 on Build GRCh38
rs755506697 Chr.9: 35738127-35738127 on Build GRCh38
rs199765267 Chr.9: 35737900-35737900 on Build GRCh38
rs772886623 Chr.9: 35737918-35737918 on Build GRCh38
rs746421010 Chr.9: 35737774-35737774 on Build GRCh38
rs772670813 Chr.9: 35737957-35737963 on Build GRCh38
rs547965402 Chr.9: 35737955-35737955 on Build GRCh38
rs916407293 Chr.9: 35738054-35738054 on Build GRCh38
rs958126086 Chr.9: 35738022-35738022 on Build GRCh38
rs752958819 Chr.9: 35737735-35737735 on Build GRCh38
rs766035120 Chr.9: 35738007-35738008 on Build GRCh38
rs940141611 Chr.9: 35737787-35737787 on Build GRCh38
rs371965145 Chr.9: 35737961-35737961 on Build GRCh38
rs748969125 Chr.9: 35738036-35738036 on Build GRCh38
rs763842418 Chr.9: 35738021-35738022 on Build GRCh38
rs763269505 Chr.9: 35738113-35738113 on Build GRCh38
rs756161551 Chr.9: 35737884-35737884 on Build GRCh38
rs972531833 Chr.9: 35737939-35737939 on Build GRCh38
rs771767001 Chr.9: 35737867-35737868 on Build GRCh38
rs772024917 Chr.9: 35738047-35738047 on Build GRCh38
rs113327265 Chr.9: 35737742-35737742 on Build GRCh38
rs529175802 Chr.9: 35737805-35737805 on Build GRCh38
rs775468187 Chr.9: 35738050-35738050 on Build GRCh38
rs746367894 Chr.9: 35737902-35737902 on Build GRCh38
rs368660186 Chr.9: 35737984-35737984 on Build GRCh38
rs758834761 Chr.9: 35737907-35737907 on Build GRCh38
rs764122231 Chr.9: 35737971-35737971 on Build GRCh38
rs752759752 Chr.9: 35737882-35737882 on Build GRCh38
rs562375787 Chr.9: 35737914-35737914 on Build GRCh38
rs760579817 Chr.9: 35738057-35738057 on Build GRCh38
rs776241385 Chr.9: 35737692-35737692 on Build GRCh38
rs202031683 Chr.9: 35738111-35738111 on Build GRCh38
rs1026317768 Chr.9: 35737956-35737956 on Build GRCh38
rs777387371 Chr.9: 35737922-35737922 on Build GRCh38
rs201258683 Chr.9: 35737719-35737719 on Build GRCh38
rs780552477 Chr.9: 35737911-35737911 on Build GRCh38
rs144531146 Chr.9: 35737915-35737915 on Build GRCh38
rs777863053 Chr.9: 35737754-35737754 on Build GRCh38
rs372001753 Chr.9: 35738021-35738021 on Build GRCh38
rs776190100 Chr.9: 35737776-35737776 on Build GRCh38
rs756428994 Chr.9: 35737745-35737745 on Build GRCh38
rs766315448 Chr.9: 35737836-35737836 on Build GRCh38
rs370957405 Chr.9: 35738000-35738000 on Build GRCh38
rs779354356 Chr.9: 35737773-35737773 on Build GRCh38
rs770839470 Chr.9: 35737942-35737942 on Build GRCh38
rs749610229 Chr.9: 35737763-35737763 on Build GRCh38
rs775313194 Chr.9: 35737960-35737960 on Build GRCh38
rs568813842 Chr.9: 35738091-35738091 on Build GRCh38
rs747824980 Chr.9: 35737735-35737736 on Build GRCh38
rs776507361 Chr.9: 35738094-35738094 on Build GRCh38
rs759515533 Chr.9: 35738070-35738071 on Build GRCh38
rs765448520 Chr.9: 35737986-35737986 on Build GRCh38
rs766366640 Chr.9: 35737716-35737716 on Build GRCh38
rs745609492 Chr.9: 35738046-35738046 on Build GRCh38
rs767703677 Chr.9: 35737880-35737880 on Build GRCh38
rs754039859 Chr.9: 35737891-35737891 on Build GRCh38
rs772992019 Chr.9: 35737715-35737715 on Build GRCh38
rs1008414642 Chr.9: 35737923-35737923 on Build GRCh38
rs774358705 Chr.9: 35737947-35737947 on Build GRCh38
rs200544152 Chr.9: 35737885-35737885 on Build GRCh38
rs928671211 Chr.9: 35738048-35738050 on Build GRCh38
rs938749085 Chr.9: 35738080-35738080 on Build GRCh38

View Full Product Details