Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37800991-37801519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF10
Entrez Gene ID
79269
Gene Name
DDB1 and CUL4 associated factor 10
Gene Aliases
WDR32
UniGene
Hs.118394
Gene Chromosome Location
Chr.9: 37800554-37867669 on Build GRCh38
Transcripts
NM_001286810.1 NM_024345.4 XM_005251577.3 XM_005251578.3 XM_017015128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6411363 c.280C>A p.P94T Chr.9: 37801146-37801146 on Build GRCh38
5559958 c.212T>C p.L71P Chr.9: 37801078-37801078 on Build GRCh38
7273100 c.286C>T p.R96W Chr.9: 37801152-37801152 on Build GRCh38
6409053 c.295G>A p.G99R Chr.9: 37801161-37801161 on Build GRCh38
6423825 c.433T>A p.F145I Chr.9: 37801299-37801299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370190180 Chr.9: 37801388-37801388 on Build GRCh38
rs746844095 Chr.9: 37801229-37801229 on Build GRCh38
rs765347190 Chr.9: 37801100-37801100 on Build GRCh38
rs938266050 Chr.9: 37801202-37801202 on Build GRCh38
rs532027208 Chr.9: 37801186-37801186 on Build GRCh38
rs762896759 Chr.9: 37801241-37801241 on Build GRCh38
rs116379925 Chr.9: 37801465-37801465 on Build GRCh38
rs199884641 Chr.9: 37801235-37801235 on Build GRCh38
rs751201578 Chr.9: 37801295-37801295 on Build GRCh38
rs1006933986 Chr.9: 37801345-37801345 on Build GRCh38
rs753239159 Chr.9: 37801155-37801155 on Build GRCh38
rs760907572 Chr.9: 37801411-37801411 on Build GRCh38
rs573368194 Chr.9: 37801336-37801336 on Build GRCh38
rs769555483 Chr.9: 37801239-37801239 on Build GRCh38
rs764459431 Chr.9: 37801283-37801283 on Build GRCh38
rs188933688 Chr.9: 37801370-37801370 on Build GRCh38
rs761258998 Chr.9: 37801275-37801275 on Build GRCh38
rs778807521 Chr.9: 37801358-37801358 on Build GRCh38
rs149691291 Chr.9: 37801350-37801350 on Build GRCh38
rs768184466 Chr.9: 37801148-37801148 on Build GRCh38
rs887543631 Chr.9: 37801030-37801030 on Build GRCh38
rs370374517 Chr.9: 37801226-37801226 on Build GRCh38
rs973684771 Chr.9: 37801430-37801430 on Build GRCh38
rs990783446 Chr.9: 37801481-37801481 on Build GRCh38
rs902897178 Chr.9: 37801070-37801070 on Build GRCh38
rs749174659 Chr.9: 37801449-37801449 on Build GRCh38
rs751998238 Chr.9: 37801123-37801123 on Build GRCh38
rs866015307 Chr.9: 37801029-37801029 on Build GRCh38
rs963227758 Chr.9: 37801426-37801426 on Build GRCh38
rs927939983 Chr.9: 37801195-37801195 on Build GRCh38
rs756767243 Chr.9: 37801054-37801054 on Build GRCh38
rs1010014405 Chr.9: 37801277-37801277 on Build GRCh38
rs866956931 Chr.9: 37801090-37801090 on Build GRCh38
rs751906495 Chr.9: 37801017-37801017 on Build GRCh38
rs757021631 Chr.9: 37801161-37801161 on Build GRCh38
rs74987730 Chr.9: 37801447-37801447 on Build GRCh38
rs948228870 Chr.9: 37801421-37801421 on Build GRCh38
rs200916317 Chr.9: 37801042-37801042 on Build GRCh38
rs751004535 Chr.9: 37801433-37801433 on Build GRCh38
rs777534271 Chr.9: 37801365-37801365 on Build GRCh38
rs930104489 Chr.9: 37801185-37801185 on Build GRCh38
rs550914416 Chr.9: 37801233-37801233 on Build GRCh38
rs755837354 Chr.9: 37801446-37801446 on Build GRCh38
rs908948299 Chr.9: 37801193-37801193 on Build GRCh38
rs760033376 Chr.9: 37801269-37801269 on Build GRCh38
rs764701106 Chr.9: 37801414-37801414 on Build GRCh38
rs374346144 Chr.9: 37801400-37801400 on Build GRCh38
rs755556944 Chr.9: 37801032-37801032 on Build GRCh38
rs766654030 Chr.9: 37801115-37801115 on Build GRCh38
rs1022658292 Chr.9: 37801036-37801036 on Build GRCh38
rs938344323 Chr.9: 37801441-37801441 on Build GRCh38
rs965376632 Chr.9: 37801361-37801361 on Build GRCh38
rs780969667 Chr.9: 37801314-37801314 on Build GRCh38
rs776609526 Chr.9: 37801238-37801238 on Build GRCh38
rs745311749 Chr.9: 37801207-37801207 on Build GRCh38
rs184138579 Chr.9: 37801273-37801273 on Build GRCh38
rs1011054832 Chr.9: 37801053-37801053 on Build GRCh38
rs145533200 Chr.9: 37801373-37801373 on Build GRCh38
rs574334180 Chr.9: 37801462-37801462 on Build GRCh38
rs976994870 Chr.9: 37801153-37801153 on Build GRCh38
rs779142860 Chr.9: 37801454-37801454 on Build GRCh38
rs75986742 Chr.9: 37801488-37801488 on Build GRCh38
rs754834552 Chr.9: 37801303-37801303 on Build GRCh38
rs551735178 Chr.9: 37801196-37801196 on Build GRCh38
rs985859394 Chr.9: 37801128-37801128 on Build GRCh38
rs753748989 Chr.9: 37801065-37801065 on Build GRCh38
rs1056956462 Chr.9: 37801258-37801258 on Build GRCh38
rs148430370 Chr.9: 37801385-37801385 on Build GRCh38
rs575473033 Chr.9: 37801386-37801386 on Build GRCh38
rs998606843 Chr.9: 37801297-37801297 on Build GRCh38
rs571881171 Chr.9: 37801217-37801217 on Build GRCh38
rs905772451 Chr.9: 37801037-37801037 on Build GRCh38
rs946874198 Chr.9: 37801240-37801240 on Build GRCh38
rs778487474 Chr.9: 37801058-37801058 on Build GRCh38
rs201208093 Chr.9: 37801377-37801377 on Build GRCh38
rs746754132 Chr.9: 37801080-37801080 on Build GRCh38
rs776521677 Chr.9: 37801091-37801091 on Build GRCh38
rs1002967034 Chr.9: 37801247-37801247 on Build GRCh38
rs868317373 Chr.9: 37801027-37801027 on Build GRCh38
rs765870449 Chr.9: 37801422-37801422 on Build GRCh38
rs774690008 Chr.9: 37801268-37801268 on Build GRCh38
rs781703007 Chr.9: 37801033-37801034 on Build GRCh38
rs982556875 Chr.9: 37801189-37801189 on Build GRCh38
rs376076163 Chr.9: 37801288-37801288 on Build GRCh38
rs778486047 Chr.9: 37801192-37801192 on Build GRCh38
rs770921332 Chr.9: 37801451-37801451 on Build GRCh38
rs761848631 Chr.9: 37801095-37801095 on Build GRCh38
rs772963081 Chr.9: 37801403-37801403 on Build GRCh38
rs745529237 Chr.9: 37801061-37801061 on Build GRCh38
rs781746797 Chr.9: 37801041-37801041 on Build GRCh38
rs202086101 Chr.9: 37801274-37801274 on Build GRCh38
rs771553791 Chr.9: 37801063-37801063 on Build GRCh38
rs772358218 Chr.9: 37801391-37801391 on Build GRCh38
rs929405295 Chr.9: 37801175-37801175 on Build GRCh38
rs559843565 Chr.9: 37801470-37801470 on Build GRCh38
rs768609407 Chr.9: 37801082-37801082 on Build GRCh38
rs973227491 Chr.9: 37801152-37801152 on Build GRCh38
rs762510038 Chr.9: 37801416-37801416 on Build GRCh38
rs946984942 Chr.9: 37801489-37801489 on Build GRCh38
rs752571640 Chr.9: 37801443-37801443 on Build GRCh38
rs766741781 Chr.9: 37801260-37801260 on Build GRCh38
rs1029995450 Chr.9: 37801076-37801076 on Build GRCh38
rs985993220 Chr.9: 37801125-37801125 on Build GRCh38
rs758090582 Chr.9: 37801208-37801208 on Build GRCh38
rs773373985 Chr.9: 37801102-37801102 on Build GRCh38
rs181616247 Chr.9: 37801436-37801436 on Build GRCh38
rs1018781166 Chr.9: 37801351-37801351 on Build GRCh38
rs1017451713 Chr.9: 37801135-37801135 on Build GRCh38
rs1021805011 Chr.9: 37801056-37801056 on Build GRCh38
rs775273508 Chr.9: 37801071-37801071 on Build GRCh38
rs919340667 Chr.9: 37801160-37801160 on Build GRCh38
rs763245828 Chr.9: 37801111-37801111 on Build GRCh38
rs977176099 Chr.9: 37801367-37801367 on Build GRCh38

View Full Product Details