Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112486623-112487111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf147
Entrez Gene ID
100133204
Gene Name
chromosome 9 open reading frame 147
Gene Aliases
bA32M23.3
UniGene
Hs.662552
Gene Chromosome Location
Chr.9: 112409446-112490453 on Build GRCh38
Transcripts
NM_001287036.1 NM_001287038.1 NM_133465.3

Gene Symbol
KIAA1958
Entrez Gene ID
158405
Gene Name
KIAA1958
Gene Aliases
-
UniGene
Hs.656619
Gene Chromosome Location
Chr.9: 112486968-112665311 on Build GRCh38
Transcripts
NM_001287036.1 NM_001287038.1 NM_133465.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1624433 c.226+2T>C p.? Chr.9: 112486977-112486977 on Build GRCh38
5787148 c.187G>A p.A63T Chr.9: 112487018-112487018 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1017066461 Chr.9: 112486954-112486954 on Build GRCh38
rs776931728 Chr.9: 112487043-112487043 on Build GRCh38
rs567309161 Chr.9: 112487001-112487001 on Build GRCh38
rs536919014 Chr.9: 112486947-112486947 on Build GRCh38
rs1036389092 Chr.9: 112486856-112486883 on Build GRCh38
rs536390881 Chr.9: 112487013-112487013 on Build GRCh38
rs1007259430 Chr.9: 112486643-112486643 on Build GRCh38
rs551736914 Chr.9: 112486688-112486688 on Build GRCh38
rs910010962 Chr.9: 112486727-112486727 on Build GRCh38
rs1017694917 Chr.9: 112486651-112486651 on Build GRCh38
rs571537400 Chr.9: 112486711-112486711 on Build GRCh38
rs557123302 Chr.9: 112486996-112486996 on Build GRCh38
rs1033657529 Chr.9: 112486971-112486971 on Build GRCh38
rs1007722527 Chr.9: 112486916-112486916 on Build GRCh38
rs1028283809 Chr.9: 112486804-112486804 on Build GRCh38
rs887276031 Chr.9: 112486946-112486946 on Build GRCh38
rs866152278 Chr.9: 112486647-112486647 on Build GRCh38
rs762317958 Chr.9: 112487047-112487047 on Build GRCh38
rs962787476 Chr.9: 112486957-112486957 on Build GRCh38
rs1038671527 Chr.9: 112486922-112486922 on Build GRCh38
rs962450371 Chr.9: 112486928-112486928 on Build GRCh38
rs769100439 Chr.9: 112486988-112486988 on Build GRCh38
rs998859521 Chr.9: 112486657-112486657 on Build GRCh38
rs981282295 Chr.9: 112486963-112486963 on Build GRCh38
rs982812805 Chr.9: 112486686-112486686 on Build GRCh38
rs913717980 Chr.9: 112486801-112486801 on Build GRCh38
rs939259305 Chr.9: 112486787-112486787 on Build GRCh38
rs868199302 Chr.9: 112486870-112486870 on Build GRCh38
rs1029758941 Chr.9: 112486666-112486666 on Build GRCh38
rs6477940 Chr.9: 112486982-112486982 on Build GRCh38
rs7871916 Chr.9: 112486652-112486652 on Build GRCh38
rs149585063 Chr.9: 112486681-112486681 on Build GRCh38
rs758243573 Chr.9: 112487012-112487019 on Build GRCh38
rs911872509 Chr.9: 112486896-112486896 on Build GRCh38
rs987907111 Chr.9: 112486825-112486825 on Build GRCh38
rs980763896 Chr.9: 112486740-112486740 on Build GRCh38
rs553027026 Chr.9: 112487032-112487032 on Build GRCh38
rs1029895181 Chr.9: 112486944-112486944 on Build GRCh38
rs975330150 Chr.9: 112486799-112486799 on Build GRCh38
rs955267507 Chr.9: 112486809-112486809 on Build GRCh38
rs992073405 Chr.9: 112486993-112486993 on Build GRCh38
rs903569138 Chr.9: 112486897-112486897 on Build GRCh38
rs998436692 Chr.9: 112486937-112486937 on Build GRCh38
rs925124181 Chr.9: 112487074-112487074 on Build GRCh38
rs765788212 Chr.9: 112487072-112487072 on Build GRCh38
rs957160976 Chr.9: 112486786-112486786 on Build GRCh38
rs1043486723 Chr.9: 112486874-112486874 on Build GRCh38
rs963026631 Chr.9: 112486728-112486728 on Build GRCh38
rs746108473 Chr.9: 112487069-112487069 on Build GRCh38
rs1033274109 Chr.9: 112486775-112486775 on Build GRCh38
rs1056424397 Chr.9: 112486800-112486800 on Build GRCh38
rs781202169 Chr.9: 112487068-112487069 on Build GRCh38
rs903819367 Chr.9: 112486683-112486683 on Build GRCh38

View Full Product Details