Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112486960-112487499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf147
Entrez Gene ID
100133204
Gene Name
chromosome 9 open reading frame 147
Gene Aliases
bA32M23.3
UniGene
Hs.662552
Gene Chromosome Location
Chr.9: 112409446-112490453 on Build GRCh38
Transcripts
NM_001287036.1 NM_001287038.1 NM_133465.3

Gene Symbol
KIAA1958
Entrez Gene ID
158405
Gene Name
KIAA1958
Gene Aliases
-
UniGene
Hs.656619
Gene Chromosome Location
Chr.9: 112486968-112665311 on Build GRCh38
Transcripts
NM_001287036.1 NM_001287038.1 NM_133465.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1624433 c.226+2T>C p.? Chr.9: 112486977-112486977 on Build GRCh38
5787148 c.187G>A p.A63T Chr.9: 112487018-112487018 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs898673268 Chr.9: 112487194-112487194 on Build GRCh38
rs572934785 Chr.9: 112487097-112487097 on Build GRCh38
rs896526297 Chr.9: 112487424-112487424 on Build GRCh38
rs903784156 Chr.9: 112487405-112487405 on Build GRCh38
rs1014078702 Chr.9: 112487451-112487451 on Build GRCh38
rs575894483 Chr.9: 112487225-112487225 on Build GRCh38
rs933022261 Chr.9: 112487129-112487129 on Build GRCh38
rs913704820 Chr.9: 112487345-112487345 on Build GRCh38
rs920965502 Chr.9: 112487319-112487319 on Build GRCh38
rs776931728 Chr.9: 112487043-112487043 on Build GRCh38
rs961062624 Chr.9: 112487240-112487240 on Build GRCh38
rs558652101 Chr.9: 112487168-112487168 on Build GRCh38
rs1002636259 Chr.9: 112487217-112487217 on Build GRCh38
rs1002156021 Chr.9: 112487417-112487417 on Build GRCh38
rs1045828630 Chr.9: 112487360-112487360 on Build GRCh38
rs190321439 Chr.9: 112487228-112487228 on Build GRCh38
rs567309161 Chr.9: 112487001-112487001 on Build GRCh38
rs1017033956 Chr.9: 112487175-112487175 on Build GRCh38
rs930850809 Chr.9: 112487213-112487213 on Build GRCh38
rs536390881 Chr.9: 112487013-112487013 on Build GRCh38
rs574933155 Chr.9: 112487291-112487291 on Build GRCh38
rs557123302 Chr.9: 112486996-112486996 on Build GRCh38
rs950847641 Chr.9: 112487461-112487461 on Build GRCh38
rs943102878 Chr.9: 112487094-112487094 on Build GRCh38
rs962382283 Chr.9: 112487303-112487303 on Build GRCh38
rs901494063 Chr.9: 112487122-112487122 on Build GRCh38
rs564680078 Chr.9: 112487421-112487421 on Build GRCh38
rs762317958 Chr.9: 112487047-112487047 on Build GRCh38
rs56045355 Chr.9: 112487334-112487334 on Build GRCh38
rs886126213 Chr.9: 112487360-112487361 on Build GRCh38
rs940248080 Chr.9: 112487361-112487361 on Build GRCh38
rs551418476 Chr.9: 112487418-112487418 on Build GRCh38
rs757733579 Chr.9: 112487258-112487258 on Build GRCh38
rs769100439 Chr.9: 112486988-112486988 on Build GRCh38
rs1043643448 Chr.9: 112487388-112487388 on Build GRCh38
rs1006363005 Chr.9: 112487463-112487463 on Build GRCh38
rs992280412 Chr.9: 112487242-112487242 on Build GRCh38
rs1004425205 Chr.9: 112487154-112487154 on Build GRCh38
rs751830228 Chr.9: 112487181-112487181 on Build GRCh38
rs1051280303 Chr.9: 112487152-112487152 on Build GRCh38
rs6477940 Chr.9: 112486982-112486982 on Build GRCh38
rs950310105 Chr.9: 112487346-112487346 on Build GRCh38
rs1026439368 Chr.9: 112487273-112487273 on Build GRCh38
rs950918455 Chr.9: 112487278-112487278 on Build GRCh38
rs561567983 Chr.9: 112487279-112487279 on Build GRCh38
rs1050868978 Chr.9: 112487341-112487341 on Build GRCh38
rs758243573 Chr.9: 112487012-112487019 on Build GRCh38
rs553027026 Chr.9: 112487032-112487032 on Build GRCh38
rs1002818326 Chr.9: 112487403-112487403 on Build GRCh38
rs1016439834 Chr.9: 112487478-112487478 on Build GRCh38
rs374420957 Chr.9: 112487458-112487458 on Build GRCh38
rs992073405 Chr.9: 112486993-112486993 on Build GRCh38
rs558145333 Chr.9: 112487411-112487411 on Build GRCh38
rs925124181 Chr.9: 112487074-112487074 on Build GRCh38
rs765788212 Chr.9: 112487072-112487072 on Build GRCh38
rs528163179 Chr.9: 112487385-112487385 on Build GRCh38
rs542773987 Chr.9: 112487317-112487317 on Build GRCh38
rs898660627 Chr.9: 112487392-112487392 on Build GRCh38
rs974518310 Chr.9: 112487301-112487301 on Build GRCh38
rs1020967633 Chr.9: 112487254-112487254 on Build GRCh38
rs1034219056 Chr.9: 112487238-112487238 on Build GRCh38
rs746108473 Chr.9: 112487069-112487069 on Build GRCh38
rs992125515 Chr.9: 112487249-112487249 on Build GRCh38
rs7031191 Chr.9: 112487153-112487153 on Build GRCh38
rs966726945 Chr.9: 112487475-112487475 on Build GRCh38
rs781202169 Chr.9: 112487068-112487069 on Build GRCh38

View Full Product Details