Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113275242-113275779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC26
Entrez Gene ID
246184
Gene Name
cell division cycle 26
Gene Aliases
ANAPC12, APC12, C9orf17
UniGene
Hs.727648
Gene Chromosome Location
Chr.9: 113267008-113275589 on Build GRCh38
Transcripts
NM_139286.3 XM_017014573.1 XM_017014575.1 NM_001244926.1 NM_001322266.1 NM_001322267.1 NM_004697.4 NR_136265.1 NR_136266.1

Gene Symbol
PRPF4
Entrez Gene ID
9128
Gene Name
pre-mRNA processing factor 4
Gene Aliases
HPRP4, HPRP4P, PRP4, Prp4p, RP70, SNRNP60
UniGene
Hs.744014
Gene Chromosome Location
Chr.9: 113275634-113292905 on Build GRCh38
Transcripts
NM_139286.3 XM_017014573.1 XM_017014575.1 NM_001244926.1 NM_001322266.1 NM_001322267.1 NM_004697.4 NR_136265.1 NR_136266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6407747 c.26C>T p.T9M Chr.9: 113275769-113275769 on Build GRCh38
295351 c.13C>T p.R5* Chr.9: 113275756-113275756 on Build GRCh38
1239956 c.19T>A p.S7T Chr.9: 113275762-113275762 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs908262908 Chr.9: 113275587-113275587 on Build GRCh38
rs796462753 Chr.9: 113275582-113275582 on Build GRCh38
rs375406089 Chr.9: 113275693-113275693 on Build GRCh38
rs964743979 Chr.9: 113275416-113275416 on Build GRCh38
rs16931980 Chr.9: 113275689-113275689 on Build GRCh38
rs1048147319 Chr.9: 113275682-113275682 on Build GRCh38
rs573327598 Chr.9: 113275360-113275360 on Build GRCh38
rs575234795 Chr.9: 113275334-113275334 on Build GRCh38
rs758702618 Chr.9: 113275721-113275721 on Build GRCh38
rs937713220 Chr.9: 113275536-113275536 on Build GRCh38
rs764710245 Chr.9: 113275706-113275706 on Build GRCh38
rs954719024 Chr.9: 113275748-113275748 on Build GRCh38
rs531642746 Chr.9: 113275572-113275572 on Build GRCh38
rs574604249 Chr.9: 113275723-113275723 on Build GRCh38
rs757688074 Chr.9: 113275480-113275480 on Build GRCh38
rs947568504 Chr.9: 113275496-113275496 on Build GRCh38
rs375348528 Chr.9: 113275652-113275652 on Build GRCh38
rs558857722 Chr.9: 113275673-113275673 on Build GRCh38
rs371893889 Chr.9: 113275555-113275555 on Build GRCh38
rs1042429439 Chr.9: 113275535-113275535 on Build GRCh38
rs1001806473 Chr.9: 113275354-113275354 on Build GRCh38
rs903962842 Chr.9: 113275547-113275547 on Build GRCh38
rs896130269 Chr.9: 113275543-113275543 on Build GRCh38
rs150504332 Chr.9: 113275743-113275743 on Build GRCh38
rs755591237 Chr.9: 113275742-113275742 on Build GRCh38
rs867677453 Chr.9: 113275521-113275521 on Build GRCh38
rs1042212901 Chr.9: 113275638-113275638 on Build GRCh38
rs775989074 Chr.9: 113275697-113275697 on Build GRCh38
rs545752859 Chr.9: 113275539-113275539 on Build GRCh38
rs41276777 Chr.9: 113275412-113275412 on Build GRCh38
rs950550283 Chr.9: 113275399-113275399 on Build GRCh38
rs891925515 Chr.9: 113275306-113275306 on Build GRCh38
rs3810919 Chr.9: 113275614-113275614 on Build GRCh38
rs548132520 Chr.9: 113275592-113275592 on Build GRCh38
rs200111116 Chr.9: 113275751-113275751 on Build GRCh38
rs373384413 Chr.9: 113275428-113275428 on Build GRCh38
rs923232402 Chr.9: 113275425-113275425 on Build GRCh38
rs896399784 Chr.9: 113275365-113275365 on Build GRCh38
rs780236028 Chr.9: 113275630-113275630 on Build GRCh38
rs747618362 Chr.9: 113275737-113275737 on Build GRCh38
rs750585880 Chr.9: 113275720-113275720 on Build GRCh38
rs996276688 Chr.9: 113275714-113275714 on Build GRCh38
rs1035271899 Chr.9: 113275338-113275338 on Build GRCh38
rs962488585 Chr.9: 113275707-113275707 on Build GRCh38
rs950881444 Chr.9: 113275573-113275573 on Build GRCh38
rs985309350 Chr.9: 113275402-113275402 on Build GRCh38
rs983636673 Chr.9: 113275574-113275574 on Build GRCh38
rs903610985 Chr.9: 113275533-113275533 on Build GRCh38
rs886294586 Chr.9: 113275564-113275564 on Build GRCh38
rs1039028284 Chr.9: 113275667-113275667 on Build GRCh38
rs775658659 Chr.9: 113275756-113275756 on Build GRCh38
rs762010635 Chr.9: 113275715-113275715 on Build GRCh38
rs1017829515 Chr.9: 113275405-113275405 on Build GRCh38
rs558555669 Chr.9: 113275321-113275321 on Build GRCh38
rs879132037 Chr.9: 113275735-113275735 on Build GRCh38
rs1026583216 Chr.9: 113275392-113275392 on Build GRCh38
rs541873609 Chr.9: 113275630-113275647 on Build GRCh38
rs575561116 Chr.9: 113275674-113275674 on Build GRCh38
rs377499486 Chr.9: 113275672-113275672 on Build GRCh38
rs113213009 Chr.9: 113275681-113275681 on Build GRCh38
rs765445102 Chr.9: 113275719-113275719 on Build GRCh38
rs772304068 Chr.9: 113275695-113275695 on Build GRCh38
rs748886389 Chr.9: 113275747-113275747 on Build GRCh38
rs913468773 Chr.9: 113275642-113275642 on Build GRCh38
rs937930050 Chr.9: 113275619-113275619 on Build GRCh38
rs532859078 Chr.9: 113275423-113275423 on Build GRCh38
rs964796011 Chr.9: 113275599-113275599 on Build GRCh38
rs41276779 Chr.9: 113275427-113275427 on Build GRCh38
rs1030562035 Chr.9: 113275687-113275687 on Build GRCh38
rs114893079 Chr.9: 113275319-113275319 on Build GRCh38
rs886347703 Chr.9: 113275688-113275688 on Build GRCh38
rs1056293928 Chr.9: 113275481-113275481 on Build GRCh38
rs1043890336 Chr.9: 113275525-113275525 on Build GRCh38
rs937766910 Chr.9: 113275656-113275656 on Build GRCh38
rs377765365 Chr.9: 113275691-113275691 on Build GRCh38
rs371047661 Chr.9: 113275710-113275710 on Build GRCh38
rs12343330 Chr.9: 113275417-113275417 on Build GRCh38
rs550478749 Chr.9: 113275703-113275703 on Build GRCh38
rs954666966 Chr.9: 113275622-113275622 on Build GRCh38
rs376054253 Chr.9: 113275734-113275734 on Build GRCh38
rs553952252 Chr.9: 113275647-113275647 on Build GRCh38
rs1026217691 Chr.9: 113275559-113275559 on Build GRCh38
rs1001399155 Chr.9: 113275666-113275666 on Build GRCh38
rs903548477 Chr.9: 113275342-113275342 on Build GRCh38
rs544222413 Chr.9: 113275339-113275339 on Build GRCh38
rs1016792887 Chr.9: 113275585-113275585 on Build GRCh38
rs533995971 Chr.9: 113275623-113275623 on Build GRCh38
rs550706483 Chr.9: 113275692-113275692 on Build GRCh38
rs778416183 Chr.9: 113275609-113275609 on Build GRCh38
rs568140896 Chr.9: 113275669-113275669 on Build GRCh38
rs896353849 Chr.9: 113275495-113275495 on Build GRCh38
rs927433526 Chr.9: 113275654-113275654 on Build GRCh38
rs974784342 Chr.9: 113275601-113275601 on Build GRCh38
rs751801962 Chr.9: 113275424-113275424 on Build GRCh38
rs1006048122 Chr.9: 113275567-113275567 on Build GRCh38
rs754855066 Chr.9: 113275727-113275727 on Build GRCh38
rs531785428 Chr.9: 113275625-113275625 on Build GRCh38
rs973724645 Chr.9: 113275606-113275606 on Build GRCh38
rs1018026509 Chr.9: 113275550-113275550 on Build GRCh38
rs182309345 Chr.9: 113275369-113275369 on Build GRCh38
rs1000842150 Chr.9: 113275549-113275549 on Build GRCh38
rs377644636 Chr.9: 113275698-113275698 on Build GRCh38

View Full Product Details