Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113389520-113390024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALAD
Entrez Gene ID
210
Gene Name
aminolevulinate dehydratase
Gene Aliases
ALADH, PBGS
UniGene
Hs.1227
Gene Chromosome Location
Chr.9: 113386312-113401338 on Build GRCh38
Transcripts
NM_000031.5 NM_001003945.2 NM_001317745.1 XM_011518364.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1624447 c.763C>T p.L255L Chr.9: 113389637-113389637 on Build GRCh38
1624449 c.762G>T p.Q254H Chr.9: 113389638-113389638 on Build GRCh38
1674737 c.662G>A p.R221H Chr.9: 113389651-113389651 on Build GRCh38
4588364 c.606C>T p.S202S Chr.9: 113389793-113389793 on Build GRCh38
1624451 c.684A>G p.K228K Chr.9: 113389802-113389802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750137137 Chr.9: 113389751-113389751 on Build GRCh38
rs760621556 Chr.9: 113389720-113389720 on Build GRCh38
rs767827395 Chr.9: 113389754-113389754 on Build GRCh38
rs143028530 Chr.9: 113389651-113389652 on Build GRCh38
rs763960444 Chr.9: 113389733-113389733 on Build GRCh38
rs753037931 Chr.9: 113389823-113389823 on Build GRCh38
rs748174951 Chr.9: 113389567-113389567 on Build GRCh38
rs764647305 Chr.9: 113389827-113389827 on Build GRCh38
rs763700966 Chr.9: 113389607-113389607 on Build GRCh38
rs761237322 Chr.9: 113389738-113389738 on Build GRCh38
rs761010522 Chr.9: 113389824-113389824 on Build GRCh38
rs773271903 Chr.9: 113389568-113389568 on Build GRCh38
rs377694098 Chr.9: 113389581-113389581 on Build GRCh38
rs769331098 Chr.9: 113389800-113389800 on Build GRCh38
rs762540635 Chr.9: 113389810-113389810 on Build GRCh38
rs372240962 Chr.9: 113389836-113389836 on Build GRCh38
rs375646177 Chr.9: 113389778-113389778 on Build GRCh38
rs779850152 Chr.9: 113389775-113389775 on Build GRCh38
rs755424043 Chr.9: 113389549-113389549 on Build GRCh38
rs1000211567 Chr.9: 113389743-113389743 on Build GRCh38
rs775624486 Chr.9: 113389603-113389603 on Build GRCh38
rs781669395 Chr.9: 113389552-113389552 on Build GRCh38
rs201127458 Chr.9: 113389773-113389773 on Build GRCh38
rs759409230 Chr.9: 113389587-113389587 on Build GRCh38
rs891982366 Chr.9: 113389966-113389966 on Build GRCh38
rs368043911 Chr.9: 113389734-113389734 on Build GRCh38
rs751493955 Chr.9: 113389591-113389591 on Build GRCh38
rs8177809 Chr.9: 113389545-113389545 on Build GRCh38
rs1004413407 Chr.9: 113389914-113389914 on Build GRCh38
rs781646677 Chr.9: 113389645-113389645 on Build GRCh38
rs780147839 Chr.9: 113389874-113389874 on Build GRCh38
rs549420461 Chr.9: 113389991-113389991 on Build GRCh38
rs758636742 Chr.9: 113389871-113389871 on Build GRCh38
rs1049787340 Chr.9: 113389898-113389898 on Build GRCh38
rs1055687089 Chr.9: 113389650-113389650 on Build GRCh38
rs764868798 Chr.9: 113389745-113389745 on Build GRCh38
rs756582105 Chr.9: 113389755-113389755 on Build GRCh38
rs776503307 Chr.9: 113389792-113389792 on Build GRCh38
rs374269304 Chr.9: 113389774-113389774 on Build GRCh38
rs766029772 Chr.9: 113389812-113389812 on Build GRCh38
rs774229934 Chr.9: 113389586-113389586 on Build GRCh38
rs767455387 Chr.9: 113389750-113389751 on Build GRCh38
rs201822144 Chr.9: 113389862-113389862 on Build GRCh38
rs1007287379 Chr.9: 113389571-113389571 on Build GRCh38
rs1021864550 Chr.9: 113389993-113389993 on Build GRCh38
rs956235996 Chr.9: 113389648-113389648 on Build GRCh38
rs973089315 Chr.9: 113389622-113389622 on Build GRCh38
rs753231528 Chr.9: 113389842-113389842 on Build GRCh38
rs368684988 Chr.9: 113389797-113389797 on Build GRCh38
rs777855664 Chr.9: 113389652-113389652 on Build GRCh38
rs955527533 Chr.9: 113389562-113389562 on Build GRCh38
rs997277368 Chr.9: 113389956-113389956 on Build GRCh38
rs778146437 Chr.9: 113389758-113389758 on Build GRCh38
rs903298299 Chr.9: 113389742-113389742 on Build GRCh38
rs749341441 Chr.9: 113389663-113389663 on Build GRCh38
rs1016159156 Chr.9: 113389926-113389926 on Build GRCh38
rs748726105 Chr.9: 113389649-113389649 on Build GRCh38
rs753321173 Chr.9: 113389615-113389615 on Build GRCh38
rs887460352 Chr.9: 113389566-113389566 on Build GRCh38
rs753946930 Chr.9: 113389762-113389762 on Build GRCh38
rs529525799 Chr.9: 113389939-113389939 on Build GRCh38
rs920234618 Chr.9: 113389646-113389646 on Build GRCh38
rs774779771 Chr.9: 113389688-113389688 on Build GRCh38
rs772736028 Chr.9: 113389809-113389809 on Build GRCh38
rs201420740 Chr.9: 113389621-113389621 on Build GRCh38
rs779097677 Chr.9: 113389861-113389861 on Build GRCh38
rs1030249826 Chr.9: 113389561-113389561 on Build GRCh38
rs527407905 Chr.9: 113389594-113389594 on Build GRCh38
rs768577018 Chr.9: 113389785-113389785 on Build GRCh38
rs371066673 Chr.9: 113389637-113389637 on Build GRCh38
rs759860695 Chr.9: 113389752-113389752 on Build GRCh38
rs550232973 Chr.9: 113389719-113389719 on Build GRCh38
rs767556393 Chr.9: 113389601-113389601 on Build GRCh38
rs190497588 Chr.9: 113389635-113389635 on Build GRCh38
rs371809975 Chr.9: 113389764-113389764 on Build GRCh38
rs375689609 Chr.9: 113389669-113389669 on Build GRCh38
rs760083050 Chr.9: 113389606-113389606 on Build GRCh38
rs779253166 Chr.9: 113389767-113389767 on Build GRCh38
rs1805258 Chr.9: 113389717-113389717 on Build GRCh38
rs771075750 Chr.9: 113389584-113389584 on Build GRCh38
rs745642261 Chr.9: 113389709-113389709 on Build GRCh38
rs757242659 Chr.9: 113389735-113389736 on Build GRCh38
rs970062215 Chr.9: 113389701-113389701 on Build GRCh38
rs1031810001 Chr.9: 113389959-113389959 on Build GRCh38
rs756607034 Chr.9: 113389651-113389651 on Build GRCh38

View Full Product Details