Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113391185-113391719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALAD
Entrez Gene ID
210
Gene Name
aminolevulinate dehydratase
Gene Aliases
ALADH, PBGS
UniGene
Hs.1227
Gene Chromosome Location
Chr.9: 113386312-113401338 on Build GRCh38
Transcripts
NM_000031.5 NM_001003945.2 NM_001317745.1 XM_011518364.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660485 c.190A>G p.M64V Chr.9: 113391598-113391598 on Build GRCh38
4163234 c.168T>C p.Y56Y Chr.9: 113391620-113391620 on Build GRCh38
1701543 c.244C>T p.P82S Chr.9: 113391544-113391544 on Build GRCh38
3996247 c.276G>A p.E92E Chr.9: 113391599-113391599 on Build GRCh38
4885153 c.288G>T p.K96N Chr.9: 113391527-113391527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759495380 Chr.9: 113391647-113391647 on Build GRCh38
rs933972600 Chr.9: 113391617-113391617 on Build GRCh38
rs776689746 Chr.9: 113391514-113391514 on Build GRCh38
rs747557477 Chr.9: 113391595-113391595 on Build GRCh38
rs545672911 Chr.9: 113391508-113391508 on Build GRCh38
rs890313016 Chr.9: 113391220-113391220 on Build GRCh38
rs924104717 Chr.9: 113391444-113391444 on Build GRCh38
rs934068162 Chr.9: 113391401-113391401 on Build GRCh38
rs372484944 Chr.9: 113391484-113391484 on Build GRCh38
rs764993649 Chr.9: 113391264-113391264 on Build GRCh38
rs760170818 Chr.9: 113391672-113391672 on Build GRCh38
rs1052732171 Chr.9: 113391229-113391229 on Build GRCh38
rs571116408 Chr.9: 113391266-113391266 on Build GRCh38
rs971399312 Chr.9: 113391643-113391643 on Build GRCh38
rs759085292 Chr.9: 113391548-113391548 on Build GRCh38
rs763659395 Chr.9: 113391568-113391568 on Build GRCh38
rs748364562 Chr.9: 113391673-113391673 on Build GRCh38
rs919925984 Chr.9: 113391368-113391368 on Build GRCh38
rs200963049 Chr.9: 113391547-113391547 on Build GRCh38
rs368315304 Chr.9: 113391481-113391481 on Build GRCh38
rs200280568 Chr.9: 113391609-113391609 on Build GRCh38
rs1800435 Chr.9: 113391611-113391611 on Build GRCh38
rs968659370 Chr.9: 113391231-113391231 on Build GRCh38
rs199501308 Chr.9: 113391329-113391329 on Build GRCh38
rs111915485 Chr.9: 113391500-113391500 on Build GRCh38
rs999864980 Chr.9: 113391256-113391256 on Build GRCh38
rs774997435 Chr.9: 113391661-113391661 on Build GRCh38
rs372911804 Chr.9: 113391262-113391262 on Build GRCh38
rs73538845 Chr.9: 113391221-113391221 on Build GRCh38
rs537189228 Chr.9: 113391269-113391269 on Build GRCh38
rs548784490 Chr.9: 113391680-113391680 on Build GRCh38
rs934419023 Chr.9: 113391449-113391449 on Build GRCh38
rs755751594 Chr.9: 113391479-113391479 on Build GRCh38
rs8177802 Chr.9: 113391288-113391288 on Build GRCh38
rs892770510 Chr.9: 113391239-113391239 on Build GRCh38
rs1041614321 Chr.9: 113391454-113391454 on Build GRCh38
rs780517883 Chr.9: 113391575-113391575 on Build GRCh38
rs780383993 Chr.9: 113391524-113391524 on Build GRCh38
rs8177801 Chr.9: 113391406-113391406 on Build GRCh38
rs565649579 Chr.9: 113391214-113391214 on Build GRCh38
rs771915829 Chr.9: 113391660-113391660 on Build GRCh38
rs992999506 Chr.9: 113391265-113391265 on Build GRCh38
rs1010066311 Chr.9: 113391445-113391445 on Build GRCh38
rs747080421 Chr.9: 113391490-113391490 on Build GRCh38
rs892583430 Chr.9: 113391439-113391439 on Build GRCh38
rs770886658 Chr.9: 113391637-113391637 on Build GRCh38
rs758374172 Chr.9: 113391649-113391652 on Build GRCh38
rs530804739 Chr.9: 113391571-113391571 on Build GRCh38
rs1139488 Chr.9: 113391620-113391620 on Build GRCh38
rs149572240 Chr.9: 113391537-113391537 on Build GRCh38
rs1019427903 Chr.9: 113391294-113391294 on Build GRCh38
rs8177803 Chr.9: 113391263-113391263 on Build GRCh38
rs1052764681 Chr.9: 113391457-113391457 on Build GRCh38
rs752170688 Chr.9: 113391559-113391559 on Build GRCh38
rs575419014 Chr.9: 113391353-113391353 on Build GRCh38
rs1020200706 Chr.9: 113391227-113391227 on Build GRCh38
rs952726962 Chr.9: 113391376-113391376 on Build GRCh38
rs749066913 Chr.9: 113391634-113391634 on Build GRCh38
rs935908863 Chr.9: 113391681-113391681 on Build GRCh38
rs530096718 Chr.9: 113391633-113391633 on Build GRCh38
rs574313359 Chr.9: 113391330-113391330 on Build GRCh38
rs774317722 Chr.9: 113391646-113391646 on Build GRCh38
rs767011177 Chr.9: 113391551-113391551 on Build GRCh38
rs755472728 Chr.9: 113391603-113391603 on Build GRCh38
rs750761222 Chr.9: 113391386-113391386 on Build GRCh38
rs962958665 Chr.9: 113391594-113391594 on Build GRCh38
rs910127676 Chr.9: 113391322-113391322 on Build GRCh38
rs780605594 Chr.9: 113391489-113391489 on Build GRCh38
rs961781407 Chr.9: 113391317-113391317 on Build GRCh38
rs368846456 Chr.9: 113391567-113391567 on Build GRCh38
rs990809569 Chr.9: 113391377-113391377 on Build GRCh38
rs748364087 Chr.9: 113391520-113391520 on Build GRCh38
rs371476277 Chr.9: 113391569-113391569 on Build GRCh38
rs769936286 Chr.9: 113391521-113391521 on Build GRCh38
rs746243171 Chr.9: 113391483-113391483 on Build GRCh38

View Full Product Details