Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121567029-121567528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB2IP
Entrez Gene ID
153090
Gene Name
DAB2 interacting protein
Gene Aliases
AF9Q34, AIP-1, AIP1, DIP1/2
UniGene
Hs.522378
Gene Chromosome Location
Chr.9: 121566883-121785530 on Build GRCh38
Transcripts
NM_032552.3 XM_005251721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245314 c.40+7C>T p.? Chr.9: 121567235-121567235 on Build GRCh38
4636789 c.30C>T p.D10D Chr.9: 121567218-121567218 on Build GRCh38
7418434 c.35C>A p.S12Y Chr.9: 121567223-121567223 on Build GRCh38
1292781 c.39C>T p.Y13Y Chr.9: 121567227-121567227 on Build GRCh38
5410304 c.15C>T p.S5S Chr.9: 121567203-121567203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547556932 Chr.9: 121567253-121567253 on Build GRCh38
rs35827615 Chr.9: 121567409-121567410 on Build GRCh38
rs953347463 Chr.9: 121567130-121567130 on Build GRCh38
rs763833463 Chr.9: 121567476-121567476 on Build GRCh38
rs577832190 Chr.9: 121567459-121567459 on Build GRCh38
rs1051724010 Chr.9: 121567080-121567080 on Build GRCh38
rs957528667 Chr.9: 121567091-121567091 on Build GRCh38
rs200496124 Chr.9: 121567198-121567198 on Build GRCh38
rs971858268 Chr.9: 121567159-121567159 on Build GRCh38
rs537953259 Chr.9: 121567419-121567419 on Build GRCh38
rs779342806 Chr.9: 121567145-121567145 on Build GRCh38
rs369955581 Chr.9: 121567175-121567175 on Build GRCh38
rs765763166 Chr.9: 121567179-121567179 on Build GRCh38
rs148409496 Chr.9: 121567171-121567171 on Build GRCh38
rs768595008 Chr.9: 121567169-121567169 on Build GRCh38
rs759183425 Chr.9: 121567265-121567265 on Build GRCh38
rs567283350 Chr.9: 121567418-121567418 on Build GRCh38
rs538896763 Chr.9: 121567496-121567496 on Build GRCh38
rs35128663 Chr.9: 121567126-121567127 on Build GRCh38
rs1045107716 Chr.9: 121567061-121567061 on Build GRCh38
rs779450388 Chr.9: 121567187-121567187 on Build GRCh38
rs779359789 Chr.9: 121567197-121567197 on Build GRCh38
rs560588941 Chr.9: 121567114-121567114 on Build GRCh38
rs572607870 Chr.9: 121567302-121567302 on Build GRCh38
rs904980003 Chr.9: 121567079-121567079 on Build GRCh38
rs775075469 Chr.9: 121567380-121567380 on Build GRCh38
rs1000609041 Chr.9: 121567087-121567087 on Build GRCh38
rs1043564075 Chr.9: 121567376-121567376 on Build GRCh38
rs757734023 Chr.9: 121567195-121567195 on Build GRCh38
rs773087793 Chr.9: 121567248-121567248 on Build GRCh38
rs976130964 Chr.9: 121567110-121567110 on Build GRCh38
rs898053159 Chr.9: 121567059-121567059 on Build GRCh38
rs966680617 Chr.9: 121567107-121567107 on Build GRCh38
rs1032061041 Chr.9: 121567088-121567088 on Build GRCh38
rs1020546159 Chr.9: 121567095-121567095 on Build GRCh38
rs376712260 Chr.9: 121567465-121567465 on Build GRCh38
rs769527237 Chr.9: 121567235-121567235 on Build GRCh38
rs142520673 Chr.9: 121567185-121567185 on Build GRCh38
rs968981559 Chr.9: 121567127-121567127 on Build GRCh38
rs201252302 Chr.9: 121567227-121567227 on Build GRCh38
rs949212451 Chr.9: 121567494-121567494 on Build GRCh38
rs189328616 Chr.9: 121567259-121567259 on Build GRCh38
rs984845100 Chr.9: 121567131-121567131 on Build GRCh38
rs536546763 Chr.9: 121567276-121567276 on Build GRCh38
rs549151363 Chr.9: 121567379-121567379 on Build GRCh38
rs909279139 Chr.9: 121567142-121567142 on Build GRCh38
rs745399903 Chr.9: 121567295-121567295 on Build GRCh38
rs998665006 Chr.9: 121567411-121567411 on Build GRCh38
rs369466458 Chr.9: 121567147-121567147 on Build GRCh38
rs953400027 Chr.9: 121567426-121567426 on Build GRCh38
rs771262362 Chr.9: 121567144-121567144 on Build GRCh38
rs373859727 Chr.9: 121567443-121567443 on Build GRCh38
rs186002930 Chr.9: 121567126-121567126 on Build GRCh38
rs984728322 Chr.9: 121567429-121567429 on Build GRCh38
rs752719524 Chr.9: 121567332-121567332 on Build GRCh38
rs772343538 Chr.9: 121567202-121567202 on Build GRCh38
rs372088944 Chr.9: 121567150-121567150 on Build GRCh38
rs749679985 Chr.9: 121567140-121567140 on Build GRCh38
rs917734260 Chr.9: 121567490-121567490 on Build GRCh38
rs778480636 Chr.9: 121567194-121567194 on Build GRCh38
rs751020175 Chr.9: 121567262-121567262 on Build GRCh38
rs199630836 Chr.9: 121567219-121567219 on Build GRCh38
rs747154084 Chr.9: 121567218-121567218 on Build GRCh38
rs962085257 Chr.9: 121567464-121567464 on Build GRCh38
rs746858434 Chr.9: 121567317-121567317 on Build GRCh38
rs1010382787 Chr.9: 121567094-121567094 on Build GRCh38
rs749835580 Chr.9: 121567089-121567089 on Build GRCh38
rs756630816 Chr.9: 121567187-121567187 on Build GRCh38
rs1010436511 Chr.9: 121567298-121567298 on Build GRCh38
rs776397734 Chr.9: 121567170-121567170 on Build GRCh38
rs771999384 Chr.9: 121567146-121567146 on Build GRCh38
rs750089842 Chr.9: 121567178-121567178 on Build GRCh38
rs936282742 Chr.9: 121567203-121567203 on Build GRCh38
rs35115829 Chr.9: 121567190-121567191 on Build GRCh38

View Full Product Details