Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 123379570-123380071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRB2
Entrez Gene ID
286204
Gene Name
crumbs 2, cell polarity complex component
Gene Aliases
FSGS9, VMCKD
UniGene
Hs.568340
Gene Chromosome Location
Chr.9: 123354065-123380335 on Build GRCh38
Transcripts
NR_104603.1 NM_020946.1 XM_005252109.2 XM_005252111.4 XM_011518882.2 XM_011518883.2 XM_011518884.2 XM_011518885.2 XM_011518886.2 XM_017014947.1 XM_017014949.1 XM_017014950.1 XM_017014952.1

Gene Symbol
DENND1A
Entrez Gene ID
57706
Gene Name
DENN domain containing 1A
Gene Aliases
FAM31A, KIAA1608
UniGene
Hs.744995
Gene Chromosome Location
Chr.9: 123379654-123930158 on Build GRCh38
Transcripts
NR_104603.1 NM_020946.1 XM_005252109.2 XM_005252111.4 XM_011518882.2 XM_011518883.2 XM_011518884.2 XM_011518885.2 XM_011518886.2 XM_017014947.1 XM_017014949.1 XM_017014950.1 XM_017014952.1

SNP Detail

SNP ID Chromosome Location
rs528142254 Chr.9: 123379868-123379868 on Build GRCh38
rs951525091 Chr.9: 123379600-123379600 on Build GRCh38
rs925716385 Chr.9: 123379930-123379930 on Build GRCh38
rs562988355 Chr.9: 123379765-123379765 on Build GRCh38
rs982893588 Chr.9: 123379604-123379604 on Build GRCh38
rs867841934 Chr.9: 123379833-123379833 on Build GRCh38
rs1015208167 Chr.9: 123379859-123379859 on Build GRCh38
rs530373026 Chr.9: 123379925-123379925 on Build GRCh38
rs745472441 Chr.9: 123379694-123379694 on Build GRCh38
rs1008212304 Chr.9: 123379620-123379620 on Build GRCh38
rs1052492750 Chr.9: 123379605-123379605 on Build GRCh38
rs569533660 Chr.9: 123379928-123379929 on Build GRCh38
rs925507482 Chr.9: 123379901-123379901 on Build GRCh38
rs1052629133 Chr.9: 123379804-123379804 on Build GRCh38
rs1055942595 Chr.9: 123379806-123379806 on Build GRCh38
rs1005990023 Chr.9: 123379856-123379856 on Build GRCh38
rs75408012 Chr.9: 123380022-123380022 on Build GRCh38
rs1045320952 Chr.9: 123379758-123379758 on Build GRCh38
rs557731039 Chr.9: 123379975-123379976 on Build GRCh38
rs962668231 Chr.9: 123379856-123379863 on Build GRCh38
rs537289395 Chr.9: 123379822-123379822 on Build GRCh38
rs947750577 Chr.9: 123379754-123379754 on Build GRCh38
rs760888887 Chr.9: 123379657-123379658 on Build GRCh38
rs192384319 Chr.9: 123379794-123379794 on Build GRCh38
rs146869671 Chr.9: 123379821-123379821 on Build GRCh38
rs758565237 Chr.9: 123379910-123379910 on Build GRCh38
rs962124551 Chr.9: 123379730-123379730 on Build GRCh38
rs1006125824 Chr.9: 123380008-123380008 on Build GRCh38
rs988319499 Chr.9: 123379946-123379946 on Build GRCh38
rs542107100 Chr.9: 123379685-123379685 on Build GRCh38
rs1004297727 Chr.9: 123379823-123379823 on Build GRCh38
rs914023469 Chr.9: 123379952-123379952 on Build GRCh38
rs907602490 Chr.9: 123379691-123379691 on Build GRCh38
rs936947752 Chr.9: 123379943-123379943 on Build GRCh38
rs951991535 Chr.9: 123379851-123379851 on Build GRCh38
rs892548658 Chr.9: 123379805-123379805 on Build GRCh38
rs899926124 Chr.9: 123379958-123379958 on Build GRCh38
rs59314301 Chr.9: 123379953-123379953 on Build GRCh38
rs951856866 Chr.9: 123379680-123379680 on Build GRCh38
rs996015183 Chr.9: 123379643-123379643 on Build GRCh38
rs917606544 Chr.9: 123379752-123379752 on Build GRCh38
rs961811103 Chr.9: 123379863-123379863 on Build GRCh38
rs1048616 Chr.9: 123379954-123379954 on Build GRCh38
rs779358074 Chr.9: 123379647-123379647 on Build GRCh38
rs116259187 Chr.9: 123379893-123379893 on Build GRCh38
rs1041488823 Chr.9: 123379803-123379803 on Build GRCh38
rs550561376 Chr.9: 123379846-123379846 on Build GRCh38
rs1008763034 Chr.9: 123379814-123379814 on Build GRCh38
rs571348554 Chr.9: 123379599-123379599 on Build GRCh38
rs965982009 Chr.9: 123379642-123379642 on Build GRCh38
rs932640675 Chr.9: 123379978-123379978 on Build GRCh38
rs1024747754 Chr.9: 123379877-123379877 on Build GRCh38
rs771101517 Chr.9: 123380025-123380025 on Build GRCh38
rs973276157 Chr.9: 123379671-123379671 on Build GRCh38
rs535842804 Chr.9: 123379988-123379988 on Build GRCh38
rs777691335 Chr.9: 123379948-123379948 on Build GRCh38
rs563862326 Chr.9: 123379847-123379847 on Build GRCh38
rs757507019 Chr.9: 123379606-123379606 on Build GRCh38
rs1052015238 Chr.9: 123379757-123379757 on Build GRCh38
rs991833007 Chr.9: 123379745-123379745 on Build GRCh38
rs115901487 Chr.9: 123379892-123379892 on Build GRCh38
rs1028349805 Chr.9: 123379860-123379860 on Build GRCh38
rs987440239 Chr.9: 123379883-123379883 on Build GRCh38
rs1048404035 Chr.9: 123379980-123379980 on Build GRCh38
rs903986188 Chr.9: 123379594-123379594 on Build GRCh38
rs1020072285 Chr.9: 123379634-123379634 on Build GRCh38
rs894735547 Chr.9: 123379809-123379809 on Build GRCh38

View Full Product Details