Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 123381131-123381630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1A
Entrez Gene ID
57706
Gene Name
DENN domain containing 1A
Gene Aliases
FAM31A, KIAA1608
UniGene
Hs.744995
Gene Chromosome Location
Chr.9: 123379654-123930158 on Build GRCh38
Transcripts
NM_020946.1 XM_005252109.2 XM_005252111.4 XM_006717195.3 XM_011518882.2 XM_011518883.2 XM_011518884.2 XM_011518885.2 XM_011518886.2 XM_017014947.1 XM_017014948.1 XM_017014949.1 XM_017014950.1 XM_017014951.1 XM_017014952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3788168 c.2960C>T p.P987L Chr.9: 123381502-123381502 on Build GRCh38
3779617 c.2937G>A p.Q979Q Chr.9: 123381525-123381525 on Build GRCh38
6439709 c.2983G>A p.D995N Chr.9: 123381512-123381512 on Build GRCh38
6114694 c.2932C>A p.P978T Chr.9: 123381563-123381563 on Build GRCh38
487025 c.2984C>A p.P995Q Chr.9: 123381478-123381478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764890114 Chr.9: 123381429-123381429 on Build GRCh38
rs757571699 Chr.9: 123381507-123381507 on Build GRCh38
rs965582229 Chr.9: 123381215-123381215 on Build GRCh38
rs779124894 Chr.9: 123381509-123381509 on Build GRCh38
rs916665967 Chr.9: 123381279-123381279 on Build GRCh38
rs759513118 Chr.9: 123381399-123381399 on Build GRCh38
rs552899156 Chr.9: 123381194-123381194 on Build GRCh38
rs568708778 Chr.9: 123381305-123381305 on Build GRCh38
rs781687359 Chr.9: 123381478-123381479 on Build GRCh38
rs776813772 Chr.9: 123381181-123381181 on Build GRCh38
rs769654098 Chr.9: 123381549-123381549 on Build GRCh38
rs768161570 Chr.9: 123381437-123381437 on Build GRCh38
rs754315383 Chr.9: 123381602-123381602 on Build GRCh38
rs772031561 Chr.9: 123381403-123381403 on Build GRCh38
rs376350165 Chr.9: 123381472-123381472 on Build GRCh38
rs763730891 Chr.9: 123381412-123381412 on Build GRCh38
rs764535042 Chr.9: 123381598-123381598 on Build GRCh38
rs757446451 Chr.9: 123381453-123381453 on Build GRCh38
rs957363658 Chr.9: 123381158-123381158 on Build GRCh38
rs747017882 Chr.9: 123381527-123381527 on Build GRCh38
rs922375671 Chr.9: 123381231-123381231 on Build GRCh38
rs756502251 Chr.9: 123381446-123381446 on Build GRCh38
rs751587146 Chr.9: 123381477-123381478 on Build GRCh38
rs766859439 Chr.9: 123381543-123381543 on Build GRCh38
rs977945815 Chr.9: 123381363-123381363 on Build GRCh38
rs397841768 Chr.9: 123381155-123381156 on Build GRCh38
rs747011867 Chr.9: 123381345-123381346 on Build GRCh38
rs567413301 Chr.9: 123381346-123381346 on Build GRCh38
rs533899152 Chr.9: 123381319-123381319 on Build GRCh38
rs150591766 Chr.9: 123381551-123381551 on Build GRCh38
rs775361470 Chr.9: 123381404-123381404 on Build GRCh38
rs58894130 Chr.9: 123381197-123381197 on Build GRCh38
rs1001583043 Chr.9: 123381328-123381328 on Build GRCh38
rs550366198 Chr.9: 123381301-123381301 on Build GRCh38
rs753309845 Chr.9: 123381499-123381499 on Build GRCh38
rs995251044 Chr.9: 123381175-123381175 on Build GRCh38
rs756438059 Chr.9: 123381485-123381490 on Build GRCh38
rs1049478580 Chr.9: 123381240-123381240 on Build GRCh38
rs372919542 Chr.9: 123381220-123381220 on Build GRCh38
rs555028534 Chr.9: 123381392-123381392 on Build GRCh38
rs557614403 Chr.9: 123381345-123381345 on Build GRCh38
rs773709278 Chr.9: 123381563-123381563 on Build GRCh38
rs746084954 Chr.9: 123381501-123381501 on Build GRCh38
rs761281711 Chr.9: 123381588-123381588 on Build GRCh38
rs201751108 Chr.9: 123381156-123381156 on Build GRCh38
rs546651080 Chr.9: 123381203-123381203 on Build GRCh38
rs568557993 Chr.9: 123381267-123381267 on Build GRCh38
rs369485557 Chr.9: 123381438-123381438 on Build GRCh38
rs201942010 Chr.9: 123381491-123381491 on Build GRCh38
rs77969061 Chr.9: 123381195-123381195 on Build GRCh38
rs749579939 Chr.9: 123381452-123381452 on Build GRCh38
rs3739837 Chr.9: 123381383-123381383 on Build GRCh38
rs774398495 Chr.9: 123381397-123381397 on Build GRCh38
rs761360081 Chr.9: 123381428-123381428 on Build GRCh38
rs779985177 Chr.9: 123381525-123381525 on Build GRCh38
rs941049676 Chr.9: 123381277-123381277 on Build GRCh38
rs1048113736 Chr.9: 123381351-123381351 on Build GRCh38
rs757940275 Chr.9: 123381434-123381434 on Build GRCh38
rs955211391 Chr.9: 123381377-123381377 on Build GRCh38
rs764268652 Chr.9: 123381475-123381476 on Build GRCh38
rs994327763 Chr.9: 123381312-123381312 on Build GRCh38
rs1034434557 Chr.9: 123381333-123381333 on Build GRCh38
rs922396489 Chr.9: 123381371-123381371 on Build GRCh38
rs897429959 Chr.9: 123381292-123381292 on Build GRCh38
rs930254388 Chr.9: 123381430-123381430 on Build GRCh38
rs142598714 Chr.9: 123381471-123381471 on Build GRCh38
rs777992283 Chr.9: 123381451-123381451 on Build GRCh38
rs185466304 Chr.9: 123381359-123381359 on Build GRCh38
rs377148532 Chr.9: 123381388-123381388 on Build GRCh38
rs748551876 Chr.9: 123381389-123381389 on Build GRCh38
rs535538892 Chr.9: 123381271-123381271 on Build GRCh38
rs967069642 Chr.9: 123381362-123381362 on Build GRCh38
rs746917881 Chr.9: 123381479-123381479 on Build GRCh38
rs758753385 Chr.9: 123381457-123381461 on Build GRCh38
rs933779631 Chr.9: 123381236-123381236 on Build GRCh38
rs955180567 Chr.9: 123381571-123381571 on Build GRCh38
rs373157481 Chr.9: 123381580-123381580 on Build GRCh38
rs753594548 Chr.9: 123381423-123381423 on Build GRCh38
rs1055858477 Chr.9: 123381488-123381488 on Build GRCh38
rs918382106 Chr.9: 123381346-123381347 on Build GRCh38
rs547365850 Chr.9: 123381483-123381483 on Build GRCh38
rs1046360902 Chr.9: 123381317-123381317 on Build GRCh38
rs780444178 Chr.9: 123381560-123381562 on Build GRCh38
rs764565454 Chr.9: 123381502-123381502 on Build GRCh38
rs758541150 Chr.9: 123381510-123381510 on Build GRCh38
rs371138017 Chr.9: 123381503-123381503 on Build GRCh38
rs765919260 Chr.9: 123381496-123381496 on Build GRCh38
rs1048620635 Chr.9: 123381180-123381180 on Build GRCh38
rs761567995 Chr.9: 123381490-123381490 on Build GRCh38
rs776590199 Chr.9: 123381528-123381528 on Build GRCh38
rs958202446 Chr.9: 123381243-123381243 on Build GRCh38
rs550772209 Chr.9: 123381322-123381322 on Build GRCh38
rs957332603 Chr.9: 123381341-123381341 on Build GRCh38
rs984964821 Chr.9: 123381365-123381365 on Build GRCh38
rs745972611 Chr.9: 123381454-123381454 on Build GRCh38
rs749400308 Chr.9: 123381394-123381394 on Build GRCh38
rs773002960 Chr.9: 123381552-123381552 on Build GRCh38
rs749921070 Chr.9: 123381431-123381431 on Build GRCh38
rs199678734 Chr.9: 123381495-123381495 on Build GRCh38
rs754188975 Chr.9: 123381506-123381506 on Build GRCh38
rs746390585 Chr.9: 123381484-123381485 on Build GRCh38
rs766040661 Chr.9: 123381558-123381558 on Build GRCh38
rs771106767 Chr.9: 123381396-123381396 on Build GRCh38
rs369785708 Chr.9: 123381406-123381406 on Build GRCh38
rs776270301 Chr.9: 123381487-123381487 on Build GRCh38
rs1038590495 Chr.9: 123381282-123381282 on Build GRCh38
rs747915223 Chr.9: 123381546-123381546 on Build GRCh38

View Full Product Details