Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14843838-14844363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs1025612305 Chr.X: 14844242-14844242 on Build GRCh38
rs749699443 Chr.X: 14843895-14843895 on Build GRCh38
rs1014242798 Chr.X: 14844301-14844301 on Build GRCh38
rs780431161 Chr.X: 14843993-14843996 on Build GRCh38
rs777566944 Chr.X: 14843971-14843971 on Build GRCh38
rs757610874 Chr.X: 14843903-14843903 on Build GRCh38
rs776118523 Chr.X: 14843960-14843960 on Build GRCh38
rs969600273 Chr.X: 14844306-14844306 on Build GRCh38
rs1000153164 Chr.X: 14844165-14844165 on Build GRCh38
rs768405501 Chr.X: 14843952-14843952 on Build GRCh38
rs1025469154 Chr.X: 14844290-14844290 on Build GRCh38
rs201070097 Chr.X: 14843918-14843918 on Build GRCh38
rs777812135 Chr.X: 14843882-14843882 on Build GRCh38
rs761783161 Chr.X: 14844186-14844186 on Build GRCh38
rs6527026 Chr.X: 14844157-14844157 on Build GRCh38
rs775265652 Chr.X: 14844228-14844228 on Build GRCh38
rs200226322 Chr.X: 14844202-14844203 on Build GRCh38
rs376894598 Chr.X: 14844119-14844119 on Build GRCh38
rs945837663 Chr.X: 14844111-14844111 on Build GRCh38
rs762993002 Chr.X: 14843988-14843988 on Build GRCh38
rs750866138 Chr.X: 14844269-14844269 on Build GRCh38
rs756112688 Chr.X: 14844022-14844022 on Build GRCh38
rs1025182355 Chr.X: 14843867-14843867 on Build GRCh38
rs6527027 Chr.X: 14844167-14844167 on Build GRCh38
rs762073276 Chr.X: 14844240-14844240 on Build GRCh38
rs770504330 Chr.X: 14844107-14844107 on Build GRCh38
rs981068174 Chr.X: 14843949-14843949 on Build GRCh38
rs868775000 Chr.X: 14844121-14844121 on Build GRCh38
rs866120454 Chr.X: 14844231-14844231 on Build GRCh38
rs140404015 Chr.X: 14844045-14844045 on Build GRCh38
rs752657026 Chr.X: 14843871-14843871 on Build GRCh38
rs756392097 Chr.X: 14843880-14843880 on Build GRCh38
rs766381118 Chr.X: 14843996-14843996 on Build GRCh38
rs749762842 Chr.X: 14843997-14843997 on Build GRCh38
rs1039643303 Chr.X: 14844196-14844196 on Build GRCh38
rs761121974 Chr.X: 14843995-14843995 on Build GRCh38
rs989738529 Chr.X: 14844042-14844042 on Build GRCh38
rs748874800 Chr.X: 14844110-14844111 on Build GRCh38
rs1040790411 Chr.X: 14844034-14844034 on Build GRCh38
rs752847575 Chr.X: 14844015-14844015 on Build GRCh38
rs898388390 Chr.X: 14844197-14844197 on Build GRCh38
rs767996684 Chr.X: 14844243-14844243 on Build GRCh38
rs765112491 Chr.X: 14843974-14843974 on Build GRCh38
rs904350828 Chr.X: 14844164-14844164 on Build GRCh38
rs769165151 Chr.X: 14844027-14844031 on Build GRCh38
rs771958941 Chr.X: 14843925-14843925 on Build GRCh38
rs370135077 Chr.X: 14844142-14844142 on Build GRCh38
rs747865842 Chr.X: 14843919-14843919 on Build GRCh38
rs779449906 Chr.X: 14843909-14843909 on Build GRCh38
rs746453967 Chr.X: 14843929-14843929 on Build GRCh38
rs922325582 Chr.X: 14844073-14844073 on Build GRCh38
rs371428199 Chr.X: 14843976-14843976 on Build GRCh38

View Full Product Details