Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15845301-15845802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1S2
Entrez Gene ID
8905
Gene Name
adaptor related protein complex 1 sigma 2 subunit
Gene Aliases
DC22, MRX59, MRXS21, MRXS5, MRXSF, PGS, SIGMA1B
UniGene
Hs.121592
Gene Chromosome Location
Chr.X: 15825806-15855014 on Build GRCh38
Transcripts
NM_001272071.1 NM_003916.4 XM_005274614.3 XM_011545599.1 XM_017029925.1 XM_017029926.1

SNP Detail

SNP ID Chromosome Location
rs746801765 Chr.X: 15845525-15845526 on Build GRCh38
rs34951696 Chr.X: 15845621-15845622 on Build GRCh38
rs769132550 Chr.X: 15845418-15845418 on Build GRCh38
rs1025799236 Chr.X: 15845618-15845618 on Build GRCh38
rs980729031 Chr.X: 15845736-15845736 on Build GRCh38
rs770804791 Chr.X: 15845536-15845537 on Build GRCh38
rs587777542 Chr.X: 15845378-15845378 on Build GRCh38
rs201372069 Chr.X: 15845547-15845547 on Build GRCh38
rs777218950 Chr.X: 15845433-15845433 on Build GRCh38
rs142340628 Chr.X: 15845710-15845710 on Build GRCh38
rs969636853 Chr.X: 15845726-15845726 on Build GRCh38
rs772683061 Chr.X: 15845526-15845526 on Build GRCh38
rs192291108 Chr.X: 15845415-15845415 on Build GRCh38
rs770199415 Chr.X: 15845524-15845524 on Build GRCh38
rs762355079 Chr.X: 15845388-15845388 on Build GRCh38
rs900361893 Chr.X: 15845613-15845613 on Build GRCh38
rs779160574 Chr.X: 15845527-15845527 on Build GRCh38
rs755358393 Chr.X: 15845514-15845514 on Build GRCh38
rs1006480772 Chr.X: 15845389-15845389 on Build GRCh38
rs951657809 Chr.X: 15845629-15845629 on Build GRCh38
rs766200992 Chr.X: 15845529-15845529 on Build GRCh38
rs747059594 Chr.X: 15845763-15845763 on Build GRCh38
rs780107799 Chr.X: 15845346-15845346 on Build GRCh38
rs772453110 Chr.X: 15845701-15845701 on Build GRCh38
rs1013780323 Chr.X: 15845665-15845665 on Build GRCh38
rs773966134 Chr.X: 15845542-15845542 on Build GRCh38
rs746104268 Chr.X: 15845328-15845328 on Build GRCh38
rs758681457 Chr.X: 15845345-15845345 on Build GRCh38
rs766119174 Chr.X: 15845612-15845612 on Build GRCh38
rs777774281 Chr.X: 15845526-15845526 on Build GRCh38

View Full Product Details