Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23868356-23868898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOO
Entrez Gene ID
79135
Gene Name
apolipoprotein O
Gene Aliases
FAM121B, MIC26, Mic23, My025
UniGene
Hs.495851
Gene Chromosome Location
Chr.X: 23833348-23907940 on Build GRCh38
Transcripts
NM_024122.4 NR_026545.2 XM_011545584.2 XM_011545585.2 XM_011545586.2 XM_011545587.2 XM_017029837.1

SNP Detail

SNP ID Chromosome Location
rs894609971 Chr.X: 23868528-23868528 on Build GRCh38
rs372937215 Chr.X: 23868638-23868638 on Build GRCh38
rs6526354 Chr.X: 23868574-23868574 on Build GRCh38
rs953161742 Chr.X: 23868761-23868761 on Build GRCh38
rs929727704 Chr.X: 23868404-23868405 on Build GRCh38
rs759754973 Chr.X: 23868728-23868728 on Build GRCh38
rs749323940 Chr.X: 23868736-23868740 on Build GRCh38
rs752763586 Chr.X: 23868735-23868736 on Build GRCh38
rs918370447 Chr.X: 23868396-23868397 on Build GRCh38
rs755276090 Chr.X: 23868715-23868716 on Build GRCh38
rs768859557 Chr.X: 23868656-23868656 on Build GRCh38
rs112869326 Chr.X: 23868459-23868459 on Build GRCh38
rs752566066 Chr.X: 23868397-23868397 on Build GRCh38
rs757634641 Chr.X: 23868645-23868645 on Build GRCh38
rs747431777 Chr.X: 23868532-23868532 on Build GRCh38
rs776307138 Chr.X: 23868712-23868712 on Build GRCh38
rs756977763 Chr.X: 23868497-23868497 on Build GRCh38
rs753270014 Chr.X: 23868490-23868490 on Build GRCh38
rs186381955 Chr.X: 23868511-23868511 on Build GRCh38
rs1030761996 Chr.X: 23868775-23868775 on Build GRCh38
rs768521438 Chr.X: 23868676-23868676 on Build GRCh38
rs369524077 Chr.X: 23868578-23868578 on Build GRCh38
rs748623374 Chr.X: 23868599-23868599 on Build GRCh38
rs200271508 Chr.X: 23868655-23868655 on Build GRCh38
rs1048118514 Chr.X: 23868464-23868464 on Build GRCh38
rs1030700096 Chr.X: 23868619-23868619 on Build GRCh38
rs915000495 Chr.X: 23868738-23868738 on Build GRCh38
rs989189130 Chr.X: 23868700-23868700 on Build GRCh38
rs769601360 Chr.X: 23868711-23868711 on Build GRCh38
rs769784742 Chr.X: 23868646-23868646 on Build GRCh38
rs1013048003 Chr.X: 23868547-23868547 on Build GRCh38
rs763248182 Chr.X: 23868548-23868548 on Build GRCh38
rs746952335 Chr.X: 23868666-23868666 on Build GRCh38
rs764560865 Chr.X: 23868618-23868618 on Build GRCh38
rs888277721 Chr.X: 23868466-23868466 on Build GRCh38
rs772313348 Chr.X: 23868732-23868732 on Build GRCh38
rs747962420 Chr.X: 23868693-23868693 on Build GRCh38
rs775648875 Chr.X: 23868733-23868733 on Build GRCh38
rs911777343 Chr.X: 23868837-23868837 on Build GRCh38
rs759576077 Chr.X: 23868576-23868576 on Build GRCh38
rs755613246 Chr.X: 23868452-23868452 on Build GRCh38
rs985979016 Chr.X: 23868821-23868821 on Build GRCh38
rs754095247 Chr.X: 23868634-23868634 on Build GRCh38
rs756652973 Chr.X: 23868614-23868614 on Build GRCh38

View Full Product Details