Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23874164-23874666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOO
Entrez Gene ID
79135
Gene Name
apolipoprotein O
Gene Aliases
FAM121B, MIC26, Mic23, My025
UniGene
Hs.495851
Gene Chromosome Location
Chr.X: 23833348-23907940 on Build GRCh38
Transcripts
NM_024122.4 NR_026545.2 XM_011545584.2 XM_011545585.2 XM_011545586.2 XM_011545587.2 XM_017029837.1

SNP Detail

SNP ID Chromosome Location
rs746494270 Chr.X: 23874382-23874382 on Build GRCh38
rs979725277 Chr.X: 23874291-23874291 on Build GRCh38
rs1004015630 Chr.X: 23874244-23874244 on Build GRCh38
rs940526811 Chr.X: 23874497-23874497 on Build GRCh38
rs1020205701 Chr.X: 23874190-23874190 on Build GRCh38
rs757873120 Chr.X: 23874357-23874357 on Build GRCh38
rs925513890 Chr.X: 23874557-23874557 on Build GRCh38
rs779552153 Chr.X: 23874426-23874426 on Build GRCh38
rs1037660508 Chr.X: 23874508-23874508 on Build GRCh38
rs751126282 Chr.X: 23874427-23874427 on Build GRCh38
rs760875349 Chr.X: 23874501-23874501 on Build GRCh38
rs997041949 Chr.X: 23874188-23874188 on Build GRCh38
rs1015442846 Chr.X: 23874269-23874269 on Build GRCh38
rs1051258212 Chr.X: 23874618-23874618 on Build GRCh38
rs966841857 Chr.X: 23874430-23874430 on Build GRCh38
rs12009399 Chr.X: 23874344-23874344 on Build GRCh38
rs768365876 Chr.X: 23874591-23874591 on Build GRCh38
rs370282991 Chr.X: 23874380-23874380 on Build GRCh38
rs932343387 Chr.X: 23874596-23874596 on Build GRCh38
rs777587929 Chr.X: 23874477-23874477 on Build GRCh38
rs754337528 Chr.X: 23874359-23874359 on Build GRCh38
rs746095920 Chr.X: 23874478-23874478 on Build GRCh38
rs757113186 Chr.X: 23874305-23874305 on Build GRCh38
rs757978777 Chr.X: 23874409-23874409 on Build GRCh38
rs775843420 Chr.X: 23874491-23874491 on Build GRCh38
rs956200336 Chr.X: 23874212-23874212 on Build GRCh38
rs947079024 Chr.X: 23874279-23874279 on Build GRCh38
rs995534173 Chr.X: 23874584-23874589 on Build GRCh38
rs1050870436 Chr.X: 23874201-23874201 on Build GRCh38
rs769499730 Chr.X: 23874455-23874455 on Build GRCh38
rs780810263 Chr.X: 23874631-23874631 on Build GRCh38
rs962625915 Chr.X: 23874290-23874290 on Build GRCh38
rs866052403 Chr.X: 23874408-23874408 on Build GRCh38
rs762130548 Chr.X: 23874365-23874365 on Build GRCh38
rs936942782 Chr.X: 23874567-23874567 on Build GRCh38
rs754478339 Chr.X: 23874444-23874444 on Build GRCh38
rs959402988 Chr.X: 23874343-23874343 on Build GRCh38
rs748097340 Chr.X: 23874453-23874453 on Build GRCh38
rs989832346 Chr.X: 23874587-23874587 on Build GRCh38
rs778716567 Chr.X: 23874222-23874222 on Build GRCh38
rs914824836 Chr.X: 23874223-23874223 on Build GRCh38
rs778548713 Chr.X: 23874314-23874314 on Build GRCh38
rs373588929 Chr.X: 23874452-23874452 on Build GRCh38
rs772366132 Chr.X: 23874489-23874489 on Build GRCh38
rs745837685 Chr.X: 23874335-23874335 on Build GRCh38

View Full Product Details