Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 27981237-27981735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF8L1
Entrez Gene ID
139425
Gene Name
DDB1 and CUL4 associated factor 8 like 1
Gene Aliases
WDR42B
UniGene
Hs.447365
Gene Chromosome Location
Chr.X: 27977993-27981449 on Build GRCh38
Transcripts
NM_001017930.1

SNP Detail

SNP ID Chromosome Location
rs12014586 Chr.X: 27981466-27981466 on Build GRCh38
rs749957050 Chr.X: 27981302-27981302 on Build GRCh38
rs771780546 Chr.X: 27981480-27981480 on Build GRCh38
rs186439125 Chr.X: 27981440-27981440 on Build GRCh38
rs1017755584 Chr.X: 27981448-27981448 on Build GRCh38
rs760962915 Chr.X: 27981259-27981259 on Build GRCh38
rs868736170 Chr.X: 27981392-27981392 on Build GRCh38
rs191756756 Chr.X: 27981475-27981475 on Build GRCh38
rs764776423 Chr.X: 27981294-27981294 on Build GRCh38
rs1038475701 Chr.X: 27981563-27981563 on Build GRCh38
rs751455513 Chr.X: 27981314-27981314 on Build GRCh38
rs201028001 Chr.X: 27981340-27981340 on Build GRCh38
rs1020744384 Chr.X: 27981425-27981425 on Build GRCh38
rs903611131 Chr.X: 27981478-27981478 on Build GRCh38
rs776215046 Chr.X: 27981276-27981276 on Build GRCh38
rs754784040 Chr.X: 27981328-27981328 on Build GRCh38
rs984748463 Chr.X: 27981607-27981607 on Build GRCh38
rs773190109 Chr.X: 27981501-27981501 on Build GRCh38
rs772117588 Chr.X: 27981659-27981659 on Build GRCh38
rs148725031 Chr.X: 27981690-27981690 on Build GRCh38
rs781055870 Chr.X: 27981380-27981380 on Build GRCh38
rs1034963493 Chr.X: 27981403-27981403 on Build GRCh38
rs780486310 Chr.X: 27981364-27981364 on Build GRCh38
rs772134980 Chr.X: 27981359-27981359 on Build GRCh38
rs749719099 Chr.X: 27981462-27981462 on Build GRCh38
rs982922138 Chr.X: 27981499-27981499 on Build GRCh38
rs761423990 Chr.X: 27981291-27981291 on Build GRCh38
rs747913922 Chr.X: 27981415-27981415 on Build GRCh38
rs998113496 Chr.X: 27981570-27981570 on Build GRCh38
rs775390599 Chr.X: 27981701-27981701 on Build GRCh38
rs1031116646 Chr.X: 27981506-27981506 on Build GRCh38
rs781034330 Chr.X: 27981337-27981337 on Build GRCh38
rs747006668 Chr.X: 27981611-27981611 on Build GRCh38
rs1016311335 Chr.X: 27981616-27981616 on Build GRCh38
rs866774220 Chr.X: 27981288-27981288 on Build GRCh38
rs1009419390 Chr.X: 27981410-27981410 on Build GRCh38
rs879082608 Chr.X: 27981429-27981429 on Build GRCh38
rs896375413 Chr.X: 27981409-27981409 on Build GRCh38
rs762988432 Chr.X: 27981367-27981367 on Build GRCh38
rs182018658 Chr.X: 27981374-27981374 on Build GRCh38
rs1056455566 Chr.X: 27981625-27981625 on Build GRCh38
rs909260045 Chr.X: 27981338-27981338 on Build GRCh38
rs918460136 Chr.X: 27981407-27981407 on Build GRCh38
rs183505313 Chr.X: 27981641-27981641 on Build GRCh38
rs903723135 Chr.X: 27981707-27981707 on Build GRCh38
rs750103037 Chr.X: 27981422-27981422 on Build GRCh38
rs1021281906 Chr.X: 27981697-27981697 on Build GRCh38
rs1056407760 Chr.X: 27981416-27981416 on Build GRCh38
rs748325291 Chr.X: 27981487-27981487 on Build GRCh38
rs941725797 Chr.X: 27981608-27981608 on Build GRCh38
rs938006486 Chr.X: 27981572-27981572 on Build GRCh38
rs964377081 Chr.X: 27981451-27981451 on Build GRCh38
rs144231744 Chr.X: 27981491-27981491 on Build GRCh38
rs773362690 Chr.X: 27981571-27981571 on Build GRCh38
rs780488710 Chr.X: 27981443-27981443 on Build GRCh38
rs908744592 Chr.X: 27981503-27981503 on Build GRCh38
rs375923258 Chr.X: 27981342-27981342 on Build GRCh38
rs941524399 Chr.X: 27981518-27981518 on Build GRCh38
rs145556673 Chr.X: 27981310-27981310 on Build GRCh38
rs757527340 Chr.X: 27981341-27981341 on Build GRCh38
rs200028096 Chr.X: 27981307-27981307 on Build GRCh38
rs769053531 Chr.X: 27981262-27981262 on Build GRCh38
rs972170233 Chr.X: 27981684-27981684 on Build GRCh38
rs896403450 Chr.X: 27981646-27981646 on Build GRCh38

View Full Product Details