Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37011219-37011751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47C
Entrez Gene ID
442444
Gene Name
family with sequence similarity 47 member C
Gene Aliases
-
UniGene
Hs.535216
Gene Chromosome Location
Chr.X: 37008359-37011666 on Build GRCh38
Transcripts
NM_001013736.2

SNP Detail

SNP ID Chromosome Location
rs782432221 Chr.X: 37011245-37011245 on Build GRCh38
rs782673621 Chr.X: 37011393-37011393 on Build GRCh38
rs375998369 Chr.X: 37011273-37011273 on Build GRCh38
rs782041320 Chr.X: 37011331-37011331 on Build GRCh38
rs373819113 Chr.X: 37011462-37011462 on Build GRCh38
rs782017607 Chr.X: 37011517-37011517 on Build GRCh38
rs782739471 Chr.X: 37011503-37011503 on Build GRCh38
rs781796853 Chr.X: 37011492-37011492 on Build GRCh38
rs782519604 Chr.X: 37011587-37011587 on Build GRCh38
rs781878008 Chr.X: 37011550-37011550 on Build GRCh38
rs782306470 Chr.X: 37011252-37011252 on Build GRCh38
rs782560952 Chr.X: 37011422-37011422 on Build GRCh38
rs782337465 Chr.X: 37011447-37011447 on Build GRCh38
rs782273856 Chr.X: 37011401-37011401 on Build GRCh38
rs782492479 Chr.X: 37011350-37011350 on Build GRCh38
rs143561128 Chr.X: 37011450-37011450 on Build GRCh38
rs781942318 Chr.X: 37011471-37011471 on Build GRCh38
rs782092998 Chr.X: 37011446-37011446 on Build GRCh38
rs782730292 Chr.X: 37011340-37011340 on Build GRCh38
rs782722000 Chr.X: 37011534-37011534 on Build GRCh38
rs782008490 Chr.X: 37011249-37011249 on Build GRCh38
rs782365655 Chr.X: 37011471-37011472 on Build GRCh38
rs200986221 Chr.X: 37011352-37011352 on Build GRCh38
rs782509331 Chr.X: 37011388-37011388 on Build GRCh38
rs782648270 Chr.X: 37011343-37011344 on Build GRCh38
rs781783357 Chr.X: 37011559-37011559 on Build GRCh38
rs189955231 Chr.X: 37011646-37011646 on Build GRCh38
rs782553697 Chr.X: 37011505-37011505 on Build GRCh38
rs782760369 Chr.X: 37011507-37011507 on Build GRCh38
rs782657203 Chr.X: 37011242-37011242 on Build GRCh38
rs28592679 Chr.X: 37011705-37011705 on Build GRCh38
rs782191586 Chr.X: 37011250-37011250 on Build GRCh38
rs781869760 Chr.X: 37011377-37011377 on Build GRCh38
rs201493707 Chr.X: 37011280-37011280 on Build GRCh38
rs782441970 Chr.X: 37011405-37011405 on Build GRCh38
rs782293631 Chr.X: 37011651-37011651 on Build GRCh38
rs782240419 Chr.X: 37011243-37011243 on Build GRCh38
rs782527841 Chr.X: 37011524-37011524 on Build GRCh38
rs782296386 Chr.X: 37011453-37011453 on Build GRCh38
rs41311787 Chr.X: 37011320-37011320 on Build GRCh38
rs782174637 Chr.X: 37011333-37011333 on Build GRCh38
rs781792626 Chr.X: 37011341-37011341 on Build GRCh38
rs146796397 Chr.X: 37011506-37011506 on Build GRCh38
rs782417722 Chr.X: 37011317-37011318 on Build GRCh38

View Full Product Details