Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41515646-41516142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASK
Entrez Gene ID
8573
Gene Name
calcium/calmodulin dependent serine protein kinase
Gene Aliases
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8
UniGene
Hs.495984
Gene Chromosome Location
Chr.X: 41514934-41923525 on Build GRCh38
Transcripts
NM_001126054.2 NM_001126055.2 NM_003688.3 XM_005272686.4 XM_006724566.3 XM_011543993.2 XM_011543994.2 XM_011543995.2 XM_011543996.2 XM_011543997.2

SNP Detail

SNP ID Chromosome Location
rs750862450 Chr.X: 41516010-41516010 on Build GRCh38
rs546776813 Chr.X: 41515792-41515792 on Build GRCh38
rs1051666393 Chr.X: 41515993-41515993 on Build GRCh38
rs930587054 Chr.X: 41516108-41516108 on Build GRCh38
rs774443711 Chr.X: 41515873-41515873 on Build GRCh38
rs928058649 Chr.X: 41515915-41515915 on Build GRCh38
rs769186373 Chr.X: 41515859-41515859 on Build GRCh38
rs1039805253 Chr.X: 41515853-41515853 on Build GRCh38
rs1008682066 Chr.X: 41515688-41515688 on Build GRCh38
rs767739558 Chr.X: 41516104-41516104 on Build GRCh38
rs940253608 Chr.X: 41516073-41516073 on Build GRCh38
rs5918193 Chr.X: 41515876-41515876 on Build GRCh38
rs989650019 Chr.X: 41515827-41515827 on Build GRCh38
rs571469394 Chr.X: 41515826-41515826 on Build GRCh38
rs1038067273 Chr.X: 41516090-41516090 on Build GRCh38
rs758656067 Chr.X: 41516026-41516026 on Build GRCh38
rs982243789 Chr.X: 41515900-41515900 on Build GRCh38
rs55749198 Chr.X: 41515943-41515943 on Build GRCh38
rs368493915 Chr.X: 41515771-41515771 on Build GRCh38
rs967092892 Chr.X: 41515756-41515756 on Build GRCh38
rs940759060 Chr.X: 41515955-41515955 on Build GRCh38
rs779272776 Chr.X: 41515911-41515912 on Build GRCh38
rs1032610680 Chr.X: 41515823-41515823 on Build GRCh38
rs1019098340 Chr.X: 41515755-41515755 on Build GRCh38
rs948210551 Chr.X: 41515877-41515877 on Build GRCh38
rs754351304 Chr.X: 41515764-41515764 on Build GRCh38

View Full Product Details