Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41516446-41516943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASK
Entrez Gene ID
8573
Gene Name
calcium/calmodulin dependent serine protein kinase
Gene Aliases
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8
UniGene
Hs.495984
Gene Chromosome Location
Chr.X: 41514934-41923525 on Build GRCh38
Transcripts
NM_001126054.2 NM_001126055.2 NM_003688.3 XM_005272686.4 XM_006724566.3 XM_011543993.2 XM_011543994.2 XM_011543995.2 XM_011543996.2 XM_011543997.2

SNP Detail

SNP ID Chromosome Location
rs186139866 Chr.X: 41516596-41516596 on Build GRCh38
rs1023827297 Chr.X: 41516525-41516525 on Build GRCh38
rs760097133 Chr.X: 41516845-41516845 on Build GRCh38
rs1020765948 Chr.X: 41516782-41516782 on Build GRCh38
rs769839344 Chr.X: 41516578-41516578 on Build GRCh38
rs376774093 Chr.X: 41516908-41516908 on Build GRCh38
rs550893314 Chr.X: 41516598-41516598 on Build GRCh38
rs1032225935 Chr.X: 41516842-41516843 on Build GRCh38
rs752259517 Chr.X: 41516757-41516757 on Build GRCh38
rs1000632606 Chr.X: 41516807-41516807 on Build GRCh38
rs969646116 Chr.X: 41516550-41516550 on Build GRCh38
rs190946012 Chr.X: 41516668-41516668 on Build GRCh38
rs975783135 Chr.X: 41516546-41516546 on Build GRCh38
rs755743147 Chr.X: 41516682-41516684 on Build GRCh38
rs5917432 Chr.X: 41516748-41516748 on Build GRCh38
rs1050358977 Chr.X: 41516689-41516689 on Build GRCh38
rs762855700 Chr.X: 41516617-41516617 on Build GRCh38
rs987026276 Chr.X: 41516601-41516601 on Build GRCh38
rs182653355 Chr.X: 41516731-41516731 on Build GRCh38
rs770668019 Chr.X: 41516618-41516618 on Build GRCh38
rs1040619007 Chr.X: 41516824-41516824 on Build GRCh38
rs937694425 Chr.X: 41516687-41516687 on Build GRCh38
rs902341075 Chr.X: 41516805-41516805 on Build GRCh38
rs930619715 Chr.X: 41516655-41516655 on Build GRCh38
rs182354197 Chr.X: 41516567-41516567 on Build GRCh38
rs1055164180 Chr.X: 41516690-41516690 on Build GRCh38
rs1038645938 Chr.X: 41516623-41516623 on Build GRCh38
rs908692687 Chr.X: 41516616-41516616 on Build GRCh38
rs972140740 Chr.X: 41516641-41516641 on Build GRCh38
rs372182896 Chr.X: 41516658-41516658 on Build GRCh38
rs920629084 Chr.X: 41516645-41516645 on Build GRCh38
rs41312112 Chr.X: 41516570-41516570 on Build GRCh38
rs975473730 Chr.X: 41516860-41516861 on Build GRCh38
rs957413395 Chr.X: 41516526-41516526 on Build GRCh38
rs774028356 Chr.X: 41516621-41516621 on Build GRCh38
rs760943244 Chr.X: 41516579-41516579 on Build GRCh38
rs879231283 Chr.X: 41516862-41516862 on Build GRCh38
rs964953837 Chr.X: 41516850-41516850 on Build GRCh38

View Full Product Details