Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41518422-41518934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASK
Entrez Gene ID
8573
Gene Name
calcium/calmodulin dependent serine protein kinase
Gene Aliases
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8
UniGene
Hs.495984
Gene Chromosome Location
Chr.X: 41514934-41923525 on Build GRCh38
Transcripts
NM_001126054.2 NM_001126055.2 NM_003688.3 XM_005272686.4 XM_006724566.3 XM_011543993.2 XM_011543994.2 XM_011543995.2 XM_011543996.2 XM_011543997.2

SNP Detail

SNP ID Chromosome Location
rs978300777 Chr.X: 41518579-41518579 on Build GRCh38
rs1036291686 Chr.X: 41518798-41518798 on Build GRCh38
rs756902704 Chr.X: 41518807-41518807 on Build GRCh38
rs775226566 Chr.X: 41518556-41518556 on Build GRCh38
rs979974932 Chr.X: 41518708-41518708 on Build GRCh38
rs959818133 Chr.X: 41518778-41518778 on Build GRCh38
rs373349122 Chr.X: 41518599-41518599 on Build GRCh38
rs1012865871 Chr.X: 41518461-41518461 on Build GRCh38
rs921684393 Chr.X: 41518605-41518605 on Build GRCh38
rs192576790 Chr.X: 41518600-41518600 on Build GRCh38
rs1031674982 Chr.X: 41518598-41518598 on Build GRCh38
rs970977460 Chr.X: 41518569-41518569 on Build GRCh38
rs41310603 Chr.X: 41518611-41518611 on Build GRCh38
rs972473987 Chr.X: 41518784-41518784 on Build GRCh38
rs887971461 Chr.X: 41518469-41518469 on Build GRCh38
rs187703004 Chr.X: 41518543-41518543 on Build GRCh38
rs765282397 Chr.X: 41518539-41518539 on Build GRCh38
rs553102748 Chr.X: 41518874-41518874 on Build GRCh38
rs767035009 Chr.X: 41518696-41518696 on Build GRCh38
rs930950060 Chr.X: 41518849-41518849 on Build GRCh38
rs1011477164 Chr.X: 41518606-41518606 on Build GRCh38
rs987750391 Chr.X: 41518655-41518655 on Build GRCh38
rs184049666 Chr.X: 41518483-41518483 on Build GRCh38
rs749358228 Chr.X: 41518557-41518557 on Build GRCh38
rs1049362681 Chr.X: 41518860-41518860 on Build GRCh38
rs369096388 Chr.X: 41518445-41518445 on Build GRCh38
rs778814275 Chr.X: 41518477-41518477 on Build GRCh38
rs746601269 Chr.X: 41518590-41518590 on Build GRCh38
rs184249905 Chr.X: 41518836-41518836 on Build GRCh38
rs1049334384 Chr.X: 41518452-41518452 on Build GRCh38
rs928422726 Chr.X: 41518713-41518713 on Build GRCh38
rs761393070 Chr.X: 41518694-41518694 on Build GRCh38
rs759330038 Chr.X: 41518497-41518497 on Build GRCh38

View Full Product Details