Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41530915-41531420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASK
Entrez Gene ID
8573
Gene Name
calcium/calmodulin dependent serine protein kinase
Gene Aliases
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8
UniGene
Hs.495984
Gene Chromosome Location
Chr.X: 41514934-41923525 on Build GRCh38
Transcripts
NM_001126054.2 NM_001126055.2 NM_003688.3 XM_005272686.4 XM_006724566.3 XM_011543993.2 XM_011543994.2 XM_011543995.2 XM_011543996.2 XM_011543997.2

SNP Detail

SNP ID Chromosome Location
rs946398902 Chr.X: 41531262-41531262 on Build GRCh38
rs371990384 Chr.X: 41531094-41531094 on Build GRCh38
rs759433935 Chr.X: 41531354-41531354 on Build GRCh38
rs993312022 Chr.X: 41531343-41531343 on Build GRCh38
rs775353846 Chr.X: 41531046-41531046 on Build GRCh38
rs138663694 Chr.X: 41531350-41531350 on Build GRCh38
rs1039408249 Chr.X: 41531249-41531249 on Build GRCh38
rs1050084691 Chr.X: 41531264-41531264 on Build GRCh38
rs1053117804 Chr.X: 41531246-41531246 on Build GRCh38
rs897656765 Chr.X: 41531321-41531321 on Build GRCh38
rs1010903804 Chr.X: 41531337-41531337 on Build GRCh38
rs150078824 Chr.X: 41531256-41531256 on Build GRCh38
rs759736132 Chr.X: 41531041-41531041 on Build GRCh38
rs751235198 Chr.X: 41531117-41531117 on Build GRCh38
rs375734729 Chr.X: 41531019-41531019 on Build GRCh38
rs1041925080 Chr.X: 41531374-41531374 on Build GRCh38
rs141158465 Chr.X: 41531085-41531085 on Build GRCh38
rs863224854 Chr.X: 41531211-41531211 on Build GRCh38
rs774433205 Chr.X: 41531038-41531038 on Build GRCh38
rs775519672 Chr.X: 41531197-41531197 on Build GRCh38
rs762911111 Chr.X: 41531288-41531288 on Build GRCh38
rs753653837 Chr.X: 41531058-41531058 on Build GRCh38
rs749742837 Chr.X: 41531135-41531135 on Build GRCh38
rs779629178 Chr.X: 41531103-41531103 on Build GRCh38
rs587783364 Chr.X: 41531042-41531042 on Build GRCh38
rs200118528 Chr.X: 41531254-41531254 on Build GRCh38
rs146936770 Chr.X: 41531147-41531147 on Build GRCh38
rs890718842 Chr.X: 41531307-41531307 on Build GRCh38
rs778572322 Chr.X: 41530966-41530966 on Build GRCh38
rs781589624 Chr.X: 41530999-41530999 on Build GRCh38
rs760957859 Chr.X: 41531202-41531202 on Build GRCh38
rs770949412 Chr.X: 41531293-41531293 on Build GRCh38
rs765293367 Chr.X: 41530988-41530988 on Build GRCh38
rs372611767 Chr.X: 41531261-41531261 on Build GRCh38
rs968871336 Chr.X: 41530965-41530965 on Build GRCh38
rs760081932 Chr.X: 41530949-41530949 on Build GRCh38
rs992804263 Chr.X: 41531022-41531022 on Build GRCh38
rs747425934 Chr.X: 41530975-41530975 on Build GRCh38
rs768962946 Chr.X: 41531018-41531018 on Build GRCh38
rs369792621 Chr.X: 41531057-41531057 on Build GRCh38
rs34998205 Chr.X: 41531373-41531374 on Build GRCh38
rs751381808 Chr.X: 41531298-41531298 on Build GRCh38
rs192044507 Chr.X: 41531045-41531045 on Build GRCh38
rs375034853 Chr.X: 41531118-41531118 on Build GRCh38
rs941141082 Chr.X: 41531199-41531199 on Build GRCh38
rs778081066 Chr.X: 41531012-41531012 on Build GRCh38
rs778228332 Chr.X: 41531121-41531121 on Build GRCh38
rs1009049426 Chr.X: 41531291-41531291 on Build GRCh38
rs758118296 Chr.X: 41531101-41531101 on Build GRCh38
rs138646715 Chr.X: 41531056-41531056 on Build GRCh38
rs55642200 Chr.X: 41531184-41531184 on Build GRCh38
rs746017736 Chr.X: 41531183-41531183 on Build GRCh38
rs761873670 Chr.X: 41531083-41531083 on Build GRCh38
rs958677925 Chr.X: 41530996-41530996 on Build GRCh38
rs372839268 Chr.X: 41531001-41531001 on Build GRCh38
rs771473384 Chr.X: 41531137-41531137 on Build GRCh38
rs1057518245 Chr.X: 41531144-41531144 on Build GRCh38

View Full Product Details