Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 105219244-105219750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RAPL2
Entrez Gene ID
26280
Gene Name
interleukin 1 receptor accessory protein like 2
Gene Aliases
IL-1R9, IL1R9, IL1RAPL-2, TIGIRR-1
UniGene
Hs.675519
Gene Chromosome Location
Chr.X: 104566315-105769729 on Build GRCh38
Transcripts
NM_017416.1 XM_017029403.1 NM_001291277.1 NM_031274.4

Gene Symbol
TEX13A
Entrez Gene ID
56157
Gene Name
testis expressed 13A
Gene Aliases
-
UniGene
Hs.567543
Gene Chromosome Location
Chr.X: 105218929-105220693 on Build GRCh38
Transcripts
NM_017416.1 XM_017029403.1 NM_001291277.1 NM_031274.4

SNP Detail

SNP ID Chromosome Location
rs377199225 Chr.X: 105219680-105219680 on Build GRCh38
rs782169422 Chr.X: 105219627-105219628 on Build GRCh38
rs782631654 Chr.X: 105219596-105219596 on Build GRCh38
rs782179258 Chr.X: 105219587-105219587 on Build GRCh38
rs782580031 Chr.X: 105219592-105219592 on Build GRCh38
rs782515691 Chr.X: 105219605-105219605 on Build GRCh38
rs781918561 Chr.X: 105219716-105219716 on Build GRCh38
rs376396802 Chr.X: 105219445-105219445 on Build GRCh38
rs782607139 Chr.X: 105219345-105219345 on Build GRCh38
rs925654106 Chr.X: 105219729-105219729 on Build GRCh38
rs367644904 Chr.X: 105219601-105219601 on Build GRCh38
rs782469368 Chr.X: 105219583-105219583 on Build GRCh38
rs782278133 Chr.X: 105219603-105219603 on Build GRCh38
rs938384055 Chr.X: 105219351-105219351 on Build GRCh38
rs782531085 Chr.X: 105219302-105219302 on Build GRCh38
rs782644087 Chr.X: 105219607-105219607 on Build GRCh38
rs938856840 Chr.X: 105219654-105219654 on Build GRCh38
rs781811258 Chr.X: 105219685-105219685 on Build GRCh38
rs370620805 Chr.X: 105219698-105219698 on Build GRCh38
rs782695080 Chr.X: 105219276-105219276 on Build GRCh38
rs376278332 Chr.X: 105219657-105219657 on Build GRCh38
rs782161125 Chr.X: 105219388-105219388 on Build GRCh38
rs782652760 Chr.X: 105219471-105219471 on Build GRCh38
rs183367827 Chr.X: 105219725-105219725 on Build GRCh38
rs782227756 Chr.X: 105219597-105219597 on Build GRCh38
rs56000396 Chr.X: 105219595-105219595 on Build GRCh38
rs980638493 Chr.X: 105219303-105219303 on Build GRCh38
rs1056932651 Chr.X: 105219371-105219371 on Build GRCh38
rs782617681 Chr.X: 105219701-105219701 on Build GRCh38
rs781880944 Chr.X: 105219507-105219507 on Build GRCh38
rs782720616 Chr.X: 105219592-105219592 on Build GRCh38
rs782108130 Chr.X: 105219486-105219486 on Build GRCh38
rs782281084 Chr.X: 105219593-105219593 on Build GRCh38
rs782402476 Chr.X: 105219390-105219390 on Build GRCh38
rs368665153 Chr.X: 105219316-105219316 on Build GRCh38
rs180688423 Chr.X: 105219453-105219453 on Build GRCh38
rs41312550 Chr.X: 105219658-105219658 on Build GRCh38
rs782687241 Chr.X: 105219355-105219355 on Build GRCh38
rs782164521 Chr.X: 105219568-105219568 on Build GRCh38
rs782437904 Chr.X: 105219588-105219588 on Build GRCh38
rs782537866 Chr.X: 105219512-105219512 on Build GRCh38
rs782632515 Chr.X: 105219380-105219380 on Build GRCh38
rs782494053 Chr.X: 105219501-105219501 on Build GRCh38
rs782741680 Chr.X: 105219546-105219547 on Build GRCh38
rs782369813 Chr.X: 105219699-105219699 on Build GRCh38
rs782401314 Chr.X: 105219613-105219613 on Build GRCh38
rs112864023 Chr.X: 105219417-105219417 on Build GRCh38
rs782082680 Chr.X: 105219530-105219530 on Build GRCh38
rs781819372 Chr.X: 105219420-105219420 on Build GRCh38
rs782755348 Chr.X: 105219578-105219578 on Build GRCh38
rs942305279 Chr.X: 105219300-105219300 on Build GRCh38
rs782062464 Chr.X: 105219333-105219333 on Build GRCh38
rs781833475 Chr.X: 105219580-105219580 on Build GRCh38
rs377312415 Chr.X: 105219552-105219552 on Build GRCh38
rs782369075 Chr.X: 105219590-105219590 on Build GRCh38
rs189616035 Chr.X: 105219392-105219392 on Build GRCh38
rs782320005 Chr.X: 105219533-105219533 on Build GRCh38
rs1030414025 Chr.X: 105219522-105219522 on Build GRCh38
rs782280904 Chr.X: 105219363-105219363 on Build GRCh38
rs891451870 Chr.X: 105219376-105219376 on Build GRCh38
rs782201089 Chr.X: 105219710-105219710 on Build GRCh38
rs781802607 Chr.X: 105219348-105219348 on Build GRCh38
rs782696967 Chr.X: 105219631-105219631 on Build GRCh38
rs781973376 Chr.X: 105219525-105219525 on Build GRCh38
rs781881356 Chr.X: 105219584-105219584 on Build GRCh38
rs781927454 Chr.X: 105219593-105219593 on Build GRCh38
rs782260670 Chr.X: 105219589-105219589 on Build GRCh38
rs782625248 Chr.X: 105219585-105219585 on Build GRCh38
rs782348932 Chr.X: 105219611-105219611 on Build GRCh38
rs782005557 Chr.X: 105219728-105219728 on Build GRCh38
rs782157629 Chr.X: 105219724-105219724 on Build GRCh38
rs983824302 Chr.X: 105219528-105219528 on Build GRCh38
rs782440581 Chr.X: 105219352-105219352 on Build GRCh38
rs372780941 Chr.X: 105219669-105219669 on Build GRCh38
rs782489830 Chr.X: 105219597-105219597 on Build GRCh38
rs782436709 Chr.X: 105219599-105219599 on Build GRCh38
rs781898459 Chr.X: 105219292-105219292 on Build GRCh38
rs781876678 Chr.X: 105219577-105219577 on Build GRCh38
rs374463333 Chr.X: 105219308-105219308 on Build GRCh38
rs782698296 Chr.X: 105219504-105219504 on Build GRCh38
rs782092010 Chr.X: 105219591-105219591 on Build GRCh38
rs782324559 Chr.X: 105219712-105219712 on Build GRCh38
rs782232598 Chr.X: 105219383-105219383 on Build GRCh38
rs781915068 Chr.X: 105219546-105219546 on Build GRCh38
rs190120031 Chr.X: 105219652-105219652 on Build GRCh38
rs950712366 Chr.X: 105219523-105219523 on Build GRCh38
rs782689683 Chr.X: 105219691-105219691 on Build GRCh38
rs782680142 Chr.X: 105219602-105219602 on Build GRCh38
rs781953376 Chr.X: 105219618-105219618 on Build GRCh38
rs782600170 Chr.X: 105219684-105219684 on Build GRCh38
rs782810571 Chr.X: 105219570-105219570 on Build GRCh38
rs781910045 Chr.X: 105219462-105219462 on Build GRCh38
rs782368474 Chr.X: 105219519-105219519 on Build GRCh38
rs367979230 Chr.X: 105219425-105219425 on Build GRCh38
rs371809236 Chr.X: 105219437-105219437 on Build GRCh38
rs782231806 Chr.X: 105219586-105219586 on Build GRCh38
rs782538724 Chr.X: 105219470-105219470 on Build GRCh38
rs111640100 Chr.X: 105219387-105219387 on Build GRCh38
rs782107353 Chr.X: 105219615-105219615 on Build GRCh38
rs782034215 Chr.X: 105219391-105219391 on Build GRCh38
rs782225912 Chr.X: 105219608-105219608 on Build GRCh38
rs782442482 Chr.X: 105219688-105219688 on Build GRCh38
rs782390890 Chr.X: 105219368-105219368 on Build GRCh38
rs372454217 Chr.X: 105219502-105219502 on Build GRCh38
rs782062617 Chr.X: 105219620-105219620 on Build GRCh38
rs185356738 Chr.X: 105219651-105219651 on Build GRCh38
rs782140073 Chr.X: 105219595-105219595 on Build GRCh38
rs202232623 Chr.X: 105219594-105219595 on Build GRCh38
rs782595598 Chr.X: 105219278-105219279 on Build GRCh38

View Full Product Details