Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 105220163-105220657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RAPL2
Entrez Gene ID
26280
Gene Name
interleukin 1 receptor accessory protein like 2
Gene Aliases
IL-1R9, IL1R9, IL1RAPL-2, TIGIRR-1
UniGene
Hs.675519
Gene Chromosome Location
Chr.X: 104566315-105769729 on Build GRCh38
Transcripts
NM_017416.1 XM_017029403.1 NM_001291277.1 NM_031274.4

Gene Symbol
TEX13A
Entrez Gene ID
56157
Gene Name
testis expressed 13A
Gene Aliases
-
UniGene
Hs.567543
Gene Chromosome Location
Chr.X: 105218929-105220693 on Build GRCh38
Transcripts
NM_017416.1 XM_017029403.1 NM_001291277.1 NM_031274.4

SNP Detail

SNP ID Chromosome Location
rs781885185 Chr.X: 105220349-105220349 on Build GRCh38
rs782100228 Chr.X: 105220439-105220439 on Build GRCh38
rs149930638 Chr.X: 105220343-105220343 on Build GRCh38
rs922291432 Chr.X: 105220468-105220468 on Build GRCh38
rs782193389 Chr.X: 105220407-105220407 on Build GRCh38
rs782210908 Chr.X: 105220265-105220265 on Build GRCh38
rs1020587782 Chr.X: 105220630-105220630 on Build GRCh38
rs782586362 Chr.X: 105220367-105220367 on Build GRCh38
rs964142491 Chr.X: 105220459-105220459 on Build GRCh38
rs782166550 Chr.X: 105220302-105220302 on Build GRCh38
rs782501337 Chr.X: 105220453-105220453 on Build GRCh38
rs782028820 Chr.X: 105220413-105220413 on Build GRCh38
rs782457588 Chr.X: 105220522-105220522 on Build GRCh38
rs781921097 Chr.X: 105220299-105220299 on Build GRCh38
rs781950967 Chr.X: 105220263-105220263 on Build GRCh38
rs782471679 Chr.X: 105220364-105220364 on Build GRCh38
rs782662637 Chr.X: 105220376-105220376 on Build GRCh38
rs781844230 Chr.X: 105220572-105220572 on Build GRCh38
rs908355767 Chr.X: 105220499-105220499 on Build GRCh38
rs372838212 Chr.X: 105220312-105220312 on Build GRCh38
rs782725405 Chr.X: 105220536-105220536 on Build GRCh38
rs6621945 Chr.X: 105220489-105220489 on Build GRCh38
rs782091176 Chr.X: 105220319-105220319 on Build GRCh38
rs1020450618 Chr.X: 105220333-105220333 on Build GRCh38
rs781807953 Chr.X: 105220441-105220441 on Build GRCh38
rs368896806 Chr.X: 105220368-105220368 on Build GRCh38
rs903342687 Chr.X: 105220632-105220632 on Build GRCh38
rs782436243 Chr.X: 105220371-105220371 on Build GRCh38
rs975010598 Chr.X: 105220465-105220465 on Build GRCh38
rs782440698 Chr.X: 105220443-105220443 on Build GRCh38
rs781877063 Chr.X: 105220401-105220401 on Build GRCh38
rs41311565 Chr.X: 105220573-105220573 on Build GRCh38
rs533463518 Chr.X: 105220212-105220212 on Build GRCh38
rs782379343 Chr.X: 105220239-105220239 on Build GRCh38
rs895847744 Chr.X: 105220599-105220599 on Build GRCh38
rs782263029 Chr.X: 105220377-105220377 on Build GRCh38
rs782732944 Chr.X: 105220440-105220440 on Build GRCh38
rs192502821 Chr.X: 105220512-105220512 on Build GRCh38
rs782704665 Chr.X: 105220342-105220342 on Build GRCh38
rs375117468 Chr.X: 105220428-105220428 on Build GRCh38
rs782701321 Chr.X: 105220183-105220183 on Build GRCh38
rs782635027 Chr.X: 105220360-105220360 on Build GRCh38
rs373094370 Chr.X: 105220416-105220416 on Build GRCh38
rs960253897 Chr.X: 105220301-105220301 on Build GRCh38
rs782459107 Chr.X: 105220226-105220226 on Build GRCh38
rs369858858 Chr.X: 105220230-105220230 on Build GRCh38
rs781800251 Chr.X: 105220332-105220332 on Build GRCh38
rs781841588 Chr.X: 105220361-105220361 on Build GRCh38
rs782789682 Chr.X: 105220417-105220417 on Build GRCh38
rs781829113 Chr.X: 105220495-105220495 on Build GRCh38
rs925759239 Chr.X: 105220410-105220410 on Build GRCh38
rs782326095 Chr.X: 105220293-105220293 on Build GRCh38
rs781911318 Chr.X: 105220192-105220192 on Build GRCh38
rs782046840 Chr.X: 105220288-105220288 on Build GRCh38
rs782592290 Chr.X: 105220633-105220633 on Build GRCh38
rs781970080 Chr.X: 105220257-105220257 on Build GRCh38
rs186710149 Chr.X: 105220535-105220535 on Build GRCh38
rs187750279 Chr.X: 105220224-105220224 on Build GRCh38
rs782305872 Chr.X: 105220411-105220411 on Build GRCh38
rs782665512 Chr.X: 105220248-105220248 on Build GRCh38
rs182068237 Chr.X: 105220513-105220513 on Build GRCh38
rs782783205 Chr.X: 105220215-105220215 on Build GRCh38
rs781979617 Chr.X: 105220402-105220402 on Build GRCh38
rs782373679 Chr.X: 105220260-105220260 on Build GRCh38
rs1014496150 Chr.X: 105220602-105220602 on Build GRCh38

View Full Product Details