Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107206181-107206674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP62CL
Entrez Gene ID
54830
Gene Name
nucleoporin 62 C-terminal like
Gene Aliases
-
UniGene
Hs.163629
Gene Chromosome Location
Chr.X: 107123427-107206440 on Build GRCh38
Transcripts
NM_017681.2 NR_033676.1 NM_001169154.1 NM_173494.1 XM_011530855.2

Gene Symbol
PIH1D3
Entrez Gene ID
139212
Gene Name
PIH1 domain containing 3
Gene Aliases
CXorf41, NYSAR97
UniGene
Hs.128836
Gene Chromosome Location
Chr.X: 107206629-107244243 on Build GRCh38
Transcripts
NM_017681.2 NR_033676.1 NM_001169154.1 NM_173494.1 XM_011530855.2

SNP Detail

SNP ID Chromosome Location
rs376279156 Chr.X: 107206407-107206407 on Build GRCh38
rs926563122 Chr.X: 107206322-107206322 on Build GRCh38
rs898501641 Chr.X: 107206224-107206224 on Build GRCh38
rs927356412 Chr.X: 107206588-107206588 on Build GRCh38
rs900377593 Chr.X: 107206395-107206395 on Build GRCh38
rs148574219 Chr.X: 107206318-107206318 on Build GRCh38
rs974220173 Chr.X: 107206624-107206624 on Build GRCh38
rs1049709489 Chr.X: 107206456-107206456 on Build GRCh38
rs958902407 Chr.X: 107206597-107206597 on Build GRCh38
rs932682512 Chr.X: 107206424-107206424 on Build GRCh38
rs1040190189 Chr.X: 107206371-107206371 on Build GRCh38
rs957270171 Chr.X: 107206446-107206446 on Build GRCh38
rs753352468 Chr.X: 107206232-107206232 on Build GRCh38
rs992014382 Chr.X: 107206375-107206375 on Build GRCh38
rs188471353 Chr.X: 107206315-107206315 on Build GRCh38
rs755707985 Chr.X: 107206264-107206264 on Build GRCh38
rs982033316 Chr.X: 107206219-107206219 on Build GRCh38
rs1022504633 Chr.X: 107206652-107206652 on Build GRCh38
rs371666759 Chr.X: 107206313-107206313 on Build GRCh38
rs981517247 Chr.X: 107206584-107206584 on Build GRCh38
rs995962881 Chr.X: 107206400-107206400 on Build GRCh38
rs956185896 Chr.X: 107206278-107206278 on Build GRCh38
rs939839563 Chr.X: 107206399-107206399 on Build GRCh38
rs193105958 Chr.X: 107206480-107206480 on Build GRCh38
rs971893059 Chr.X: 107206583-107206583 on Build GRCh38
rs891375058 Chr.X: 107206240-107206240 on Build GRCh38
rs1009744705 Chr.X: 107206279-107206279 on Build GRCh38
rs1053271701 Chr.X: 107206574-107206574 on Build GRCh38
rs937586012 Chr.X: 107206241-107206241 on Build GRCh38
rs1008608160 Chr.X: 107206365-107206365 on Build GRCh38
rs930329322 Chr.X: 107206303-107206303 on Build GRCh38
rs919982192 Chr.X: 107206403-107206403 on Build GRCh38
rs373656501 Chr.X: 107206207-107206207 on Build GRCh38
rs920174458 Chr.X: 107206272-107206272 on Build GRCh38
rs757168632 Chr.X: 107206296-107206296 on Build GRCh38
rs1022013586 Chr.X: 107206280-107206280 on Build GRCh38
rs185846932 Chr.X: 107206482-107206482 on Build GRCh38
rs2298179 Chr.X: 107206413-107206413 on Build GRCh38
rs112770553 Chr.X: 107206360-107206360 on Build GRCh38
rs763039632 Chr.X: 107206567-107206567 on Build GRCh38
rs1010664828 Chr.X: 107206472-107206472 on Build GRCh38
rs187879072 Chr.X: 107206489-107206489 on Build GRCh38
rs968122738 Chr.X: 107206286-107206286 on Build GRCh38

View Full Product Details