Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120255235-120255741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM255A
Entrez Gene ID
55026
Gene Name
transmembrane protein 255A
Gene Aliases
FAM70A
UniGene
Hs.437563
Gene Chromosome Location
Chr.X: 120250984-120311556 on Build GRCh38
Transcripts
XM_017029619.1 NM_001184742.1 NM_006777.3

Gene Symbol
ZBTB33
Entrez Gene ID
10009
Gene Name
zinc finger and BTB domain containing 33
Gene Aliases
ZNF-kaiso, ZNF348
UniGene
Hs.143604
Gene Chromosome Location
Chr.X: 120250752-120258396 on Build GRCh38
Transcripts
XM_017029619.1 NM_001184742.1 NM_006777.3

SNP Detail

SNP ID Chromosome Location
rs782337560 Chr.X: 120255443-120255443 on Build GRCh38
rs781855059 Chr.X: 120255317-120255317 on Build GRCh38
rs782008220 Chr.X: 120255262-120255262 on Build GRCh38
rs782691586 Chr.X: 120255478-120255478 on Build GRCh38
rs557274880 Chr.X: 120255287-120255287 on Build GRCh38
rs781978909 Chr.X: 120255394-120255394 on Build GRCh38
rs782273880 Chr.X: 120255361-120255361 on Build GRCh38
rs185674874 Chr.X: 120255303-120255303 on Build GRCh38
rs782323742 Chr.X: 120255698-120255698 on Build GRCh38
rs1025109821 Chr.X: 120255524-120255524 on Build GRCh38
rs374847229 Chr.X: 120255523-120255523 on Build GRCh38
rs1004993482 Chr.X: 120255574-120255574 on Build GRCh38
rs782498946 Chr.X: 120255320-120255320 on Build GRCh38
rs781906430 Chr.X: 120255515-120255515 on Build GRCh38
rs782283979 Chr.X: 120255286-120255286 on Build GRCh38
rs782436598 Chr.X: 120255349-120255349 on Build GRCh38
rs782244755 Chr.X: 120255646-120255646 on Build GRCh38
rs782218082 Chr.X: 120255438-120255438 on Build GRCh38
rs782173779 Chr.X: 120255459-120255459 on Build GRCh38
rs782195625 Chr.X: 120255332-120255332 on Build GRCh38
rs782535735 Chr.X: 120255306-120255306 on Build GRCh38
rs970420105 Chr.X: 120255537-120255537 on Build GRCh38
rs782211281 Chr.X: 120255667-120255667 on Build GRCh38
rs79151935 Chr.X: 120255288-120255288 on Build GRCh38
rs782706407 Chr.X: 120255301-120255301 on Build GRCh38
rs782374358 Chr.X: 120255334-120255334 on Build GRCh38
rs782536589 Chr.X: 120255534-120255534 on Build GRCh38
rs782000872 Chr.X: 120255472-120255472 on Build GRCh38
rs912573988 Chr.X: 120255324-120255324 on Build GRCh38
rs139977503 Chr.X: 120255614-120255614 on Build GRCh38
rs961956166 Chr.X: 120255657-120255657 on Build GRCh38
rs782604657 Chr.X: 120255327-120255327 on Build GRCh38
rs782123269 Chr.X: 120255279-120255279 on Build GRCh38
rs1000725191 Chr.X: 120255428-120255428 on Build GRCh38
rs974300245 Chr.X: 120255660-120255660 on Build GRCh38
rs782803950 Chr.X: 120255471-120255471 on Build GRCh38
rs782680446 Chr.X: 120255357-120255357 on Build GRCh38
rs190526366 Chr.X: 120255369-120255369 on Build GRCh38
rs781927736 Chr.X: 120255449-120255449 on Build GRCh38
rs782793665 Chr.X: 120255308-120255308 on Build GRCh38

View Full Product Details