Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120873814-120874325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CT47B1
Entrez Gene ID
643311
Gene Name
cancer/testis antigen family 47, member B1
Gene Aliases
CT47.13, CT47A13
UniGene
Hs.729566
Gene Chromosome Location
Chr.X: 120872554-120875925 on Build GRCh38
Transcripts
NM_001145718.1 XM_017029734.1

SNP Detail

SNP ID Chromosome Location
rs1028972047 Chr.X: 120874061-120874061 on Build GRCh38
rs761868467 Chr.X: 120873922-120873922 on Build GRCh38
rs41311647 Chr.X: 120873956-120873956 on Build GRCh38
rs1006697106 Chr.X: 120874226-120874226 on Build GRCh38
rs953432046 Chr.X: 120874094-120874094 on Build GRCh38
rs753445955 Chr.X: 120874068-120874068 on Build GRCh38
rs776806847 Chr.X: 120873894-120873894 on Build GRCh38
rs774173039 Chr.X: 120873909-120873909 on Build GRCh38
rs5956259 Chr.X: 120873838-120873838 on Build GRCh38
rs962512827 Chr.X: 120874241-120874241 on Build GRCh38
rs753346461 Chr.X: 120873940-120873940 on Build GRCh38
rs771240316 Chr.X: 120873982-120873982 on Build GRCh38
rs754450640 Chr.X: 120873941-120873941 on Build GRCh38
rs928809243 Chr.X: 120874270-120874270 on Build GRCh38
rs972170611 Chr.X: 120874263-120874263 on Build GRCh38
rs767296022 Chr.X: 120873930-120873930 on Build GRCh38
rs901052165 Chr.X: 120874033-120874033 on Build GRCh38
rs761627554 Chr.X: 120874041-120874041 on Build GRCh38
rs768575428 Chr.X: 120873904-120873904 on Build GRCh38
rs760492298 Chr.X: 120874063-120874063 on Build GRCh38
rs749280119 Chr.X: 120874063-120874064 on Build GRCh38
rs771315740 Chr.X: 120873864-120873864 on Build GRCh38
rs745907069 Chr.X: 120874002-120874002 on Build GRCh38
rs778249348 Chr.X: 120873964-120873964 on Build GRCh38
rs775707046 Chr.X: 120874040-120874040 on Build GRCh38
rs772575224 Chr.X: 120873849-120873849 on Build GRCh38
rs1016386781 Chr.X: 120874241-120874242 on Build GRCh38
rs772048023 Chr.X: 120874052-120874052 on Build GRCh38
rs747403054 Chr.X: 120873965-120873965 on Build GRCh38
rs778335477 Chr.X: 120873851-120873851 on Build GRCh38
rs750260118 Chr.X: 120873931-120873931 on Build GRCh38
rs747227235 Chr.X: 120873853-120873853 on Build GRCh38
rs779414722 Chr.X: 120873949-120873949 on Build GRCh38
rs765950011 Chr.X: 120874064-120874064 on Build GRCh38
rs759567971 Chr.X: 120873899-120873899 on Build GRCh38
rs779807535 Chr.X: 120873928-120873929 on Build GRCh38
rs760300034 Chr.X: 120873935-120873935 on Build GRCh38
rs777911161 Chr.X: 120874295-120874297 on Build GRCh38
rs769750173 Chr.X: 120874031-120874031 on Build GRCh38
rs774932604 Chr.X: 120874000-120874000 on Build GRCh38
rs5957502 Chr.X: 120874103-120874103 on Build GRCh38
rs760206366 Chr.X: 120874198-120874198 on Build GRCh38
rs772941256 Chr.X: 120874058-120874058 on Build GRCh38

View Full Product Details