Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134781743-134782249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM122B
Entrez Gene ID
159090
Gene Name
family with sequence similarity 122B
Gene Aliases
SPACIA2
UniGene
Hs.404706
Gene Chromosome Location
Chr.X: 134769566-134797232 on Build GRCh38
Transcripts
NM_001166599.2 NM_001166600.2 NM_001170756.1 NM_001170757.1 NM_145284.5 XM_006724730.2 XM_006724732.2 XM_006724733.3 XM_011531282.1 XM_011531283.1 XM_011531284.1 XM_011531285.1 XM_011531286.1 XM_011531287.1 XM_011531288.1 XM_011531289.1 XM_011531290.1 XM_011531291.1 XM_011531292.1 XM_011531293.1 XM_011531294.1 XM_017029306.1 XM_017029307.1

SNP Detail

SNP ID Chromosome Location
rs376577809 Chr.X: 134782205-134782205 on Build GRCh38
rs941501244 Chr.X: 134782144-134782144 on Build GRCh38
rs746379669 Chr.X: 134781798-134781798 on Build GRCh38
rs888813219 Chr.X: 134781895-134781895 on Build GRCh38
rs764057426 Chr.X: 134782222-134782222 on Build GRCh38
rs766984262 Chr.X: 134781969-134781969 on Build GRCh38
rs149511692 Chr.X: 134782181-134782181 on Build GRCh38
rs776440947 Chr.X: 134781856-134781856 on Build GRCh38
rs756782459 Chr.X: 134781968-134781968 on Build GRCh38
rs771524036 Chr.X: 134781811-134781812 on Build GRCh38
rs1056494893 Chr.X: 134781785-134781785 on Build GRCh38
rs762705593 Chr.X: 134781889-134781889 on Build GRCh38
rs939406598 Chr.X: 134782213-134782213 on Build GRCh38
rs748223906 Chr.X: 134781847-134781847 on Build GRCh38
rs945498122 Chr.X: 134781795-134781795 on Build GRCh38
rs137982213 Chr.X: 134781844-134781844 on Build GRCh38
rs751336171 Chr.X: 134781838-134781838 on Build GRCh38
rs114190003 Chr.X: 134782098-134782098 on Build GRCh38
rs374009398 Chr.X: 134781845-134781845 on Build GRCh38
rs910135321 Chr.X: 134782130-134782130 on Build GRCh38
rs150742745 Chr.X: 134782061-134782061 on Build GRCh38
rs763785225 Chr.X: 134781930-134781930 on Build GRCh38
rs368857530 Chr.X: 134781935-134781935 on Build GRCh38
rs771375026 Chr.X: 134781812-134781812 on Build GRCh38
rs1056582621 Chr.X: 134782220-134782220 on Build GRCh38
rs41304526 Chr.X: 134781858-134781858 on Build GRCh38
rs192108895 Chr.X: 134781991-134781991 on Build GRCh38
rs1043927082 Chr.X: 134781809-134781809 on Build GRCh38
rs983047875 Chr.X: 134782116-134782116 on Build GRCh38
rs765043561 Chr.X: 134781859-134781859 on Build GRCh38
rs1017267864 Chr.X: 134781884-134781884 on Build GRCh38
rs781609127 Chr.X: 134781787-134781787 on Build GRCh38
rs775229809 Chr.X: 134781873-134781873 on Build GRCh38
rs926638378 Chr.X: 134782198-134782198 on Build GRCh38
rs756771148 Chr.X: 134781770-134781770 on Build GRCh38
rs1019070431 Chr.X: 134782055-134782055 on Build GRCh38
rs760593693 Chr.X: 134782114-134782114 on Build GRCh38
rs965784512 Chr.X: 134781902-134781902 on Build GRCh38
rs755553879 Chr.X: 134782086-134782086 on Build GRCh38
rs772175799 Chr.X: 134781850-134781850 on Build GRCh38
rs372885090 Chr.X: 134781951-134781951 on Build GRCh38
rs951942839 Chr.X: 134782159-134782159 on Build GRCh38
rs980955219 Chr.X: 134782168-134782168 on Build GRCh38
rs757171918 Chr.X: 134781846-134781846 on Build GRCh38
rs756437393 Chr.X: 134781823-134781823 on Build GRCh38

View Full Product Details