Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134796556-134797085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM122B
Entrez Gene ID
159090
Gene Name
family with sequence similarity 122B
Gene Aliases
SPACIA2
UniGene
Hs.404706
Gene Chromosome Location
Chr.X: 134769566-134797232 on Build GRCh38
Transcripts
NM_001166599.2 NM_001166600.2 NM_001170757.1 XM_006724730.2 XM_006724732.2 XM_006724733.3 XM_011531282.1 XM_011531283.1 XM_011531284.1 XM_011531285.1 XM_011531286.1 XM_011531287.1 XM_011531288.1 XM_011531289.1 XM_011531290.1 XM_011531291.1 XM_011531292.1 XM_011531293.1 XM_011531294.1 XM_017029306.1 XM_017029307.1 NM_001170780.1

Gene Symbol
FAM122C
Entrez Gene ID
159091
Gene Name
family with sequence similarity 122C
Gene Aliases
-
UniGene
Hs.269127
Gene Chromosome Location
Chr.X: 134796413-134855137 on Build GRCh38
Transcripts
NM_001166599.2 NM_001166600.2 NM_001170757.1 XM_006724730.2 XM_006724732.2 XM_006724733.3 XM_011531282.1 XM_011531283.1 XM_011531284.1 XM_011531285.1 XM_011531286.1 XM_011531287.1 XM_011531288.1 XM_011531289.1 XM_011531290.1 XM_011531291.1 XM_011531292.1 XM_011531293.1 XM_011531294.1 XM_017029306.1 XM_017029307.1 NM_001170780.1

SNP Detail

SNP ID Chromosome Location
rs963107147 Chr.X: 134796956-134796956 on Build GRCh38
rs765344830 Chr.X: 134796829-134796829 on Build GRCh38
rs999317761 Chr.X: 134796955-134796955 on Build GRCh38
rs923274546 Chr.X: 134796621-134796621 on Build GRCh38
rs1010703637 Chr.X: 134796999-134796999 on Build GRCh38
rs893602057 Chr.X: 134796971-134796971 on Build GRCh38
rs775095537 Chr.X: 134796917-134796917 on Build GRCh38
rs906262532 Chr.X: 134796941-134796941 on Build GRCh38
rs762927411 Chr.X: 134796900-134796900 on Build GRCh38
rs947677591 Chr.X: 134796879-134796879 on Build GRCh38
rs974764298 Chr.X: 134796615-134796615 on Build GRCh38
rs983345575 Chr.X: 134796812-134796812 on Build GRCh38
rs777087404 Chr.X: 134796727-134796727 on Build GRCh38
rs954502884 Chr.X: 134796948-134796948 on Build GRCh38
rs976735822 Chr.X: 134796742-134796742 on Build GRCh38
rs938276956 Chr.X: 134796784-134796784 on Build GRCh38
rs907740595 Chr.X: 134796952-134796952 on Build GRCh38
rs983295693 Chr.X: 134796950-134796950 on Build GRCh38
rs941646556 Chr.X: 134796681-134796681 on Build GRCh38
rs894741668 Chr.X: 134796858-134796858 on Build GRCh38
rs941526347 Chr.X: 134796832-134796832 on Build GRCh38
rs896618193 Chr.X: 134796805-134796805 on Build GRCh38
rs926977251 Chr.X: 134796741-134796741 on Build GRCh38
rs973602437 Chr.X: 134797032-134797032 on Build GRCh38
rs930102375 Chr.X: 134796799-134796799 on Build GRCh38
rs943665103 Chr.X: 134796609-134796609 on Build GRCh38
rs1018120447 Chr.X: 134797024-134797024 on Build GRCh38
rs188270507 Chr.X: 134796739-134796739 on Build GRCh38
rs926973556 Chr.X: 134797046-134797046 on Build GRCh38
rs368201434 Chr.X: 134796856-134796856 on Build GRCh38
rs976704781 Chr.X: 134797061-134797061 on Build GRCh38
rs933381046 Chr.X: 134796625-134796625 on Build GRCh38
rs922667329 Chr.X: 134796777-134796777 on Build GRCh38
rs996071602 Chr.X: 134796924-134796924 on Build GRCh38
rs902592537 Chr.X: 134796831-134796831 on Build GRCh38
rs963949365 Chr.X: 134797028-134797028 on Build GRCh38
rs1000928044 Chr.X: 134796848-134796848 on Build GRCh38
rs937011920 Chr.X: 134797048-134797048 on Build GRCh38
rs1013710194 Chr.X: 134796823-134796823 on Build GRCh38
rs1056021803 Chr.X: 134796846-134796846 on Build GRCh38
rs771599651 Chr.X: 134796593-134796593 on Build GRCh38
rs1032341654 Chr.X: 134796878-134796878 on Build GRCh38
rs1031250170 Chr.X: 134796970-134796970 on Build GRCh38
rs769520509 Chr.X: 134796881-134796881 on Build GRCh38
rs911140872 Chr.X: 134796703-134796703 on Build GRCh38
rs989397097 Chr.X: 134796700-134796700 on Build GRCh38
rs753029567 Chr.X: 134796873-134796873 on Build GRCh38

View Full Product Details