Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134809497-134810046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM122C
Entrez Gene ID
159091
Gene Name
family with sequence similarity 122C
Gene Aliases
-
UniGene
Hs.269127
Gene Chromosome Location
Chr.X: 134796413-134855137 on Build GRCh38
Transcripts
NM_001170779.1 NM_001170780.1 NM_001170781.1 NM_001170782.1 NM_001170783.1 NM_001170784.1 NM_138819.3 XM_005262382.1 XM_005262383.2 XM_005262384.4 XM_005262386.3 XM_005262387.3 XM_006724734.3 XM_006724735.1 XM_006724736.2 XM_011531295.1 XM_011531296.2 XM_011531297.2 XM_011531298.2 XM_011531305.2 XM_011531306.2 XM_017029308.1 XM_017029310.1 XM_017029311.1

SNP Detail

SNP ID Chromosome Location
rs759442084 Chr.X: 134809546-134809547 on Build GRCh38
rs191339413 Chr.X: 134809879-134809879 on Build GRCh38
rs539562032 Chr.X: 134809757-134809757 on Build GRCh38
rs963078681 Chr.X: 134809918-134809918 on Build GRCh38
rs755395352 Chr.X: 134809946-134809946 on Build GRCh38
rs948721365 Chr.X: 134809695-134809695 on Build GRCh38
rs745590679 Chr.X: 134809577-134809577 on Build GRCh38
rs961247852 Chr.X: 134809604-134809604 on Build GRCh38
rs1053183289 Chr.X: 134809886-134809886 on Build GRCh38
rs1042781266 Chr.X: 134809663-134809663 on Build GRCh38
rs917982983 Chr.X: 134809575-134809575 on Build GRCh38
rs949521550 Chr.X: 134809608-134809608 on Build GRCh38
rs980239585 Chr.X: 134809733-134809733 on Build GRCh38
rs1004669659 Chr.X: 134809871-134809871 on Build GRCh38
rs995996658 Chr.X: 134809776-134809776 on Build GRCh38
rs183333227 Chr.X: 134809688-134809688 on Build GRCh38
rs947702831 Chr.X: 134809982-134809982 on Build GRCh38
rs754200044 Chr.X: 134809890-134809890 on Build GRCh38
rs757981845 Chr.X: 134809717-134809717 on Build GRCh38
rs7059630 Chr.X: 134810020-134810020 on Build GRCh38
rs763445836 Chr.X: 134809765-134809765 on Build GRCh38
rs954611214 Chr.X: 134809858-134809858 on Build GRCh38
rs974273549 Chr.X: 134809923-134809923 on Build GRCh38
rs1030192285 Chr.X: 134809811-134809811 on Build GRCh38
rs73568758 Chr.X: 134809607-134809607 on Build GRCh38
rs920192703 Chr.X: 134809983-134809983 on Build GRCh38
rs892050938 Chr.X: 134809939-134809939 on Build GRCh38
rs186922062 Chr.X: 134809859-134809859 on Build GRCh38
rs779653547 Chr.X: 134809793-134809793 on Build GRCh38
rs1015532137 Chr.X: 134809571-134809571 on Build GRCh38
rs900306484 Chr.X: 134809740-134809740 on Build GRCh38
rs751110428 Chr.X: 134809846-134809846 on Build GRCh38
rs34014290 Chr.X: 134810020-134810021 on Build GRCh38
rs771937701 Chr.X: 134809916-134809916 on Build GRCh38
rs543992372 Chr.X: 134809845-134809845 on Build GRCh38

View Full Product Details