Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135021520-135022010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM127C
Entrez Gene ID
441518
Gene Name
family with sequence similarity 127 member C
Gene Aliases
CXX1c, MAR8B
UniGene
Hs.378813
Gene Chromosome Location
Chr.X: 135020504-135022536 on Build GRCh38
Transcripts
NM_001078173.1

SNP Detail

SNP ID Chromosome Location
rs761569843 Chr.X: 135021813-135021813 on Build GRCh38
rs777944597 Chr.X: 135021970-135021970 on Build GRCh38
rs1050311978 Chr.X: 135021910-135021910 on Build GRCh38
rs745354797 Chr.X: 135021576-135021576 on Build GRCh38
rs749997664 Chr.X: 135021826-135021826 on Build GRCh38
rs779123018 Chr.X: 135021895-135021895 on Build GRCh38
rs773465325 Chr.X: 135021713-135021713 on Build GRCh38
rs772385107 Chr.X: 135021886-135021886 on Build GRCh38
rs953117866 Chr.X: 135021671-135021671 on Build GRCh38
rs768996052 Chr.X: 135021649-135021649 on Build GRCh38
rs762855307 Chr.X: 135021745-135021745 on Build GRCh38
rs1040110464 Chr.X: 135021574-135021574 on Build GRCh38
rs756694751 Chr.X: 135021702-135021702 on Build GRCh38
rs1005805929 Chr.X: 135021677-135021677 on Build GRCh38
rs753862347 Chr.X: 135021578-135021593 on Build GRCh38
rs748458060 Chr.X: 135021897-135021897 on Build GRCh38
rs928057817 Chr.X: 135021796-135021796 on Build GRCh38
rs769910446 Chr.X: 135021739-135021739 on Build GRCh38
rs749716107 Chr.X: 135021642-135021642 on Build GRCh38
rs762906531 Chr.X: 135021919-135021919 on Build GRCh38
rs756741600 Chr.X: 135021547-135021547 on Build GRCh38
rs780530094 Chr.X: 135021635-135021635 on Build GRCh38
rs763902167 Chr.X: 135021766-135021766 on Build GRCh38
rs62599865 Chr.X: 135021874-135021874 on Build GRCh38
rs748428579 Chr.X: 135021882-135021882 on Build GRCh38
rs946586101 Chr.X: 135021909-135021909 on Build GRCh38
rs956719866 Chr.X: 135021808-135021808 on Build GRCh38
rs760814351 Chr.X: 135021715-135021715 on Build GRCh38
rs149477038 Chr.X: 135021559-135021559 on Build GRCh38
rs755611864 Chr.X: 135021834-135021834 on Build GRCh38
rs767220394 Chr.X: 135021969-135021969 on Build GRCh38
rs755318319 Chr.X: 135021878-135021878 on Build GRCh38
rs969559925 Chr.X: 135021764-135021764 on Build GRCh38
rs764336023 Chr.X: 135021870-135021870 on Build GRCh38
rs7880514 Chr.X: 135021782-135021782 on Build GRCh38
rs191134459 Chr.X: 135021599-135021599 on Build GRCh38
rs757488260 Chr.X: 135021888-135021892 on Build GRCh38
rs779013151 Chr.X: 135021661-135021661 on Build GRCh38
rs767058504 Chr.X: 135021821-135021821 on Build GRCh38
rs766827867 Chr.X: 135021741-135021741 on Build GRCh38
rs372704415 Chr.X: 135021675-135021675 on Build GRCh38
rs761496733 Chr.X: 135021939-135021939 on Build GRCh38
rs747032694 Chr.X: 135021977-135021977 on Build GRCh38
rs756666795 Chr.X: 135021597-135021597 on Build GRCh38
rs745660674 Chr.X: 135021992-135021992 on Build GRCh38
rs763729185 Chr.X: 135021572-135021575 on Build GRCh38
rs747090263 Chr.X: 135021901-135021901 on Build GRCh38
rs994758847 Chr.X: 135021947-135021947 on Build GRCh38
rs763093530 Chr.X: 135021798-135021799 on Build GRCh38
rs771076737 Chr.X: 135021906-135021906 on Build GRCh38
rs993592045 Chr.X: 135021601-135021601 on Build GRCh38
rs774247910 Chr.X: 135021935-135021935 on Build GRCh38
rs751817155 Chr.X: 135021880-135021883 on Build GRCh38
rs772073241 Chr.X: 135021710-135021710 on Build GRCh38
rs74634293 Chr.X: 135021923-135021923 on Build GRCh38
rs780540158 Chr.X: 135021868-135021868 on Build GRCh38
rs775591843 Chr.X: 135021908-135021908 on Build GRCh38
rs915158852 Chr.X: 135021905-135021905 on Build GRCh38

View Full Product Details