Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 147936272-147936795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMR1
Entrez Gene ID
2332
Gene Name
fragile X mental retardation 1
Gene Aliases
FMRP, FRAXA, POF, POF1
UniGene
Hs.103183
Gene Chromosome Location
Chr.X: 147911951-147951127 on Build GRCh38
Transcripts
NM_001185075.1 NM_001185076.1 NM_001185081.1 NM_001185082.1 NM_002024.5 NR_033699.1 NR_033700.1

SNP Detail

SNP ID Chromosome Location
rs902947990 Chr.X: 147936391-147936391 on Build GRCh38
rs974059679 Chr.X: 147936591-147936591 on Build GRCh38
rs782163090 Chr.X: 147936656-147936656 on Build GRCh38
rs782630605 Chr.X: 147936626-147936626 on Build GRCh38
rs899529633 Chr.X: 147936357-147936357 on Build GRCh38
rs782386085 Chr.X: 147936617-147936617 on Build GRCh38
rs25712 Chr.X: 147936465-147936465 on Build GRCh38
rs782265632 Chr.X: 147936590-147936590 on Build GRCh38
rs25713 Chr.X: 147936754-147936754 on Build GRCh38
rs943992537 Chr.X: 147936336-147936336 on Build GRCh38
rs982884389 Chr.X: 147936701-147936701 on Build GRCh38
rs782004372 Chr.X: 147936686-147936686 on Build GRCh38
rs947018350 Chr.X: 147936381-147936381 on Build GRCh38
rs782232058 Chr.X: 147936642-147936642 on Build GRCh38
rs111870942 Chr.X: 147936446-147936446 on Build GRCh38
rs782530601 Chr.X: 147936341-147936341 on Build GRCh38
rs1031590314 Chr.X: 147936421-147936421 on Build GRCh38
rs947257168 Chr.X: 147936753-147936753 on Build GRCh38
rs121434622 Chr.X: 147936534-147936534 on Build GRCh38
rs782684689 Chr.X: 147936491-147936491 on Build GRCh38
rs943873900 Chr.X: 147936709-147936709 on Build GRCh38
rs782446757 Chr.X: 147936416-147936416 on Build GRCh38
rs372138591 Chr.X: 147936464-147936464 on Build GRCh38
rs782402980 Chr.X: 147936660-147936660 on Build GRCh38
rs921200017 Chr.X: 147936724-147936724 on Build GRCh38
rs896647478 Chr.X: 147936424-147936424 on Build GRCh38
rs782382658 Chr.X: 147936644-147936644 on Build GRCh38
rs886041089 Chr.X: 147936614-147936614 on Build GRCh38
rs782679317 Chr.X: 147936388-147936388 on Build GRCh38
rs29284 Chr.X: 147936410-147936410 on Build GRCh38
rs376794313 Chr.X: 147936473-147936473 on Build GRCh38
rs781857570 Chr.X: 147936454-147936454 on Build GRCh38
rs782644007 Chr.X: 147936418-147936418 on Build GRCh38
rs1039616862 Chr.X: 147936344-147936344 on Build GRCh38
rs1014286057 Chr.X: 147936430-147936430 on Build GRCh38
rs912361545 Chr.X: 147936307-147936307 on Build GRCh38
rs782737667 Chr.X: 147936433-147936434 on Build GRCh38
rs930035984 Chr.X: 147936301-147936301 on Build GRCh38
rs1023827356 Chr.X: 147936442-147936442 on Build GRCh38
rs25714 Chr.X: 147936627-147936627 on Build GRCh38
rs951551613 Chr.X: 147936694-147936694 on Build GRCh38
rs912419608 Chr.X: 147936704-147936704 on Build GRCh38
rs983032282 Chr.X: 147936304-147936304 on Build GRCh38

View Full Product Details