Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201204802-201205340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFN1
Entrez Gene ID
91156
Gene Name
immunoglobulin-like and fibronectin type III domain containing 1
Gene Aliases
EEF1A2BP1
UniGene
Hs.519024
Gene Chromosome Location
Chr.1: 201190780-201228952 on Build GRCh38
Transcripts
NM_001164586.1 XM_005245580.3 XM_006711618.3 XM_011510120.2 XM_011510121.2 XM_017002787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5532584 c.1142C>T p.S381L Chr.1: 201205307-201205307 on Build GRCh38
4764339 c.1018G>A p.A340T Chr.1: 201205183-201205183 on Build GRCh38
7352685 c.1019C>G p.A340G Chr.1: 201205184-201205184 on Build GRCh38
5505706 c.1002C>A p.S334R Chr.1: 201205167-201205167 on Build GRCh38
6237364 c.920T>C p.I307T Chr.1: 201205085-201205085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987090706 Chr.1: 201204968-201204968 on Build GRCh38
rs774621227 Chr.1: 201205196-201205196 on Build GRCh38
rs777338105 Chr.1: 201205279-201205279 on Build GRCh38
rs777473357 Chr.1: 201205089-201205089 on Build GRCh38
rs746556247 Chr.1: 201205094-201205094 on Build GRCh38
rs370332278 Chr.1: 201205213-201205213 on Build GRCh38
rs867065319 Chr.1: 201205188-201205188 on Build GRCh38
rs575197790 Chr.1: 201205108-201205108 on Build GRCh38
rs977548372 Chr.1: 201205262-201205262 on Build GRCh38
rs531557599 Chr.1: 201205295-201205295 on Build GRCh38
rs868736745 Chr.1: 201205307-201205307 on Build GRCh38
rs12725611 Chr.1: 201205076-201205076 on Build GRCh38
rs367631324 Chr.1: 201205248-201205248 on Build GRCh38
rs890775050 Chr.1: 201205124-201205124 on Build GRCh38
rs764837921 Chr.1: 201205007-201205007 on Build GRCh38
rs553969713 Chr.1: 201205035-201205035 on Build GRCh38
rs1050204753 Chr.1: 201205308-201205308 on Build GRCh38
rs902299967 Chr.1: 201205317-201205317 on Build GRCh38
rs1042599104 Chr.1: 201205202-201205202 on Build GRCh38
rs949494971 Chr.1: 201204885-201204885 on Build GRCh38
rs372183161 Chr.1: 201205151-201205151 on Build GRCh38
rs540024608 Chr.1: 201205201-201205201 on Build GRCh38
rs1000805061 Chr.1: 201205064-201205064 on Build GRCh38
rs756751848 Chr.1: 201205283-201205283 on Build GRCh38
rs753551170 Chr.1: 201204900-201204900 on Build GRCh38
rs188913362 Chr.1: 201204834-201204834 on Build GRCh38
rs571886984 Chr.1: 201204913-201204913 on Build GRCh38
rs61743109 Chr.1: 201205230-201205230 on Build GRCh38
rs142181533 Chr.1: 201205242-201205242 on Build GRCh38
rs61749969 Chr.1: 201205184-201205184 on Build GRCh38
rs1007991710 Chr.1: 201205132-201205132 on Build GRCh38
rs1050256250 Chr.1: 201205172-201205172 on Build GRCh38
rs959338679 Chr.1: 201204845-201204845 on Build GRCh38
rs377463155 Chr.1: 201205077-201205077 on Build GRCh38
rs1042095047 Chr.1: 201205091-201205091 on Build GRCh38
rs199966486 Chr.1: 201205275-201205275 on Build GRCh38
rs201805154 Chr.1: 201205140-201205140 on Build GRCh38
rs1027911574 Chr.1: 201205212-201205212 on Build GRCh38
rs1024987817 Chr.1: 201205206-201205206 on Build GRCh38
rs973693490 Chr.1: 201205042-201205042 on Build GRCh38
rs971111623 Chr.1: 201205209-201205209 on Build GRCh38
rs996434828 Chr.1: 201204921-201204921 on Build GRCh38
rs781592987 Chr.1: 201205112-201205112 on Build GRCh38
rs184463517 Chr.1: 201204830-201204830 on Build GRCh38
rs371722890 Chr.1: 201205282-201205282 on Build GRCh38
rs942163519 Chr.1: 201205101-201205101 on Build GRCh38
rs999475089 Chr.1: 201204879-201204879 on Build GRCh38
rs866843664 Chr.1: 201205086-201205086 on Build GRCh38
rs902428652 Chr.1: 201204860-201204860 on Build GRCh38
rs1008675472 Chr.1: 201205183-201205183 on Build GRCh38
rs1014614965 Chr.1: 201204977-201204977 on Build GRCh38
rs193152194 Chr.1: 201204920-201204920 on Build GRCh38
rs770358996 Chr.1: 201205318-201205318 on Build GRCh38
rs1047914834 Chr.1: 201204888-201204888 on Build GRCh38
rs962595031 Chr.1: 201205041-201205041 on Build GRCh38
rs546783067 Chr.1: 201204829-201204829 on Build GRCh38
rs932282727 Chr.1: 201205249-201205249 on Build GRCh38
rs1041753596 Chr.1: 201204849-201204849 on Build GRCh38
rs1020816289 Chr.1: 201204974-201204974 on Build GRCh38
rs185501571 Chr.1: 201204982-201204982 on Build GRCh38
rs908068518 Chr.1: 201205096-201205096 on Build GRCh38
rs539218791 Chr.1: 201204955-201204955 on Build GRCh38
rs534100902 Chr.1: 201204993-201204993 on Build GRCh38
rs780702463 Chr.1: 201205319-201205319 on Build GRCh38
rs141169586 Chr.1: 201204914-201204914 on Build GRCh38
rs752551820 Chr.1: 201205274-201205274 on Build GRCh38
rs922221166 Chr.1: 201205110-201205110 on Build GRCh38
rs577682608 Chr.1: 201205195-201205195 on Build GRCh38
rs746395288 Chr.1: 201205300-201205300 on Build GRCh38
rs893602784 Chr.1: 201205142-201205142 on Build GRCh38
rs935217493 Chr.1: 201205113-201205113 on Build GRCh38
rs112649677 Chr.1: 201204883-201204883 on Build GRCh38
rs942299196 Chr.1: 201204905-201204905 on Build GRCh38
rs562864069 Chr.1: 201205261-201205261 on Build GRCh38
rs536759117 Chr.1: 201205175-201205175 on Build GRCh38
rs970864704 Chr.1: 201205090-201205090 on Build GRCh38
rs893655365 Chr.1: 201204884-201204884 on Build GRCh38
rs145066841 Chr.1: 201204936-201204936 on Build GRCh38
rs898170422 Chr.1: 201204917-201204917 on Build GRCh38
rs890919282 Chr.1: 201204964-201204964 on Build GRCh38
rs991535544 Chr.1: 201205085-201205085 on Build GRCh38
rs931939801 Chr.1: 201205302-201205302 on Build GRCh38
rs11584104 Chr.1: 201205087-201205087 on Build GRCh38

View Full Product Details