Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201205229-201205738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFN1
Entrez Gene ID
91156
Gene Name
immunoglobulin-like and fibronectin type III domain containing 1
Gene Aliases
EEF1A2BP1
UniGene
Hs.519024
Gene Chromosome Location
Chr.1: 201190780-201228952 on Build GRCh38
Transcripts
NM_001164586.1 XM_005245580.3 XM_006711618.3 XM_011510120.2 XM_011510121.2 XM_017002787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5532584 c.1142C>T p.S381L Chr.1: 201205307-201205307 on Build GRCh38
6846676 c.1084G>T p.G362W Chr.1: 201205249-201205249 on Build GRCh38
4458266 c.1078C>T p.P360S Chr.1: 201205243-201205243 on Build GRCh38
5731353 c.1076C>T p.S359F Chr.1: 201205241-201205241 on Build GRCh38
2212020 c.1171G>A p.A391T Chr.1: 201205336-201205336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764019158 Chr.1: 201205447-201205447 on Build GRCh38
rs777338105 Chr.1: 201205279-201205279 on Build GRCh38
rs114767524 Chr.1: 201205551-201205551 on Build GRCh38
rs553997517 Chr.1: 201205702-201205702 on Build GRCh38
rs531557599 Chr.1: 201205295-201205295 on Build GRCh38
rs868736745 Chr.1: 201205307-201205307 on Build GRCh38
rs977548372 Chr.1: 201205262-201205262 on Build GRCh38
rs557755793 Chr.1: 201205689-201205689 on Build GRCh38
rs571457395 Chr.1: 201205595-201205595 on Build GRCh38
rs576389837 Chr.1: 201205336-201205336 on Build GRCh38
rs1013204897 Chr.1: 201205427-201205427 on Build GRCh38
rs1050204753 Chr.1: 201205308-201205308 on Build GRCh38
rs536907891 Chr.1: 201205453-201205453 on Build GRCh38
rs902299967 Chr.1: 201205317-201205317 on Build GRCh38
rs977773878 Chr.1: 201205566-201205566 on Build GRCh38
rs906482006 Chr.1: 201205683-201205683 on Build GRCh38
rs1029131730 Chr.1: 201205516-201205516 on Build GRCh38
rs756751848 Chr.1: 201205283-201205283 on Build GRCh38
rs374608026 Chr.1: 201205654-201205654 on Build GRCh38
rs138354288 Chr.1: 201205690-201205690 on Build GRCh38
rs573925950 Chr.1: 201205706-201205706 on Build GRCh38
rs547571730 Chr.1: 201205479-201205479 on Build GRCh38
rs776798944 Chr.1: 201205699-201205699 on Build GRCh38
rs571625451 Chr.1: 201205392-201205392 on Build GRCh38
rs199966486 Chr.1: 201205275-201205275 on Build GRCh38
rs1036350450 Chr.1: 201205692-201205692 on Build GRCh38
rs1027361218 Chr.1: 201205400-201205400 on Build GRCh38
rs930409672 Chr.1: 201205670-201205670 on Build GRCh38
rs772056865 Chr.1: 201205385-201205385 on Build GRCh38
rs556394124 Chr.1: 201205554-201205554 on Build GRCh38
rs990164525 Chr.1: 201205553-201205553 on Build GRCh38
rs371722890 Chr.1: 201205282-201205282 on Build GRCh38
rs946315212 Chr.1: 201205565-201205565 on Build GRCh38
rs1014876519 Chr.1: 201205467-201205467 on Build GRCh38
rs748351879 Chr.1: 201205365-201205365 on Build GRCh38
rs150450722 Chr.1: 201205331-201205331 on Build GRCh38
rs4488061 Chr.1: 201205680-201205680 on Build GRCh38
rs931870964 Chr.1: 201205564-201205564 on Build GRCh38
rs1053508165 Chr.1: 201205643-201205643 on Build GRCh38
rs770358996 Chr.1: 201205318-201205318 on Build GRCh38
rs936417065 Chr.1: 201205606-201205606 on Build GRCh38
rs780229965 Chr.1: 201205335-201205335 on Build GRCh38
rs973447688 Chr.1: 201205489-201205489 on Build GRCh38
rs4396148 Chr.1: 201205694-201205694 on Build GRCh38
rs986029111 Chr.1: 201205567-201205567 on Build GRCh38
rs932282727 Chr.1: 201205249-201205249 on Build GRCh38
rs1029454467 Chr.1: 201205707-201205707 on Build GRCh38
rs774466143 Chr.1: 201205357-201205357 on Build GRCh38
rs749295099 Chr.1: 201205383-201205383 on Build GRCh38
rs780702463 Chr.1: 201205319-201205319 on Build GRCh38
rs115347169 Chr.1: 201205676-201205676 on Build GRCh38
rs769270207 Chr.1: 201205503-201205503 on Build GRCh38
rs752551820 Chr.1: 201205274-201205274 on Build GRCh38
rs746395288 Chr.1: 201205300-201205300 on Build GRCh38
rs772565230 Chr.1: 201205571-201205571 on Build GRCh38
rs562864069 Chr.1: 201205261-201205261 on Build GRCh38
rs773676086 Chr.1: 201205658-201205658 on Build GRCh38
rs527816935 Chr.1: 201205410-201205410 on Build GRCh38
rs977871627 Chr.1: 201205487-201205487 on Build GRCh38
rs1012182638 Chr.1: 201205374-201205374 on Build GRCh38
rs931939801 Chr.1: 201205302-201205302 on Build GRCh38
rs1031764776 Chr.1: 201205519-201205519 on Build GRCh38

View Full Product Details