Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206596958-206597498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2D
Entrez Gene ID
1939
Gene Name
eukaryotic translation initiation factor 2D
Gene Aliases
HCA56, LGTN
UniGene
Hs.497581
Gene Chromosome Location
Chr.1: 206572485-206612551 on Build GRCh38
Transcripts
NM_001201478.1 NM_006893.2 XM_005277509.3 XM_006711196.3 XM_011509257.2 XM_017000530.1 XM_017000531.1 XM_017000532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3803671 c.1326C>T p.C442C Chr.1: 206597162-206597162 on Build GRCh38
1180409 c.1314C>A p.I438I Chr.1: 206597174-206597174 on Build GRCh38
5053016 c.1357A>G p.M453V Chr.1: 206597131-206597131 on Build GRCh38
1180408 c.1315C>A p.L439I Chr.1: 206597173-206597173 on Build GRCh38
4448861 c.1378_1379delCT p.L460fs*32 Chr.1: 206597109-206597110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048191052 Chr.1: 206597253-206597253 on Build GRCh38
rs372538151 Chr.1: 206597212-206597212 on Build GRCh38
rs782030079 Chr.1: 206597094-206597094 on Build GRCh38
rs201352069 Chr.1: 206597194-206597194 on Build GRCh38
rs782121005 Chr.1: 206597128-206597128 on Build GRCh38
rs1020360925 Chr.1: 206597382-206597382 on Build GRCh38
rs782068936 Chr.1: 206597057-206597057 on Build GRCh38
rs1040278782 Chr.1: 206597275-206597275 on Build GRCh38
rs782813126 Chr.1: 206597135-206597135 on Build GRCh38
rs567680991 Chr.1: 206597118-206597118 on Build GRCh38
rs547888785 Chr.1: 206597102-206597102 on Build GRCh38
rs538557900 Chr.1: 206597229-206597229 on Build GRCh38
rs554534144 Chr.1: 206597389-206597389 on Build GRCh38
rs782097617 Chr.1: 206597116-206597116 on Build GRCh38
rs782523276 Chr.1: 206597166-206597166 on Build GRCh38
rs782202453 Chr.1: 206597064-206597064 on Build GRCh38
rs375984315 Chr.1: 206597211-206597211 on Build GRCh38
rs533906408 Chr.1: 206597095-206597095 on Build GRCh38
rs782183155 Chr.1: 206597203-206597203 on Build GRCh38
rs113368228 Chr.1: 206597111-206597111 on Build GRCh38
rs886476770 Chr.1: 206597262-206597262 on Build GRCh38
rs782372469 Chr.1: 206597090-206597090 on Build GRCh38
rs369017810 Chr.1: 206597297-206597297 on Build GRCh38
rs150077489 Chr.1: 206597119-206597119 on Build GRCh38
rs371484311 Chr.1: 206597062-206597062 on Build GRCh38
rs372760750 Chr.1: 206597068-206597068 on Build GRCh38
rs781842982 Chr.1: 206597160-206597160 on Build GRCh38
rs373669376 Chr.1: 206597224-206597224 on Build GRCh38
rs186861196 Chr.1: 206597360-206597360 on Build GRCh38
rs920650661 Chr.1: 206597185-206597185 on Build GRCh38
rs111533231 Chr.1: 206597279-206597279 on Build GRCh38
rs536755773 Chr.1: 206597431-206597431 on Build GRCh38
rs782322660 Chr.1: 206597096-206597096 on Build GRCh38
rs781956564 Chr.1: 206597206-206597206 on Build GRCh38
rs782113463 Chr.1: 206597169-206597169 on Build GRCh38
rs201324890 Chr.1: 206597252-206597252 on Build GRCh38
rs377303536 Chr.1: 206597142-206597142 on Build GRCh38
rs112313312 Chr.1: 206597381-206597381 on Build GRCh38
rs938141826 Chr.1: 206597072-206597072 on Build GRCh38
rs782531373 Chr.1: 206597176-206597176 on Build GRCh38
rs996013270 Chr.1: 206596996-206596996 on Build GRCh38
rs782452874 Chr.1: 206597143-206597143 on Build GRCh38
rs574490109 Chr.1: 206597414-206597414 on Build GRCh38
rs782048813 Chr.1: 206597415-206597415 on Build GRCh38
rs1010024867 Chr.1: 206597361-206597361 on Build GRCh38
rs555979007 Chr.1: 206597171-206597171 on Build GRCh38
rs782156040 Chr.1: 206597234-206597234 on Build GRCh38
rs995983693 Chr.1: 206597304-206597304 on Build GRCh38
rs782257577 Chr.1: 206597087-206597087 on Build GRCh38
rs572031566 Chr.1: 206597268-206597268 on Build GRCh38
rs1032876263 Chr.1: 206597317-206597317 on Build GRCh38
rs782603266 Chr.1: 206597078-206597078 on Build GRCh38
rs142664077 Chr.1: 206597157-206597157 on Build GRCh38
rs559986692 Chr.1: 206597025-206597025 on Build GRCh38
rs781925699 Chr.1: 206597015-206597015 on Build GRCh38
rs782352335 Chr.1: 206597205-206597205 on Build GRCh38

View Full Product Details