Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209609760-209610261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1G
Entrez Gene ID
57172
Gene Name
calcium/calmodulin dependent protein kinase IG
Gene Aliases
CLICK3, CLICKIII, VWS1, dJ272L16.1
UniGene
Hs.199068
Gene Chromosome Location
Chr.1: 209583700-209613939 on Build GRCh38
Transcripts
NM_020439.2 XM_017001866.1 XM_017001867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359333 c.775G>C p.E259Q Chr.1: 209609877-209609877 on Build GRCh38
319219 c.784C>G p.P262A Chr.1: 209609886-209609886 on Build GRCh38
5845328 c.796T>C p.Y266H Chr.1: 209609898-209609898 on Build GRCh38
358325 c.820C>T p.H274Y Chr.1: 209609922-209609922 on Build GRCh38
5379509 c.804T>A p.C268* Chr.1: 209609906-209609906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005214420 Chr.1: 209610060-209610060 on Build GRCh38
rs777093190 Chr.1: 209609827-209609827 on Build GRCh38
rs146703783 Chr.1: 209609784-209609784 on Build GRCh38
rs540439163 Chr.1: 209609801-209609801 on Build GRCh38
rs753238483 Chr.1: 209609959-209609959 on Build GRCh38
rs980743552 Chr.1: 209609902-209609902 on Build GRCh38
rs139313701 Chr.1: 209609916-209609916 on Build GRCh38
rs754075910 Chr.1: 209609955-209609955 on Build GRCh38
rs747374065 Chr.1: 209609969-209609969 on Build GRCh38
rs1001943619 Chr.1: 209610219-209610219 on Build GRCh38
rs958008588 Chr.1: 209610216-209610216 on Build GRCh38
rs573878918 Chr.1: 209609824-209609824 on Build GRCh38
rs778900198 Chr.1: 209609958-209609958 on Build GRCh38
rs771323567 Chr.1: 209609970-209609970 on Build GRCh38
rs774655486 Chr.1: 209609837-209609838 on Build GRCh38
rs149187822 Chr.1: 209609891-209609891 on Build GRCh38
rs769369395 Chr.1: 209609896-209609896 on Build GRCh38
rs761635113 Chr.1: 209609946-209609946 on Build GRCh38
rs755043860 Chr.1: 209609852-209609852 on Build GRCh38
rs55853242 Chr.1: 209609985-209609985 on Build GRCh38
rs775955322 Chr.1: 209609805-209609805 on Build GRCh38
rs762170245 Chr.1: 209609847-209609847 on Build GRCh38
rs370981947 Chr.1: 209610159-209610159 on Build GRCh38
rs368478956 Chr.1: 209609956-209609956 on Build GRCh38
rs986813124 Chr.1: 209610059-209610059 on Build GRCh38
rs904936142 Chr.1: 209610178-209610178 on Build GRCh38
rs576435401 Chr.1: 209610164-209610164 on Build GRCh38
rs562853883 Chr.1: 209609932-209609932 on Build GRCh38
rs546339380 Chr.1: 209610012-209610012 on Build GRCh38
rs200728472 Chr.1: 209609954-209609954 on Build GRCh38
rs144647481 Chr.1: 209609780-209609780 on Build GRCh38
rs1052744397 Chr.1: 209610167-209610167 on Build GRCh38
rs914111474 Chr.1: 209610228-209610228 on Build GRCh38
rs974768691 Chr.1: 209610032-209610032 on Build GRCh38
rs532267705 Chr.1: 209610089-209610089 on Build GRCh38
rs892395326 Chr.1: 209610171-209610171 on Build GRCh38
rs367680723 Chr.1: 209609825-209609825 on Build GRCh38
rs759895279 Chr.1: 209609846-209609847 on Build GRCh38
rs1015447502 Chr.1: 209610064-209610064 on Build GRCh38
rs918847486 Chr.1: 209610041-209610041 on Build GRCh38
rs776318186 Chr.1: 209609894-209609894 on Build GRCh38
rs201972342 Chr.1: 209609882-209609882 on Build GRCh38
rs375139559 Chr.1: 209609968-209609968 on Build GRCh38
rs903720509 Chr.1: 209610129-209610129 on Build GRCh38
rs1046069370 Chr.1: 209610176-209610176 on Build GRCh38
rs544223522 Chr.1: 209610102-209610102 on Build GRCh38
rs1014474065 Chr.1: 209610103-209610103 on Build GRCh38
rs35561962 Chr.1: 209609877-209609877 on Build GRCh38
rs762942646 Chr.1: 209609915-209609915 on Build GRCh38
rs780296684 Chr.1: 209609835-209609835 on Build GRCh38
rs1051143395 Chr.1: 209609802-209609802 on Build GRCh38
rs765307107 Chr.1: 209609814-209609814 on Build GRCh38
rs759065954 Chr.1: 209609806-209609806 on Build GRCh38
rs562690709 Chr.1: 209610104-209610104 on Build GRCh38
rs962984411 Chr.1: 209610026-209610026 on Build GRCh38
rs56379151 Chr.1: 209609888-209609888 on Build GRCh38
rs535846107 Chr.1: 209610125-209610125 on Build GRCh38
rs759185718 Chr.1: 209609895-209609895 on Build GRCh38
rs752666731 Chr.1: 209609818-209609818 on Build GRCh38
rs770539963 Chr.1: 209609892-209609892 on Build GRCh38
rs57823603 Chr.1: 209610042-209610042 on Build GRCh38
rs775571475 Chr.1: 209609897-209609897 on Build GRCh38
rs188908583 Chr.1: 209609828-209609828 on Build GRCh38
rs1015498515 Chr.1: 209609844-209609844 on Build GRCh38
rs868357603 Chr.1: 209610027-209610027 on Build GRCh38
rs199573100 Chr.1: 209609974-209609974 on Build GRCh38
rs554566758 Chr.1: 209610172-209610172 on Build GRCh38
rs143363832 Chr.1: 209609964-209609964 on Build GRCh38
rs375640747 Chr.1: 209610082-209610093 on Build GRCh38

View Full Product Details