Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 219979899-219980399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPRS
Entrez Gene ID
2058
Gene Name
glutamyl-prolyl-tRNA synthetase
Gene Aliases
EARS, GLUPRORS, PARS, PIG32, QARS, QPRS
UniGene
Hs.497788
Gene Chromosome Location
Chr.1: 219968600-220046658 on Build GRCh38
Transcripts
NM_004446.2 XM_017000614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3360676 c.3604G>A p.A1202T Chr.1: 219980192-219980192 on Build GRCh38
310911 c.3578A>G p.Y1193C Chr.1: 219980218-219980218 on Build GRCh38
5951466 c.3675A>C p.I1225I Chr.1: 219980121-219980121 on Build GRCh38
3934490 c.3633G>A p.T1211T Chr.1: 219980163-219980163 on Build GRCh38
1668487 c.3679G>T p.A1227S Chr.1: 219980117-219980117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3835619 Chr.1: 219979990-219979991 on Build GRCh38
rs775396857 Chr.1: 219980093-219980093 on Build GRCh38
rs928776541 Chr.1: 219980296-219980296 on Build GRCh38
rs774011588 Chr.1: 219980057-219980058 on Build GRCh38
rs778513654 Chr.1: 219980057-219980057 on Build GRCh38
rs747787523 Chr.1: 219980215-219980215 on Build GRCh38
rs747413517 Chr.1: 219980060-219980060 on Build GRCh38
rs78464168 Chr.1: 219979993-219979993 on Build GRCh38
rs559445163 Chr.1: 219979986-219979986 on Build GRCh38
rs751802894 Chr.1: 219980121-219980121 on Build GRCh38
rs567407362 Chr.1: 219980342-219980342 on Build GRCh38
rs985832281 Chr.1: 219979944-219979944 on Build GRCh38
rs550854491 Chr.1: 219980283-219980283 on Build GRCh38
rs963166400 Chr.1: 219979965-219979965 on Build GRCh38
rs754451619 Chr.1: 219980204-219980204 on Build GRCh38
rs950430230 Chr.1: 219980052-219980052 on Build GRCh38
rs749001631 Chr.1: 219980042-219980042 on Build GRCh38
rs768840036 Chr.1: 219980281-219980281 on Build GRCh38
rs763262450 Chr.1: 219980101-219980101 on Build GRCh38
rs571460083 Chr.1: 219980134-219980134 on Build GRCh38
rs112749480 Chr.1: 219980146-219980146 on Build GRCh38
rs890056202 Chr.1: 219980335-219980335 on Build GRCh38
rs386369689 Chr.1: 219979991-219979992 on Build GRCh38
rs1053444135 Chr.1: 219979950-219979950 on Build GRCh38
rs981926756 Chr.1: 219980237-219980237 on Build GRCh38
rs763168857 Chr.1: 219980271-219980271 on Build GRCh38
rs532166962 Chr.1: 219980005-219980005 on Build GRCh38
rs552975173 Chr.1: 219980374-219980374 on Build GRCh38
rs964514595 Chr.1: 219980006-219980006 on Build GRCh38
rs746422231 Chr.1: 219980232-219980232 on Build GRCh38
rs919129762 Chr.1: 219979990-219979990 on Build GRCh38
rs762055553 Chr.1: 219980285-219980285 on Build GRCh38
rs768247332 Chr.1: 219980051-219980051 on Build GRCh38
rs868402451 Chr.1: 219980084-219980084 on Build GRCh38
rs901380368 Chr.1: 219980231-219980231 on Build GRCh38
rs117101042 Chr.1: 219980061-219980061 on Build GRCh38
rs781345508 Chr.1: 219980228-219980228 on Build GRCh38
rs759903052 Chr.1: 219980067-219980067 on Build GRCh38
rs926623662 Chr.1: 219980024-219980024 on Build GRCh38
rs146522175 Chr.1: 219980348-219980348 on Build GRCh38
rs770155747 Chr.1: 219980233-219980233 on Build GRCh38
rs1028508388 Chr.1: 219980325-219980325 on Build GRCh38
rs34917248 Chr.1: 219979992-219979993 on Build GRCh38
rs756764132 Chr.1: 219980164-219980164 on Build GRCh38
rs997185880 Chr.1: 219980036-219980036 on Build GRCh38
rs764462607 Chr.1: 219980115-219980115 on Build GRCh38
rs867470942 Chr.1: 219980186-219980186 on Build GRCh38
rs953187357 Chr.1: 219979928-219979928 on Build GRCh38
rs373651698 Chr.1: 219980037-219980037 on Build GRCh38
rs934642236 Chr.1: 219980359-219980359 on Build GRCh38
rs7537559 Chr.1: 219979964-219979964 on Build GRCh38
rs145176480 Chr.1: 219980163-219980163 on Build GRCh38
rs546954085 Chr.1: 219980314-219980314 on Build GRCh38
rs761754930 Chr.1: 219980132-219980132 on Build GRCh38
rs868611544 Chr.1: 219979996-219979996 on Build GRCh38
rs908773497 Chr.1: 219980062-219980062 on Build GRCh38
rs771300035 Chr.1: 219980219-219980219 on Build GRCh38
rs750375315 Chr.1: 219980145-219980145 on Build GRCh38
rs776046358 Chr.1: 219980238-219980238 on Build GRCh38
rs753337644 Chr.1: 219980199-219980199 on Build GRCh38
rs778601693 Chr.1: 219980209-219980209 on Build GRCh38
rs370984363 Chr.1: 219980081-219980081 on Build GRCh38

View Full Product Details