Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 224159644-224160139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO28
Entrez Gene ID
23219
Gene Name
F-box protein 28
Gene Aliases
CENP-30, Fbx28
UniGene
Hs.64691
Gene Chromosome Location
Chr.1: 224114087-224162047 on Build GRCh38
Transcripts
NM_001136115.2 NM_015176.3 NR_049764.1

SNP Detail

SNP ID Chromosome Location
rs531224400 Chr.1: 224160094-224160094 on Build GRCh38
rs1036267397 Chr.1: 224159982-224159982 on Build GRCh38
rs966367984 Chr.1: 224160046-224160046 on Build GRCh38
rs550470053 Chr.1: 224160104-224160104 on Build GRCh38
rs11804140 Chr.1: 224159689-224159689 on Build GRCh38
rs991880347 Chr.1: 224159693-224159693 on Build GRCh38
rs1048693400 Chr.1: 224159950-224159950 on Build GRCh38
rs955266919 Chr.1: 224160113-224160113 on Build GRCh38
rs1016667967 Chr.1: 224160061-224160062 on Build GRCh38
rs959125435 Chr.1: 224159986-224159986 on Build GRCh38
rs562457995 Chr.1: 224159751-224159751 on Build GRCh38
rs932500809 Chr.1: 224159768-224159768 on Build GRCh38
rs59738860 Chr.1: 224159952-224159952 on Build GRCh38
rs1005336434 Chr.1: 224159990-224159990 on Build GRCh38
rs944116991 Chr.1: 224159827-224159827 on Build GRCh38
rs891813809 Chr.1: 224159867-224159867 on Build GRCh38
rs554992797 Chr.1: 224159724-224159724 on Build GRCh38
rs142832248 Chr.1: 224159777-224159777 on Build GRCh38
rs960523093 Chr.1: 224160018-224160018 on Build GRCh38
rs532932931 Chr.1: 224159762-224159762 on Build GRCh38
rs922544211 Chr.1: 224159795-224159795 on Build GRCh38
rs145695250 Chr.1: 224159678-224159678 on Build GRCh38
rs560901650 Chr.1: 224160066-224160066 on Build GRCh38
rs551609138 Chr.1: 224159887-224159887 on Build GRCh38
rs933825406 Chr.1: 224159806-224159806 on Build GRCh38
rs1010235388 Chr.1: 224159873-224159873 on Build GRCh38
rs973087077 Chr.1: 224159778-224159778 on Build GRCh38
rs770284547 Chr.1: 224159989-224159989 on Build GRCh38
rs897165119 Chr.1: 224159897-224159897 on Build GRCh38
rs755983217 Chr.1: 224160093-224160093 on Build GRCh38
rs912670496 Chr.1: 224159826-224159826 on Build GRCh38
rs148933112 Chr.1: 224159749-224159749 on Build GRCh38
rs907419162 Chr.1: 224159921-224159921 on Build GRCh38
rs780845967 Chr.1: 224159719-224159720 on Build GRCh38
rs527429039 Chr.1: 224159851-224159851 on Build GRCh38
rs769513455 Chr.1: 224159696-224159696 on Build GRCh38
rs11804144 Chr.1: 224159731-224159731 on Build GRCh38
rs147740350 Chr.1: 224159818-224159818 on Build GRCh38
rs774280582 Chr.1: 224160065-224160065 on Build GRCh38
rs1050011845 Chr.1: 224159782-224159782 on Build GRCh38
rs1013763048 Chr.1: 224160024-224160024 on Build GRCh38
rs191747171 Chr.1: 224159670-224159670 on Build GRCh38
rs913564634 Chr.1: 224159721-224159721 on Build GRCh38
rs549295970 Chr.1: 224159894-224159895 on Build GRCh38
rs1049509904 Chr.1: 224159808-224159808 on Build GRCh38
rs140734908 Chr.1: 224160048-224160050 on Build GRCh38
rs1037069382 Chr.1: 224159857-224159857 on Build GRCh38
rs762867835 Chr.1: 224159821-224159821 on Build GRCh38
rs995465449 Chr.1: 224159914-224159914 on Build GRCh38
rs1020621955 Chr.1: 224160028-224160028 on Build GRCh38
rs979321174 Chr.1: 224159743-224159743 on Build GRCh38
rs1001735506 Chr.1: 224159968-224159968 on Build GRCh38

View Full Product Details