Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236718816-236719329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN2
Entrez Gene ID
88
Gene Name
actinin alpha 2
Gene Aliases
CMD1AA, CMH23
UniGene
Hs.498178
Gene Chromosome Location
Chr.1: 236686454-236764631 on Build GRCh38
Transcripts
NM_001103.3 NM_001278343.1 NM_001278344.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4890674 c.358G>T p.E120* Chr.1: 236719010-236719010 on Build GRCh38
5640601 c.318C>A p.Y106* Chr.1: 236718970-236718970 on Build GRCh38
6541998 c.262G>T p.D88Y Chr.1: 236718914-236718914 on Build GRCh38
5892514 c.260C>T p.P87L Chr.1: 236718912-236718912 on Build GRCh38
6898041 c.253C>T p.P85S Chr.1: 236718905-236718905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569950571 Chr.1: 236719060-236719060 on Build GRCh38
rs555617678 Chr.1: 236719075-236719075 on Build GRCh38
rs778513501 Chr.1: 236718842-236718842 on Build GRCh38
rs374362248 Chr.1: 236719031-236719031 on Build GRCh38
rs770193305 Chr.1: 236719247-236719247 on Build GRCh38
rs1044929547 Chr.1: 236719081-236719081 on Build GRCh38
rs115970611 Chr.1: 236719268-236719268 on Build GRCh38
rs780383236 Chr.1: 236719059-236719059 on Build GRCh38
rs750239412 Chr.1: 236718866-236718866 on Build GRCh38
rs372584949 Chr.1: 236719052-236719052 on Build GRCh38
rs908976310 Chr.1: 236719155-236719155 on Build GRCh38
rs546541463 Chr.1: 236719292-236719292 on Build GRCh38
rs397516579 Chr.1: 236718995-236718995 on Build GRCh38
rs965620689 Chr.1: 236719140-236719140 on Build GRCh38
rs938348281 Chr.1: 236719248-236719248 on Build GRCh38
rs794728960 Chr.1: 236719005-236719005 on Build GRCh38
rs1031641418 Chr.1: 236719139-236719139 on Build GRCh38
rs998476148 Chr.1: 236719149-236719149 on Build GRCh38
rs761746349 Chr.1: 236719112-236719112 on Build GRCh38
rs772390012 Chr.1: 236718918-236718918 on Build GRCh38
rs187232670 Chr.1: 236719250-236719250 on Build GRCh38
rs951603467 Chr.1: 236718863-236718863 on Build GRCh38
rs727502886 Chr.1: 236719007-236719007 on Build GRCh38
rs147527370 Chr.1: 236718849-236718849 on Build GRCh38
rs946683716 Chr.1: 236719286-236719286 on Build GRCh38
rs149871432 Chr.1: 236718953-236718953 on Build GRCh38
rs532583106 Chr.1: 236719113-236719113 on Build GRCh38
rs199665859 Chr.1: 236719200-236719200 on Build GRCh38
rs749041916 Chr.1: 236718880-236718880 on Build GRCh38
rs915124748 Chr.1: 236719276-236719276 on Build GRCh38
rs918885561 Chr.1: 236719238-236719238 on Build GRCh38
rs770703958 Chr.1: 236718891-236718891 on Build GRCh38
rs727502885 Chr.1: 236718976-236718976 on Build GRCh38
rs1012737816 Chr.1: 236719095-236719095 on Build GRCh38
rs553114211 Chr.1: 236719122-236719122 on Build GRCh38
rs369629689 Chr.1: 236718917-236718917 on Build GRCh38
rs756072126 Chr.1: 236718871-236718871 on Build GRCh38
rs1044912228 Chr.1: 236719290-236719290 on Build GRCh38
rs753872930 Chr.1: 236719037-236719037 on Build GRCh38
rs780724098 Chr.1: 236718854-236718854 on Build GRCh38
rs953126488 Chr.1: 236719220-236719220 on Build GRCh38
rs759787554 Chr.1: 236719027-236719027 on Build GRCh38
rs148628141 Chr.1: 236718983-236718983 on Build GRCh38
rs376981679 Chr.1: 236718901-236718901 on Build GRCh38
rs748210204 Chr.1: 236719236-236719236 on Build GRCh38
rs777728987 Chr.1: 236718874-236718874 on Build GRCh38
rs777124854 Chr.1: 236718930-236718930 on Build GRCh38
rs142899258 Chr.1: 236719158-236719158 on Build GRCh38
rs894353081 Chr.1: 236719088-236719088 on Build GRCh38
rs984289322 Chr.1: 236719154-236719154 on Build GRCh38
rs561234020 Chr.1: 236719281-236719281 on Build GRCh38
rs373987899 Chr.1: 236719283-236719283 on Build GRCh38
rs765948528 Chr.1: 236718848-236718848 on Build GRCh38
rs750757768 Chr.1: 236718962-236718962 on Build GRCh38
rs528474133 Chr.1: 236719289-236719289 on Build GRCh38
rs202213263 Chr.1: 236719032-236719032 on Build GRCh38
rs760787515 Chr.1: 236718929-236718929 on Build GRCh38
rs867564727 Chr.1: 236718920-236718920 on Build GRCh38
rs781400031 Chr.1: 236719190-236719190 on Build GRCh38
rs139424431 Chr.1: 236718896-236718896 on Build GRCh38
rs754609390 Chr.1: 236718850-236718850 on Build GRCh38
rs115748872 Chr.1: 236719150-236719150 on Build GRCh38
rs370020363 Chr.1: 236719042-236719042 on Build GRCh38
rs546727550 Chr.1: 236718978-236718978 on Build GRCh38
rs567437714 Chr.1: 236719287-236719287 on Build GRCh38
rs905266034 Chr.1: 236719085-236719085 on Build GRCh38
rs986321378 Chr.1: 236719028-236719028 on Build GRCh38
rs1057005244 Chr.1: 236719249-236719249 on Build GRCh38
rs973145308 Chr.1: 236718970-236718970 on Build GRCh38
rs963214658 Chr.1: 236718934-236718934 on Build GRCh38
rs1017614949 Chr.1: 236719129-236719129 on Build GRCh38
rs987221795 Chr.1: 236719222-236719222 on Build GRCh38
rs536983633 Chr.1: 236719063-236719063 on Build GRCh38
rs560136212 Chr.1: 236719180-236719180 on Build GRCh38
rs41269355 Chr.1: 236719109-236719109 on Build GRCh38
rs762095615 Chr.1: 236718931-236718931 on Build GRCh38
rs775761356 Chr.1: 236718927-236718927 on Build GRCh38
rs1004436349 Chr.1: 236719125-236719125 on Build GRCh38
rs1341864 Chr.1: 236719003-236719003 on Build GRCh38
rs539250948 Chr.1: 236719006-236719006 on Build GRCh38
rs921392642 Chr.1: 236718955-236718955 on Build GRCh38
rs140092735 Chr.1: 236719151-236719151 on Build GRCh38
rs377715235 Chr.1: 236719034-236719034 on Build GRCh38
rs905064442 Chr.1: 236719295-236719295 on Build GRCh38
rs878951518 Chr.1: 236718895-236718895 on Build GRCh38
rs557641600 Chr.1: 236719047-236719047 on Build GRCh38
rs786204949 Chr.1: 236718984-236718984 on Build GRCh38
rs373067784 Chr.1: 236719043-236719043 on Build GRCh38
rs181799482 Chr.1: 236718841-236718841 on Build GRCh38

View Full Product Details