Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241629977-241630490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHML
Entrez Gene ID
1122
Gene Name
CHM like, Rab escort protein 2
Gene Aliases
REP2
UniGene
Hs.654545
Gene Chromosome Location
Chr.1: 241628864-241635930 on Build GRCh38
Transcripts
NM_001821.3 NM_014322.2

Gene Symbol
OPN3
Entrez Gene ID
23596
Gene Name
opsin 3
Gene Aliases
ECPN, PPP1R116
UniGene
Hs.409081
Gene Chromosome Location
Chr.1: 241593150-241640399 on Build GRCh38
Transcripts
NM_001821.3 NM_014322.2

SNP Detail

SNP ID Chromosome Location
rs991579845 Chr.1: 241630022-241630022 on Build GRCh38
rs557760002 Chr.1: 241630357-241630357 on Build GRCh38
rs776445432 Chr.1: 241630161-241630161 on Build GRCh38
rs1000585332 Chr.1: 241630010-241630010 on Build GRCh38
rs991299321 Chr.1: 241630192-241630192 on Build GRCh38
rs148061875 Chr.1: 241630251-241630251 on Build GRCh38
rs1033027938 Chr.1: 241630013-241630013 on Build GRCh38
rs761517701 Chr.1: 241630174-241630174 on Build GRCh38
rs971534670 Chr.1: 241630370-241630370 on Build GRCh38
rs901633802 Chr.1: 241630221-241630221 on Build GRCh38
rs993631438 Chr.1: 241630147-241630147 on Build GRCh38
rs143529414 Chr.1: 241630201-241630201 on Build GRCh38
rs2756225 Chr.1: 241630375-241630375 on Build GRCh38
rs150584159 Chr.1: 241630174-241630175 on Build GRCh38
rs1000467306 Chr.1: 241630269-241630269 on Build GRCh38
rs369233646 Chr.1: 241630155-241630155 on Build GRCh38
rs919155261 Chr.1: 241630093-241630093 on Build GRCh38
rs915798230 Chr.1: 241630193-241630193 on Build GRCh38
rs943082990 Chr.1: 241630180-241630180 on Build GRCh38
rs764694167 Chr.1: 241630206-241630206 on Build GRCh38
rs930601452 Chr.1: 241630122-241630122 on Build GRCh38
rs1050628319 Chr.1: 241630074-241630074 on Build GRCh38
rs538472015 Chr.1: 241630133-241630133 on Build GRCh38
rs371953669 Chr.1: 241630331-241630331 on Build GRCh38
rs919116915 Chr.1: 241630413-241630413 on Build GRCh38
rs889347877 Chr.1: 241630407-241630410 on Build GRCh38
rs910225293 Chr.1: 241630167-241630167 on Build GRCh38
rs758131803 Chr.1: 241630291-241630294 on Build GRCh38
rs958700851 Chr.1: 241630293-241630293 on Build GRCh38
rs145498333 Chr.1: 241630047-241630047 on Build GRCh38
rs12038871 Chr.1: 241630024-241630024 on Build GRCh38
rs553575234 Chr.1: 241630249-241630249 on Build GRCh38
rs1009191137 Chr.1: 241630417-241630417 on Build GRCh38
rs574186579 Chr.1: 241630244-241630244 on Build GRCh38
rs1018969567 Chr.1: 241630126-241630126 on Build GRCh38
rs543003113 Chr.1: 241630246-241630246 on Build GRCh38
rs952021812 Chr.1: 241630418-241630418 on Build GRCh38
rs914327330 Chr.1: 241630353-241630353 on Build GRCh38
rs768512687 Chr.1: 241630063-241630063 on Build GRCh38
rs998745102 Chr.1: 241630233-241630233 on Build GRCh38
rs148880894 Chr.1: 241630165-241630165 on Build GRCh38
rs531080726 Chr.1: 241630059-241630059 on Build GRCh38
rs955688777 Chr.1: 241630440-241630440 on Build GRCh38
rs1054171877 Chr.1: 241630298-241630298 on Build GRCh38
rs984674439 Chr.1: 241630449-241630449 on Build GRCh38
rs958751009 Chr.1: 241630019-241630019 on Build GRCh38
rs962435562 Chr.1: 241630129-241630129 on Build GRCh38
rs1032980525 Chr.1: 241630273-241630273 on Build GRCh38
rs543547045 Chr.1: 241630389-241630389 on Build GRCh38

View Full Product Details