Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241630323-241630871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHML
Entrez Gene ID
1122
Gene Name
CHM like, Rab escort protein 2
Gene Aliases
REP2
UniGene
Hs.654545
Gene Chromosome Location
Chr.1: 241628864-241635930 on Build GRCh38
Transcripts
NM_001821.3 NM_014322.2

Gene Symbol
OPN3
Entrez Gene ID
23596
Gene Name
opsin 3
Gene Aliases
ECPN, PPP1R116
UniGene
Hs.409081
Gene Chromosome Location
Chr.1: 241593150-241640399 on Build GRCh38
Transcripts
NM_001821.3 NM_014322.2

SNP Detail

SNP ID Chromosome Location
rs1049077033 Chr.1: 241630784-241630784 on Build GRCh38
rs557760002 Chr.1: 241630357-241630357 on Build GRCh38
rs142315394 Chr.1: 241630469-241630469 on Build GRCh38
rs1026526010 Chr.1: 241630757-241630757 on Build GRCh38
rs971534670 Chr.1: 241630370-241630370 on Build GRCh38
rs924533663 Chr.1: 241630615-241630615 on Build GRCh38
rs575534212 Chr.1: 241630614-241630614 on Build GRCh38
rs945162754 Chr.1: 241630571-241630571 on Build GRCh38
rs2756225 Chr.1: 241630375-241630375 on Build GRCh38
rs559088652 Chr.1: 241630543-241630543 on Build GRCh38
rs781492118 Chr.1: 241630684-241630684 on Build GRCh38
rs3765820 Chr.1: 241630754-241630754 on Build GRCh38
rs191325541 Chr.1: 241630843-241630843 on Build GRCh38
rs766906897 Chr.1: 241630519-241630519 on Build GRCh38
rs975747628 Chr.1: 241630505-241630505 on Build GRCh38
rs955694277 Chr.1: 241630634-241630634 on Build GRCh38
rs936199529 Chr.1: 241630616-241630616 on Build GRCh38
rs1012995425 Chr.1: 241630811-241630811 on Build GRCh38
rs910754374 Chr.1: 241630715-241630715 on Build GRCh38
rs146133115 Chr.1: 241630669-241630669 on Build GRCh38
rs919116915 Chr.1: 241630413-241630413 on Build GRCh38
rs542436785 Chr.1: 241630521-241630521 on Build GRCh38
rs889347877 Chr.1: 241630407-241630410 on Build GRCh38
rs1023214265 Chr.1: 241630836-241630839 on Build GRCh38
rs944877194 Chr.1: 241630737-241630737 on Build GRCh38
rs971719819 Chr.1: 241630563-241630563 on Build GRCh38
rs780547466 Chr.1: 241630841-241630842 on Build GRCh38
rs951910080 Chr.1: 241630810-241630810 on Build GRCh38
rs530819991 Chr.1: 241630835-241630835 on Build GRCh38
rs1009191137 Chr.1: 241630417-241630417 on Build GRCh38
rs1054712212 Chr.1: 241630628-241630628 on Build GRCh38
rs1042247543 Chr.1: 241630587-241630587 on Build GRCh38
rs952021812 Chr.1: 241630418-241630418 on Build GRCh38
rs943030554 Chr.1: 241630501-241630501 on Build GRCh38
rs887845883 Chr.1: 241630797-241630797 on Build GRCh38
rs543449882 Chr.1: 241630738-241630738 on Build GRCh38
rs914327330 Chr.1: 241630353-241630353 on Build GRCh38
rs989833338 Chr.1: 241630638-241630639 on Build GRCh38
rs1012910432 Chr.1: 241630730-241630730 on Build GRCh38
rs373412622 Chr.1: 241630679-241630679 on Build GRCh38
rs562117101 Chr.1: 241630740-241630740 on Build GRCh38
rs955688777 Chr.1: 241630440-241630440 on Build GRCh38
rs984674439 Chr.1: 241630449-241630449 on Build GRCh38
rs573356307 Chr.1: 241630509-241630509 on Build GRCh38
rs993460469 Chr.1: 241630499-241630500 on Build GRCh38
rs914296381 Chr.1: 241630647-241630647 on Build GRCh38
rs1050579250 Chr.1: 241630704-241630704 on Build GRCh38
rs530983924 Chr.1: 241630819-241630819 on Build GRCh38
rs543547045 Chr.1: 241630389-241630389 on Build GRCh38

View Full Product Details