Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241634108-241634616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHML
Entrez Gene ID
1122
Gene Name
CHM like, Rab escort protein 2
Gene Aliases
REP2
UniGene
Hs.654545
Gene Chromosome Location
Chr.1: 241628864-241635930 on Build GRCh38
Transcripts
NM_001821.3 NM_014322.2

Gene Symbol
OPN3
Entrez Gene ID
23596
Gene Name
opsin 3
Gene Aliases
ECPN, PPP1R116
UniGene
Hs.409081
Gene Chromosome Location
Chr.1: 241593150-241640399 on Build GRCh38
Transcripts
NM_001821.3 NM_014322.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628458 c.1541C>A p.S514Y Chr.1: 241634226-241634226 on Build GRCh38
4569035 c.1464T>A p.A488A Chr.1: 241634303-241634303 on Build GRCh38
1560749 c.1301T>G p.F434C Chr.1: 241634466-241634466 on Build GRCh38
4436994 c.1644C>T p.L548L Chr.1: 241634123-241634123 on Build GRCh38
6628455 c.1617C>T p.N539N Chr.1: 241634150-241634150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926903802 Chr.1: 241634501-241634501 on Build GRCh38
rs772929547 Chr.1: 241634559-241634559 on Build GRCh38
rs532140585 Chr.1: 241634341-241634341 on Build GRCh38
rs551842840 Chr.1: 241634427-241634427 on Build GRCh38
rs372360469 Chr.1: 241634489-241634489 on Build GRCh38
rs780997451 Chr.1: 241634503-241634503 on Build GRCh38
rs775779940 Chr.1: 241634139-241634139 on Build GRCh38
rs80336474 Chr.1: 241634517-241634517 on Build GRCh38
rs758380488 Chr.1: 241634393-241634393 on Build GRCh38
rs780047465 Chr.1: 241634404-241634404 on Build GRCh38
rs187777115 Chr.1: 241634198-241634198 on Build GRCh38
rs775871937 Chr.1: 241634221-241634221 on Build GRCh38
rs779959593 Chr.1: 241634299-241634299 on Build GRCh38
rs762564140 Chr.1: 241634560-241634560 on Build GRCh38
rs202246923 Chr.1: 241634554-241634554 on Build GRCh38
rs375680469 Chr.1: 241634333-241634333 on Build GRCh38
rs754747195 Chr.1: 241634502-241634502 on Build GRCh38
rs767840796 Chr.1: 241634136-241634136 on Build GRCh38
rs747297803 Chr.1: 241634300-241634300 on Build GRCh38
rs761801338 Chr.1: 241634326-241634326 on Build GRCh38
rs754689407 Chr.1: 241634161-241634161 on Build GRCh38
rs370499285 Chr.1: 241634163-241634163 on Build GRCh38
rs758394640 Chr.1: 241634556-241634556 on Build GRCh38
rs764883955 Chr.1: 241634487-241634487 on Build GRCh38
rs769387506 Chr.1: 241634537-241634537 on Build GRCh38
rs765655048 Chr.1: 241634256-241634256 on Build GRCh38
rs145858464 Chr.1: 241634302-241634302 on Build GRCh38
rs139853468 Chr.1: 241634298-241634298 on Build GRCh38
rs375203343 Chr.1: 241634179-241634179 on Build GRCh38
rs760842619 Chr.1: 241634140-241634140 on Build GRCh38
rs191276616 Chr.1: 241634435-241634435 on Build GRCh38
rs79951803 Chr.1: 241634539-241634539 on Build GRCh38
rs756733101 Chr.1: 241634374-241634374 on Build GRCh38
rs758772974 Chr.1: 241634263-241634263 on Build GRCh38
rs776835459 Chr.1: 241634322-241634322 on Build GRCh38
rs747736759 Chr.1: 241634171-241634171 on Build GRCh38
rs866842597 Chr.1: 241634254-241634254 on Build GRCh38
rs868426815 Chr.1: 241634167-241634167 on Build GRCh38
rs995384456 Chr.1: 241634307-241634307 on Build GRCh38
rs531402599 Chr.1: 241634462-241634462 on Build GRCh38
rs561929720 Chr.1: 241634250-241634250 on Build GRCh38
rs760764453 Chr.1: 241634233-241634233 on Build GRCh38
rs576100300 Chr.1: 241634155-241634155 on Build GRCh38
rs759826725 Chr.1: 241634451-241634451 on Build GRCh38
rs140810282 Chr.1: 241634533-241634533 on Build GRCh38
rs772357239 Chr.1: 241634536-241634536 on Build GRCh38
rs765861577 Chr.1: 241634562-241634562 on Build GRCh38
rs777390084 Chr.1: 241634279-241634279 on Build GRCh38
rs778521313 Chr.1: 241634379-241634379 on Build GRCh38
rs753464772 Chr.1: 241634360-241634360 on Build GRCh38
rs201448683 Chr.1: 241634252-241634252 on Build GRCh38
rs746772887 Chr.1: 241634408-241634408 on Build GRCh38
rs750858068 Chr.1: 241634259-241634259 on Build GRCh38
rs763651268 Chr.1: 241634359-241634359 on Build GRCh38
rs145348026 Chr.1: 241634327-241634327 on Build GRCh38
rs879175089 Chr.1: 241634371-241634371 on Build GRCh38
rs752377996 Chr.1: 241634268-241634268 on Build GRCh38
rs778999435 Chr.1: 241634294-241634294 on Build GRCh38
rs757667040 Chr.1: 241634149-241634149 on Build GRCh38
rs868049122 Chr.1: 241634550-241634550 on Build GRCh38
rs767264972 Chr.1: 241634581-241634581 on Build GRCh38
rs755690197 Chr.1: 241634270-241634270 on Build GRCh38
rs765127182 Chr.1: 241634429-241634429 on Build GRCh38
rs771927047 Chr.1: 241634216-241634216 on Build GRCh38
rs1055581232 Chr.1: 241634385-241634385 on Build GRCh38
rs777320912 Chr.1: 241634257-241634257 on Build GRCh38
rs765676878 Chr.1: 241634150-241634150 on Build GRCh38
rs745654356 Chr.1: 241634207-241634207 on Build GRCh38
rs774848074 Chr.1: 241634447-241634447 on Build GRCh38
rs771412466 Chr.1: 241634446-241634446 on Build GRCh38
rs753456268 Chr.1: 241634482-241634482 on Build GRCh38
rs533890577 Chr.1: 241634558-241634558 on Build GRCh38
rs550461531 Chr.1: 241634415-241634415 on Build GRCh38
rs758472006 Chr.1: 241634492-241634492 on Build GRCh38
rs1004033225 Chr.1: 241634180-241634180 on Build GRCh38
rs376250907 Chr.1: 241634160-241634160 on Build GRCh38
rs200021744 Chr.1: 241634165-241634165 on Build GRCh38
rs755671388 Chr.1: 241634172-241634172 on Build GRCh38
rs368722099 Chr.1: 241634356-241634356 on Build GRCh38
rs776169905 Chr.1: 241634275-241634275 on Build GRCh38
rs368941629 Chr.1: 241634534-241634534 on Build GRCh38
rs761521790 Chr.1: 241634484-241634484 on Build GRCh38
rs371441164 Chr.1: 241634547-241634547 on Build GRCh38
rs766466619 Chr.1: 241634589-241634589 on Build GRCh38
rs766680773 Chr.1: 241634265-241634265 on Build GRCh38
rs777107327 Chr.1: 241634174-241634174 on Build GRCh38
rs938015015 Chr.1: 241634513-241634513 on Build GRCh38
rs748358849 Chr.1: 241634514-241634514 on Build GRCh38
rs763762314 Chr.1: 241634264-241634264 on Build GRCh38
rs376791831 Chr.1: 241634523-241634523 on Build GRCh38
rs532237518 Chr.1: 241634246-241634246 on Build GRCh38
rs757288530 Chr.1: 241634285-241634285 on Build GRCh38
rs556621444 Chr.1: 241634147-241634147 on Build GRCh38
rs773243934 Chr.1: 241634432-241634432 on Build GRCh38
rs760144305 Chr.1: 241634242-241634242 on Build GRCh38
rs751845326 Chr.1: 241634352-241634352 on Build GRCh38
rs761610842 Chr.1: 241634295-241634295 on Build GRCh38
rs547660354 Chr.1: 241634568-241634568 on Build GRCh38
rs1028233433 Chr.1: 241634320-241634320 on Build GRCh38
rs773726766 Chr.1: 241634332-241634332 on Build GRCh38
rs750421802 Chr.1: 241634383-241634383 on Build GRCh38
rs777886665 Chr.1: 241634518-241634518 on Build GRCh38
rs780142087 Chr.1: 241634494-241634494 on Build GRCh38
rs371934916 Chr.1: 241634498-241634498 on Build GRCh38
rs372387524 Chr.1: 241634238-241634238 on Build GRCh38
rs748735293 Chr.1: 241634283-241634283 on Build GRCh38
rs754911781 Chr.1: 241634571-241634571 on Build GRCh38
rs770752839 Chr.1: 241634191-241634191 on Build GRCh38

View Full Product Details