Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241853216-241853733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXO1
Entrez Gene ID
9156
Gene Name
exonuclease 1
Gene Aliases
HEX1, hExoI
UniGene
Hs.498248
Gene Chromosome Location
Chr.1: 241847967-241889939 on Build GRCh38
Transcripts
NM_001319224.1 NM_003686.4 NM_006027.4 NM_130398.3 XM_006711840.2 XM_011544321.1 XM_011544322.1 XM_011544323.2 XM_011544324.2 XM_017002793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
333367 c.349G>T p.E117* Chr.1: 241853425-241853425 on Build GRCh38
3977356 c.282-2A>T p.? Chr.1: 241853356-241853356 on Build GRCh38
906508 c.286C>T p.R96* Chr.1: 241853362-241853362 on Build GRCh38
2050246 c.325G>C p.E109Q Chr.1: 241853401-241853401 on Build GRCh38
2050244 c.322C>T p.R108C Chr.1: 241853398-241853398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751977225 Chr.1: 241853453-241853453 on Build GRCh38
rs769401961 Chr.1: 241853500-241853500 on Build GRCh38
rs147687027 Chr.1: 241853399-241853399 on Build GRCh38
rs371763760 Chr.1: 241853423-241853423 on Build GRCh38
rs143546023 Chr.1: 241853401-241853401 on Build GRCh38
rs930104982 Chr.1: 241853629-241853629 on Build GRCh38
rs757519301 Chr.1: 241853476-241853476 on Build GRCh38
rs369994389 Chr.1: 241853351-241853351 on Build GRCh38
rs962425574 Chr.1: 241853596-241853596 on Build GRCh38
rs745735907 Chr.1: 241853644-241853644 on Build GRCh38
rs192239018 Chr.1: 241853574-241853574 on Build GRCh38
rs759905066 Chr.1: 241853455-241853455 on Build GRCh38
rs748843701 Chr.1: 241853512-241853512 on Build GRCh38
rs756251971 Chr.1: 241853402-241853402 on Build GRCh38
rs562099707 Chr.1: 241853297-241853298 on Build GRCh38
rs768005950 Chr.1: 241853456-241853456 on Build GRCh38
rs770625941 Chr.1: 241853635-241853635 on Build GRCh38
rs775266335 Chr.1: 241853336-241853336 on Build GRCh38
rs764989865 Chr.1: 241853363-241853363 on Build GRCh38
rs79036854 Chr.1: 241853435-241853435 on Build GRCh38
rs775943886 Chr.1: 241853531-241853531 on Build GRCh38
rs963947369 Chr.1: 241853429-241853429 on Build GRCh38
rs973584795 Chr.1: 241853625-241853625 on Build GRCh38
rs768725549 Chr.1: 241853433-241853433 on Build GRCh38
rs769489836 Chr.1: 241853439-241853439 on Build GRCh38
rs751980406 Chr.1: 241853396-241853396 on Build GRCh38
rs766612173 Chr.1: 241853447-241853447 on Build GRCh38
rs542092590 Chr.1: 241853328-241853328 on Build GRCh38
rs367890594 Chr.1: 241853415-241853415 on Build GRCh38
rs4149867 Chr.1: 241853288-241853288 on Build GRCh38
rs141567727 Chr.1: 241853403-241853403 on Build GRCh38
rs1018305982 Chr.1: 241853685-241853685 on Build GRCh38
rs538813878 Chr.1: 241853549-241853549 on Build GRCh38
rs4149868 Chr.1: 241853536-241853536 on Build GRCh38
rs777957014 Chr.1: 241853412-241853412 on Build GRCh38
rs766819069 Chr.1: 241853389-241853389 on Build GRCh38
rs761495515 Chr.1: 241853362-241853362 on Build GRCh38
rs753259264 Chr.1: 241853464-241853464 on Build GRCh38
rs745643236 Chr.1: 241853330-241853330 on Build GRCh38
rs750170334 Chr.1: 241853365-241853365 on Build GRCh38
rs140831046 Chr.1: 241853445-241853445 on Build GRCh38
rs778823480 Chr.1: 241853636-241853636 on Build GRCh38
rs756164023 Chr.1: 241853467-241853467 on Build GRCh38
rs779927405 Chr.1: 241853422-241853422 on Build GRCh38
rs552656525 Chr.1: 241853652-241853652 on Build GRCh38
rs550672150 Chr.1: 241853344-241853344 on Build GRCh38
rs774515338 Chr.1: 241853518-241853518 on Build GRCh38
rs755709932 Chr.1: 241853438-241853438 on Build GRCh38
rs781264960 Chr.1: 241853492-241853492 on Build GRCh38
rs937249908 Chr.1: 241853505-241853505 on Build GRCh38
rs537162676 Chr.1: 241853593-241853593 on Build GRCh38
rs368245110 Chr.1: 241853501-241853501 on Build GRCh38
rs766837333 Chr.1: 241853343-241853343 on Build GRCh38
rs777870811 Chr.1: 241853469-241853469 on Build GRCh38
rs746822778 Chr.1: 241853489-241853489 on Build GRCh38
rs374873745 Chr.1: 241853437-241853437 on Build GRCh38
rs745845276 Chr.1: 241853452-241853452 on Build GRCh38
rs761201455 Chr.1: 241853532-241853532 on Build GRCh38
rs148384257 Chr.1: 241853331-241853331 on Build GRCh38
rs767916211 Chr.1: 241853523-241853523 on Build GRCh38
rs577634324 Chr.1: 241853580-241853580 on Build GRCh38
rs1635515 Chr.1: 241853707-241853707 on Build GRCh38
rs753963521 Chr.1: 241853470-241853470 on Build GRCh38
rs759879002 Chr.1: 241853520-241853520 on Build GRCh38
rs373710484 Chr.1: 241853414-241853414 on Build GRCh38
rs748149063 Chr.1: 241853496-241853496 on Build GRCh38
rs567681802 Chr.1: 241853398-241853398 on Build GRCh38
rs779653956 Chr.1: 241853479-241853479 on Build GRCh38
rs777913418 Chr.1: 241853668-241853668 on Build GRCh38
rs768641664 Chr.1: 241853490-241853490 on Build GRCh38
rs770027738 Chr.1: 241853325-241853325 on Build GRCh38
rs377234143 Chr.1: 241853513-241853513 on Build GRCh38
rs925630651 Chr.1: 241853292-241853292 on Build GRCh38
rs372531016 Chr.1: 241853503-241853503 on Build GRCh38
rs1010356491 Chr.1: 241853320-241853320 on Build GRCh38

View Full Product Details