Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241861301-241861843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXO1
Entrez Gene ID
9156
Gene Name
exonuclease 1
Gene Aliases
HEX1, hExoI
UniGene
Hs.498248
Gene Chromosome Location
Chr.1: 241847967-241889939 on Build GRCh38
Transcripts
NM_001319224.1 NM_003686.4 NM_006027.4 NM_130398.3 XM_006711840.2 XM_011544321.1 XM_011544322.1 XM_011544323.2 XM_011544324.2 XM_011544325.2 XM_017002793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
354749 c.1020C>T p.D340D Chr.1: 241861481-241861481 on Build GRCh38
4451022 c.1024A>C p.N342H Chr.1: 241861485-241861485 on Build GRCh38
320188 c.1041+1G>T p.? Chr.1: 241861503-241861503 on Build GRCh38
906519 c.983G>T p.G328V Chr.1: 241861444-241861444 on Build GRCh38
7331810 c.992A>T p.D331V Chr.1: 241861453-241861453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023745543 Chr.1: 241861588-241861588 on Build GRCh38
rs368445876 Chr.1: 241861472-241861472 on Build GRCh38
rs746216292 Chr.1: 241861505-241861505 on Build GRCh38
rs754051450 Chr.1: 241861422-241861422 on Build GRCh38
rs562376702 Chr.1: 241861777-241861777 on Build GRCh38
rs4149920 Chr.1: 241861626-241861626 on Build GRCh38
rs910365869 Chr.1: 241861695-241861695 on Build GRCh38
rs763433772 Chr.1: 241861370-241861372 on Build GRCh38
rs77512261 Chr.1: 241861782-241861782 on Build GRCh38
rs540978517 Chr.1: 241861800-241861800 on Build GRCh38
rs4149918 Chr.1: 241861551-241861551 on Build GRCh38
rs367673354 Chr.1: 241861508-241861508 on Build GRCh38
rs768331195 Chr.1: 241861597-241861597 on Build GRCh38
rs368140134 Chr.1: 241861397-241861397 on Build GRCh38
rs181271455 Chr.1: 241861620-241861620 on Build GRCh38
rs962272292 Chr.1: 241861814-241861814 on Build GRCh38
rs992990935 Chr.1: 241861799-241861799 on Build GRCh38
rs890255017 Chr.1: 241861611-241861611 on Build GRCh38
rs112699482 Chr.1: 241861465-241861465 on Build GRCh38
rs184996983 Chr.1: 241861793-241861793 on Build GRCh38
rs766465530 Chr.1: 241861422-241861433 on Build GRCh38
rs749960068 Chr.1: 241861457-241861457 on Build GRCh38
rs772902358 Chr.1: 241861377-241861377 on Build GRCh38
rs564088202 Chr.1: 241861586-241861586 on Build GRCh38
rs75296817 Chr.1: 241861783-241861783 on Build GRCh38
rs781262681 Chr.1: 241861471-241861471 on Build GRCh38
rs751405652 Chr.1: 241861462-241861462 on Build GRCh38
rs1009235743 Chr.1: 241861612-241861612 on Build GRCh38
rs969437225 Chr.1: 241861368-241861368 on Build GRCh38
rs770586588 Chr.1: 241861486-241861486 on Build GRCh38
rs1021902557 Chr.1: 241861738-241861738 on Build GRCh38
rs905340236 Chr.1: 241861809-241861809 on Build GRCh38
rs761116678 Chr.1: 241861514-241861514 on Build GRCh38
rs1022332669 Chr.1: 241861636-241861636 on Build GRCh38
rs764660825 Chr.1: 241861445-241861445 on Build GRCh38
rs766217496 Chr.1: 241861453-241861453 on Build GRCh38
rs956697245 Chr.1: 241861618-241861618 on Build GRCh38
rs532953643 Chr.1: 241861602-241861602 on Build GRCh38
rs750732573 Chr.1: 241861540-241861540 on Build GRCh38
rs556253108 Chr.1: 241861380-241861380 on Build GRCh38
rs762061524 Chr.1: 241861526-241861526 on Build GRCh38
rs576262524 Chr.1: 241861451-241861451 on Build GRCh38
rs932074517 Chr.1: 241861594-241861594 on Build GRCh38
rs993040338 Chr.1: 241861507-241861507 on Build GRCh38
rs1035526351 Chr.1: 241861604-241861604 on Build GRCh38
rs927455927 Chr.1: 241861473-241861473 on Build GRCh38
rs4149917 Chr.1: 241861550-241861550 on Build GRCh38
rs372233535 Chr.1: 241861404-241861404 on Build GRCh38
rs775916013 Chr.1: 241861405-241861405 on Build GRCh38
rs779554284 Chr.1: 241861581-241861581 on Build GRCh38
rs774723194 Chr.1: 241861388-241861388 on Build GRCh38
rs759966857 Chr.1: 241861402-241861402 on Build GRCh38
rs1023799244 Chr.1: 241861332-241861332 on Build GRCh38
rs4149919 Chr.1: 241861621-241861621 on Build GRCh38
rs4149921 Chr.1: 241861820-241861820 on Build GRCh38
rs768730665 Chr.1: 241861522-241861522 on Build GRCh38
rs1003559187 Chr.1: 241861598-241861598 on Build GRCh38
rs372371375 Chr.1: 241861475-241861475 on Build GRCh38
rs748924199 Chr.1: 241861483-241861483 on Build GRCh38
rs944537017 Chr.1: 241861704-241861704 on Build GRCh38
rs769246356 Chr.1: 241861373-241861373 on Build GRCh38
rs767259099 Chr.1: 241861541-241861541 on Build GRCh38
rs892380667 Chr.1: 241861661-241861661 on Build GRCh38
rs376327531 Chr.1: 241861511-241861511 on Build GRCh38
rs148949114 Chr.1: 241861674-241861674 on Build GRCh38
rs968360177 Chr.1: 241861637-241861637 on Build GRCh38
rs144135764 Chr.1: 241861423-241861423 on Build GRCh38
rs541298102 Chr.1: 241861566-241861566 on Build GRCh38
rs1776133 Chr.1: 241861796-241861796 on Build GRCh38
rs776585717 Chr.1: 241861525-241861525 on Build GRCh38
rs764120105 Chr.1: 241861409-241861409 on Build GRCh38
rs969557565 Chr.1: 241861755-241861755 on Build GRCh38
rs751628811 Chr.1: 241861431-241861432 on Build GRCh38
rs370613345 Chr.1: 241861548-241861548 on Build GRCh38
rs142940117 Chr.1: 241861732-241861732 on Build GRCh38
rs376803522 Chr.1: 241861492-241861492 on Build GRCh38
rs145785261 Chr.1: 241861452-241861452 on Build GRCh38
rs765554526 Chr.1: 241861535-241861535 on Build GRCh38
rs956337452 Chr.1: 241861344-241861344 on Build GRCh38
rs199816364 Chr.1: 241861545-241861545 on Build GRCh38
rs772470060 Chr.1: 241861506-241861506 on Build GRCh38
rs760783766 Chr.1: 241861408-241861408 on Build GRCh38
rs549242942 Chr.1: 241861696-241861696 on Build GRCh38
rs1034965554 Chr.1: 241861396-241861396 on Build GRCh38

View Full Product Details