Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244408414-244408923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSS
Entrez Gene ID
159
Gene Name
adenylosuccinate synthase
Gene Aliases
ADEH, ADSS 2
UniGene
Hs.498313
Gene Chromosome Location
Chr.1: 244408492-244452134 on Build GRCh38
Transcripts
NM_001126.3 XM_011544108.2

SNP Detail

SNP ID Chromosome Location
rs944419442 Chr.1: 244408712-244408712 on Build GRCh38
rs369680128 Chr.1: 244408501-244408501 on Build GRCh38
rs1036307266 Chr.1: 244408464-244408466 on Build GRCh38
rs775108524 Chr.1: 244408690-244408690 on Build GRCh38
rs562139270 Chr.1: 244408537-244408537 on Build GRCh38
rs1034281809 Chr.1: 244408488-244408488 on Build GRCh38
rs573638170 Chr.1: 244408485-244408485 on Build GRCh38
rs745750326 Chr.1: 244408471-244408471 on Build GRCh38
rs1023671917 Chr.1: 244408532-244408532 on Build GRCh38
rs982507699 Chr.1: 244408634-244408635 on Build GRCh38
rs748845095 Chr.1: 244408443-244408443 on Build GRCh38
rs541478611 Chr.1: 244408689-244408689 on Build GRCh38
rs977875288 Chr.1: 244408579-244408579 on Build GRCh38
rs546910013 Chr.1: 244408634-244408635 on Build GRCh38
rs952805163 Chr.1: 244408513-244408513 on Build GRCh38
rs774331138 Chr.1: 244408582-244408582 on Build GRCh38
rs770396282 Chr.1: 244408491-244408491 on Build GRCh38
rs907307653 Chr.1: 244408711-244408711 on Build GRCh38
rs115523508 Chr.1: 244408512-244408512 on Build GRCh38
rs1012198922 Chr.1: 244408850-244408850 on Build GRCh38
rs944410800 Chr.1: 244408637-244408637 on Build GRCh38
rs61084783 Chr.1: 244408635-244408636 on Build GRCh38
rs1022157577 Chr.1: 244408521-244408521 on Build GRCh38
rs768749251 Chr.1: 244408733-244408733 on Build GRCh38
rs775360653 Chr.1: 244408679-244408679 on Build GRCh38
rs397962172 Chr.1: 244408644-244408645 on Build GRCh38
rs932281312 Chr.1: 244408672-244408672 on Build GRCh38
rs940867561 Chr.1: 244408700-244408700 on Build GRCh38
rs1027404248 Chr.1: 244408486-244408486 on Build GRCh38
rs527766409 Chr.1: 244408615-244408615 on Build GRCh38
rs1036452995 Chr.1: 244408701-244408701 on Build GRCh38
rs556796676 Chr.1: 244408462-244408462 on Build GRCh38
rs1034335492 Chr.1: 244408742-244408742 on Build GRCh38
rs1022631568 Chr.1: 244408856-244408856 on Build GRCh38
rs759334673 Chr.1: 244408649-244408649 on Build GRCh38
rs564441604 Chr.1: 244408744-244408744 on Build GRCh38
rs907260142 Chr.1: 244408460-244408460 on Build GRCh38
rs375505040 Chr.1: 244408635-244408635 on Build GRCh38
rs769885759 Chr.1: 244408571-244408571 on Build GRCh38
rs35885954 Chr.1: 244408848-244408849 on Build GRCh38
rs921577846 Chr.1: 244408670-244408670 on Build GRCh38
rs1014961886 Chr.1: 244408454-244408454 on Build GRCh38
rs762121079 Chr.1: 244408795-244408795 on Build GRCh38
rs1050212103 Chr.1: 244408706-244408709 on Build GRCh38
rs762855148 Chr.1: 244408707-244408707 on Build GRCh38
rs897705824 Chr.1: 244408477-244408478 on Build GRCh38
rs995126994 Chr.1: 244408754-244408754 on Build GRCh38
rs959335098 Chr.1: 244408492-244408492 on Build GRCh38

View Full Product Details