Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244420020-244420568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSS
Entrez Gene ID
159
Gene Name
adenylosuccinate synthase
Gene Aliases
ADEH, ADSS 2
UniGene
Hs.498313
Gene Chromosome Location
Chr.1: 244408492-244452134 on Build GRCh38
Transcripts
NM_001126.3 XM_011544108.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4443181 c.758G>A p.G253D Chr.1: 244420202-244420202 on Build GRCh38
906675 c.734C>A p.P245Q Chr.1: 244420226-244420226 on Build GRCh38
3486383 c.747C>T p.I249I Chr.1: 244420213-244420213 on Build GRCh38
238718 c.777A>C p.L259F Chr.1: 244420183-244420183 on Build GRCh38
6595642 c.747C>A p.I249I Chr.1: 244420213-244420213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778380869 Chr.1: 244420232-244420232 on Build GRCh38
rs760897311 Chr.1: 244420265-244420265 on Build GRCh38
rs879312718 Chr.1: 244420087-244420087 on Build GRCh38
rs11434001 Chr.1: 244420499-244420500 on Build GRCh38
rs749216992 Chr.1: 244420212-244420212 on Build GRCh38
rs72503260 Chr.1: 244420498-244420499 on Build GRCh38
rs373510004 Chr.1: 244420150-244420150 on Build GRCh38
rs975009030 Chr.1: 244420300-244420300 on Build GRCh38
rs776911337 Chr.1: 244420279-244420279 on Build GRCh38
rs745321037 Chr.1: 244420347-244420347 on Build GRCh38
rs763138349 Chr.1: 244420296-244420296 on Build GRCh38
rs1020479110 Chr.1: 244420193-244420193 on Build GRCh38
rs769037132 Chr.1: 244420269-244420269 on Build GRCh38
rs370954450 Chr.1: 244420341-244420341 on Build GRCh38
rs1027554565 Chr.1: 244420400-244420400 on Build GRCh38
rs1055777527 Chr.1: 244420425-244420425 on Build GRCh38
rs777533847 Chr.1: 244420200-244420200 on Build GRCh38
rs758788917 Chr.1: 244420124-244420124 on Build GRCh38
rs55648210 Chr.1: 244420283-244420283 on Build GRCh38
rs780893723 Chr.1: 244420407-244420407 on Build GRCh38
rs765388497 Chr.1: 244420139-244420139 on Build GRCh38
rs900070209 Chr.1: 244420349-244420349 on Build GRCh38
rs546702796 Chr.1: 244420127-244420127 on Build GRCh38
rs574365469 Chr.1: 244420368-244420368 on Build GRCh38
rs990031875 Chr.1: 244420468-244420468 on Build GRCh38
rs750462737 Chr.1: 244420295-244420295 on Build GRCh38
rs983268883 Chr.1: 244420424-244420424 on Build GRCh38
rs771643025 Chr.1: 244420253-244420253 on Build GRCh38
rs796590657 Chr.1: 244420417-244420417 on Build GRCh38
rs73129710 Chr.1: 244420438-244420438 on Build GRCh38
rs999986120 Chr.1: 244420209-244420209 on Build GRCh38
rs557526534 Chr.1: 244420385-244420385 on Build GRCh38
rs766661968 Chr.1: 244420299-244420299 on Build GRCh38
rs745448883 Chr.1: 244420246-244420246 on Build GRCh38
rs754388754 Chr.1: 244420126-244420126 on Build GRCh38
rs777599985 Chr.1: 244420304-244420304 on Build GRCh38
rs780428510 Chr.1: 244420141-244420141 on Build GRCh38
rs937239724 Chr.1: 244420462-244420462 on Build GRCh38
rs775133709 Chr.1: 244420258-244420258 on Build GRCh38
rs540303345 Chr.1: 244420370-244420370 on Build GRCh38
rs566693557 Chr.1: 244420282-244420282 on Build GRCh38
rs113091014 Chr.1: 244420481-244420481 on Build GRCh38
rs1025024586 Chr.1: 244420434-244420434 on Build GRCh38
rs369938263 Chr.1: 244420122-244420122 on Build GRCh38
rs973522090 Chr.1: 244420372-244420372 on Build GRCh38
rs1038189628 Chr.1: 244420053-244420053 on Build GRCh38
rs530164941 Chr.1: 244420058-244420058 on Build GRCh38
rs937293150 Chr.1: 244420079-244420079 on Build GRCh38
rs896048687 Chr.1: 244420100-244420100 on Build GRCh38
rs980741930 Chr.1: 244420447-244420447 on Build GRCh38
rs372593853 Chr.1: 244420497-244420498 on Build GRCh38
rs903329680 Chr.1: 244420194-244420194 on Build GRCh38
rs565765101 Chr.1: 244420052-244420052 on Build GRCh38
rs908411756 Chr.1: 244420367-244420369 on Build GRCh38
rs751840380 Chr.1: 244420302-244420302 on Build GRCh38
rs895805397 Chr.1: 244420433-244420433 on Build GRCh38
rs774793929 Chr.1: 244420303-244420303 on Build GRCh38
rs34630643 Chr.1: 244420500-244420501 on Build GRCh38
rs766784410 Chr.1: 244420140-244420140 on Build GRCh38
rs377557638 Chr.1: 244420340-244420340 on Build GRCh38
rs778822431 Chr.1: 244420345-244420345 on Build GRCh38
rs554587213 Chr.1: 244420355-244420355 on Build GRCh38
rs537424652 Chr.1: 244420353-244420353 on Build GRCh38
rs368841454 Chr.1: 244420148-244420148 on Build GRCh38
rs770820204 Chr.1: 244420224-244420224 on Build GRCh38

View Full Product Details