Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244424107-244424652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSS
Entrez Gene ID
159
Gene Name
adenylosuccinate synthase
Gene Aliases
ADEH, ADSS 2
UniGene
Hs.498313
Gene Chromosome Location
Chr.1: 244408492-244452134 on Build GRCh38
Transcripts
NM_001126.3 XM_011544108.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6400019 c.463G>T p.A155S Chr.1: 244424331-244424331 on Build GRCh38
340564 c.452G>A p.R151K Chr.1: 244424342-244424342 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755823515 Chr.1: 244424315-244424315 on Build GRCh38
rs1047692588 Chr.1: 244424529-244424529 on Build GRCh38
rs901505675 Chr.1: 244424172-244424172 on Build GRCh38
rs555887754 Chr.1: 244424278-244424278 on Build GRCh38
rs770601470 Chr.1: 244424437-244424437 on Build GRCh38
rs368627472 Chr.1: 244424449-244424449 on Build GRCh38
rs946904362 Chr.1: 244424433-244424433 on Build GRCh38
rs753435720 Chr.1: 244424580-244424580 on Build GRCh38
rs779544471 Chr.1: 244424296-244424297 on Build GRCh38
rs897946985 Chr.1: 244424445-244424445 on Build GRCh38
rs1023994614 Chr.1: 244424496-244424496 on Build GRCh38
rs992489369 Chr.1: 244424230-244424230 on Build GRCh38
rs367573748 Chr.1: 244424281-244424281 on Build GRCh38
rs925113459 Chr.1: 244424554-244424554 on Build GRCh38
rs887667834 Chr.1: 244424535-244424535 on Build GRCh38
rs922994209 Chr.1: 244424581-244424581 on Build GRCh38
rs887610583 Chr.1: 244424232-244424232 on Build GRCh38
rs772682043 Chr.1: 244424392-244424392 on Build GRCh38
rs761484302 Chr.1: 244424605-244424605 on Build GRCh38
rs766917351 Chr.1: 244424295-244424295 on Build GRCh38
rs888836716 Chr.1: 244424130-244424130 on Build GRCh38
rs772617162 Chr.1: 244424260-244424260 on Build GRCh38
rs1040114497 Chr.1: 244424455-244424455 on Build GRCh38
rs112000889 Chr.1: 244424336-244424336 on Build GRCh38
rs191529595 Chr.1: 244424616-244424616 on Build GRCh38
rs979706860 Chr.1: 244424531-244424531 on Build GRCh38
rs371893179 Chr.1: 244424219-244424219 on Build GRCh38
rs6679230 Chr.1: 244424427-244424427 on Build GRCh38
rs752311188 Chr.1: 244424304-244424304 on Build GRCh38
rs956497850 Chr.1: 244424562-244424562 on Build GRCh38
rs960732514 Chr.1: 244424330-244424330 on Build GRCh38
rs80144373 Chr.1: 244424167-244424167 on Build GRCh38
rs768811848 Chr.1: 244424400-244424400 on Build GRCh38
rs1026200261 Chr.1: 244424204-244424204 on Build GRCh38
rs988807807 Chr.1: 244424356-244424356 on Build GRCh38
rs916469315 Chr.1: 244424251-244424251 on Build GRCh38
rs201241047 Chr.1: 244424395-244424395 on Build GRCh38
rs928945115 Chr.1: 244424526-244424526 on Build GRCh38
rs535859204 Chr.1: 244424245-244424245 on Build GRCh38
rs1012893161 Chr.1: 244424468-244424468 on Build GRCh38
rs1006131171 Chr.1: 244424567-244424567 on Build GRCh38
rs979268231 Chr.1: 244424265-244424265 on Build GRCh38
rs185943178 Chr.1: 244424218-244424218 on Build GRCh38
rs969317345 Chr.1: 244424518-244424518 on Build GRCh38
rs199554228 Chr.1: 244424296-244424296 on Build GRCh38
rs944399445 Chr.1: 244424584-244424584 on Build GRCh38
rs1035685900 Chr.1: 244424151-244424151 on Build GRCh38
rs761649478 Chr.1: 244424161-244424161 on Build GRCh38
rs371336132 Chr.1: 244424329-244424329 on Build GRCh38
rs374011195 Chr.1: 244424338-244424338 on Build GRCh38
rs922941839 Chr.1: 244424327-244424327 on Build GRCh38
rs541616269 Chr.1: 244424331-244424331 on Build GRCh38
rs750267039 Chr.1: 244424257-244424257 on Build GRCh38
rs759559314 Chr.1: 244424294-244424294 on Build GRCh38
rs200336048 Chr.1: 244424412-244424412 on Build GRCh38
rs1006032827 Chr.1: 244424256-244424256 on Build GRCh38
rs572617552 Chr.1: 244424284-244424284 on Build GRCh38
rs993854619 Chr.1: 244424173-244424173 on Build GRCh38
rs993527204 Chr.1: 244424146-244424146 on Build GRCh38
rs543437584 Chr.1: 244424448-244424448 on Build GRCh38
rs943120734 Chr.1: 244424156-244424156 on Build GRCh38
rs905694967 Chr.1: 244424611-244424611 on Build GRCh38
rs767047121 Chr.1: 244424282-244424284 on Build GRCh38
rs772928790 Chr.1: 244424274-244424274 on Build GRCh38
rs746327258 Chr.1: 244424381-244424381 on Build GRCh38
rs181053396 Chr.1: 244424166-244424166 on Build GRCh38
rs148509609 Chr.1: 244424347-244424347 on Build GRCh38
rs564020511 Chr.1: 244424447-244424447 on Build GRCh38

View Full Product Details