Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5097323-5097846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1C3
Entrez Gene ID
8644
Gene Name
aldo-keto reductase family 1, member C3
Gene Aliases
DD3, DDX, HA1753, HAKRB, HAKRe, HSD17B5, PGFS, hluPGFS
UniGene
Hs.78183
Gene Chromosome Location
Chr.10: 5048766-5107686 on Build GRCh38
Transcripts
NM_001253908.1 NM_001253909.1 NM_003739.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3985634 c.331G>C p.V111L Chr.10: 5097512-5097512 on Build GRCh38
3438498 c.346A>T p.I116F Chr.10: 5097527-5097527 on Build GRCh38
3985633 c.260C>G p.S87C Chr.10: 5097441-5097441 on Build GRCh38
3438497 c.298G>A p.E100K Chr.10: 5097479-5097479 on Build GRCh38
3438496 c.272G>A p.R91Q Chr.10: 5097453-5097453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782397389 Chr.10: 5097620-5097620 on Build GRCh38
rs782433833 Chr.10: 5097477-5097477 on Build GRCh38
rs570684197 Chr.10: 5097733-5097733 on Build GRCh38
rs782303259 Chr.10: 5097528-5097528 on Build GRCh38
rs370856934 Chr.10: 5097580-5097580 on Build GRCh38
rs782219293 Chr.10: 5097460-5097460 on Build GRCh38
rs186198146 Chr.10: 5097771-5097771 on Build GRCh38
rs782049519 Chr.10: 5097416-5097416 on Build GRCh38
rs781983530 Chr.10: 5097488-5097488 on Build GRCh38
rs782620878 Chr.10: 5097503-5097503 on Build GRCh38
rs975094272 Chr.10: 5097770-5097770 on Build GRCh38
rs968016064 Chr.10: 5097732-5097732 on Build GRCh38
rs183337621 Chr.10: 5097602-5097602 on Build GRCh38
rs782642613 Chr.10: 5097568-5097568 on Build GRCh38
rs1033134049 Chr.10: 5097750-5097750 on Build GRCh38
rs960316891 Chr.10: 5097506-5097506 on Build GRCh38
rs967542977 Chr.10: 5097543-5097543 on Build GRCh38
rs567515422 Chr.10: 5097744-5097744 on Build GRCh38
rs781805668 Chr.10: 5097508-5097508 on Build GRCh38
rs782551582 Chr.10: 5097493-5097493 on Build GRCh38
rs185230294 Chr.10: 5097388-5097388 on Build GRCh38
rs4987103 Chr.10: 5097553-5097553 on Build GRCh38
rs782604138 Chr.10: 5097611-5097611 on Build GRCh38
rs782282013 Chr.10: 5097673-5097673 on Build GRCh38
rs782421321 Chr.10: 5097539-5097539 on Build GRCh38
rs782250047 Chr.10: 5097400-5097400 on Build GRCh38
rs550984088 Chr.10: 5097660-5097660 on Build GRCh38
rs782754665 Chr.10: 5097636-5097636 on Build GRCh38
rs2298305 Chr.10: 5097694-5097694 on Build GRCh38
rs942348188 Chr.10: 5097397-5097397 on Build GRCh38
rs782363753 Chr.10: 5097467-5097467 on Build GRCh38
rs955041536 Chr.10: 5097775-5097775 on Build GRCh38
rs782010496 Chr.10: 5097633-5097633 on Build GRCh38
rs577759811 Chr.10: 5097489-5097489 on Build GRCh38
rs963802926 Chr.10: 5097762-5097762 on Build GRCh38
rs80144615 Chr.10: 5097724-5097724 on Build GRCh38
rs528884532 Chr.10: 5097565-5097565 on Build GRCh38
rs782015261 Chr.10: 5097422-5097422 on Build GRCh38
rs148886004 Chr.10: 5097434-5097434 on Build GRCh38
rs2245191 Chr.10: 5097623-5097623 on Build GRCh38
rs190674338 Chr.10: 5097498-5097498 on Build GRCh38
rs781820694 Chr.10: 5097570-5097570 on Build GRCh38
rs782769157 Chr.10: 5097427-5097427 on Build GRCh38
rs782445635 Chr.10: 5097559-5097560 on Build GRCh38
rs782458578 Chr.10: 5097449-5097449 on Build GRCh38
rs781871241 Chr.10: 5097500-5097500 on Build GRCh38
rs782581919 Chr.10: 5097468-5097468 on Build GRCh38
rs552317814 Chr.10: 5097702-5097702 on Build GRCh38
rs978498949 Chr.10: 5097355-5097355 on Build GRCh38
rs782314956 Chr.10: 5097626-5097626 on Build GRCh38
rs531233762 Chr.10: 5097545-5097545 on Build GRCh38
rs782188615 Chr.10: 5097529-5097530 on Build GRCh38
rs118150330 Chr.10: 5097452-5097452 on Build GRCh38
rs781959902 Chr.10: 5097414-5097414 on Build GRCh38
rs540534605 Chr.10: 5097518-5097518 on Build GRCh38
rs201680078 Chr.10: 5097517-5097517 on Build GRCh38
rs782729191 Chr.10: 5097551-5097551 on Build GRCh38
rs533106161 Chr.10: 5097659-5097659 on Build GRCh38
rs551990617 Chr.10: 5097692-5097692 on Build GRCh38
rs563553179 Chr.10: 5097649-5097649 on Build GRCh38
rs12387 Chr.10: 5097493-5097493 on Build GRCh38
rs1001615574 Chr.10: 5097651-5097651 on Build GRCh38
rs782158776 Chr.10: 5097425-5097425 on Build GRCh38
rs782497370 Chr.10: 5097453-5097453 on Build GRCh38
rs782790991 Chr.10: 5097562-5097562 on Build GRCh38
rs895780568 Chr.10: 5097672-5097672 on Build GRCh38
rs562560711 Chr.10: 5097628-5097628 on Build GRCh38
rs782048590 Chr.10: 5097491-5097491 on Build GRCh38
rs782153871 Chr.10: 5097538-5097538 on Build GRCh38
rs1039327830 Chr.10: 5097401-5097401 on Build GRCh38
rs782202734 Chr.10: 5097629-5097629 on Build GRCh38
rs556187295 Chr.10: 5097817-5097817 on Build GRCh38
rs781855690 Chr.10: 5097479-5097480 on Build GRCh38
rs782386139 Chr.10: 5097803-5097803 on Build GRCh38
rs782603138 Chr.10: 5097520-5097520 on Build GRCh38
rs145644085 Chr.10: 5097513-5097513 on Build GRCh38
rs527951734 Chr.10: 5097703-5097703 on Build GRCh38
rs782535113 Chr.10: 5097510-5097510 on Build GRCh38
rs982835849 Chr.10: 5097777-5097777 on Build GRCh38
rs908482292 Chr.10: 5097787-5097787 on Build GRCh38
rs202144998 Chr.10: 5097470-5097470 on Build GRCh38
rs782396026 Chr.10: 5097411-5097411 on Build GRCh38
rs782661431 Chr.10: 5097457-5097457 on Build GRCh38
rs781868397 Chr.10: 5097431-5097431 on Build GRCh38
rs782744331 Chr.10: 5097641-5097641 on Build GRCh38
rs782050484 Chr.10: 5097560-5097560 on Build GRCh38
rs376418943 Chr.10: 5097383-5097383 on Build GRCh38
rs782604645 Chr.10: 5097612-5097612 on Build GRCh38
rs782493365 Chr.10: 5097607-5097607 on Build GRCh38
rs754732117 Chr.10: 5097409-5097409 on Build GRCh38
rs782466786 Chr.10: 5097431-5097431 on Build GRCh38
rs4987102 Chr.10: 5097497-5097497 on Build GRCh38
rs782136936 Chr.10: 5097548-5097548 on Build GRCh38
rs782222522 Chr.10: 5097527-5097527 on Build GRCh38
rs782155599 Chr.10: 5097634-5097634 on Build GRCh38
rs781961155 Chr.10: 5097490-5097490 on Build GRCh38
rs140441894 Chr.10: 5097502-5097502 on Build GRCh38
rs782687251 Chr.10: 5097587-5097587 on Build GRCh38
rs782773422 Chr.10: 5097518-5097519 on Build GRCh38
rs555441815 Chr.10: 5097419-5097419 on Build GRCh38
rs782495340 Chr.10: 5097571-5097571 on Build GRCh38
rs941266101 Chr.10: 5097789-5097789 on Build GRCh38
rs200981816 Chr.10: 5097450-5097450 on Build GRCh38

View Full Product Details