Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12093697-12094237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHTKD1
Entrez Gene ID
55526
Gene Name
dehydrogenase E1 and transketolase domain containing 1
Gene Aliases
AMOXAD, CMT2Q
UniGene
Hs.104980
Gene Chromosome Location
Chr.10: 12068917-12123228 on Build GRCh38
Transcripts
NM_018706.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4523471 c.1216G>A p.V406I Chr.10: 12094129-12094129 on Build GRCh38
4012085 c.1307A>T p.N436I Chr.10: 12094220-12094220 on Build GRCh38
4012084 c.1250G>A p.R417H Chr.10: 12094163-12094163 on Build GRCh38
915378 c.1177G>A p.A393T Chr.10: 12094090-12094090 on Build GRCh38
6458797 c.1240_1241insGGCC p.E414fs*14 Chr.10: 12094153-12094154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755722138 Chr.10: 12094140-12094140 on Build GRCh38
rs1024809552 Chr.10: 12093945-12093945 on Build GRCh38
rs768983753 Chr.10: 12094045-12094045 on Build GRCh38
rs1004699868 Chr.10: 12094204-12094204 on Build GRCh38
rs965252389 Chr.10: 12094158-12094158 on Build GRCh38
rs768943532 Chr.10: 12093890-12093890 on Build GRCh38
rs397514534 Chr.10: 12094141-12094141 on Build GRCh38
rs755773040 Chr.10: 12094198-12094198 on Build GRCh38
rs772581919 Chr.10: 12094063-12094063 on Build GRCh38
rs761350668 Chr.10: 12094180-12094180 on Build GRCh38
rs767323403 Chr.10: 12094105-12094105 on Build GRCh38
rs145036232 Chr.10: 12094213-12094213 on Build GRCh38
rs528683749 Chr.10: 12093978-12093978 on Build GRCh38
rs946310068 Chr.10: 12093841-12093841 on Build GRCh38
rs562162834 Chr.10: 12094055-12094055 on Build GRCh38
rs762746645 Chr.10: 12094171-12094171 on Build GRCh38
rs1014457149 Chr.10: 12094000-12094000 on Build GRCh38
rs759869851 Chr.10: 12094162-12094162 on Build GRCh38
rs373872630 Chr.10: 12094208-12094208 on Build GRCh38
rs7086420 Chr.10: 12093889-12093889 on Build GRCh38
rs775165668 Chr.10: 12094170-12094170 on Build GRCh38
rs771115245 Chr.10: 12094161-12094161 on Build GRCh38
rs758294466 Chr.10: 12094104-12094104 on Build GRCh38
rs772458687 Chr.10: 12094146-12094146 on Build GRCh38
rs148613315 Chr.10: 12094197-12094197 on Build GRCh38
rs140878433 Chr.10: 12094098-12094098 on Build GRCh38
rs879165011 Chr.10: 12093923-12093923 on Build GRCh38
rs750669119 Chr.10: 12093773-12093773 on Build GRCh38
rs748482829 Chr.10: 12094142-12094142 on Build GRCh38
rs750044852 Chr.10: 12094139-12094139 on Build GRCh38
rs773175080 Chr.10: 12094066-12094066 on Build GRCh38
rs766772195 Chr.10: 12094086-12094086 on Build GRCh38
rs776977187 Chr.10: 12094093-12094093 on Build GRCh38
rs34033944 Chr.10: 12094175-12094175 on Build GRCh38
rs749722181 Chr.10: 12094028-12094028 on Build GRCh38
rs963395488 Chr.10: 12094006-12094006 on Build GRCh38
rs973415471 Chr.10: 12094039-12094039 on Build GRCh38
rs748633070 Chr.10: 12094051-12094051 on Build GRCh38
rs376642629 Chr.10: 12094099-12094099 on Build GRCh38
rs752500384 Chr.10: 12094101-12094101 on Build GRCh38
rs921042581 Chr.10: 12093816-12093816 on Build GRCh38
rs77813085 Chr.10: 12093767-12093767 on Build GRCh38
rs564925185 Chr.10: 12093800-12093800 on Build GRCh38
rs943864101 Chr.10: 12093804-12093804 on Build GRCh38
rs780153747 Chr.10: 12093873-12093873 on Build GRCh38
rs542484980 Chr.10: 12094008-12094008 on Build GRCh38
rs779372510 Chr.10: 12094132-12094132 on Build GRCh38
rs896037466 Chr.10: 12093940-12093940 on Build GRCh38
rs150179532 Chr.10: 12094107-12094107 on Build GRCh38
rs112134857 Chr.10: 12093900-12093900 on Build GRCh38
rs932397754 Chr.10: 12093832-12093832 on Build GRCh38
rs200039178 Chr.10: 12094172-12094172 on Build GRCh38
rs970512205 Chr.10: 12093948-12093948 on Build GRCh38
rs774960836 Chr.10: 12094050-12094050 on Build GRCh38
rs760767010 Chr.10: 12094072-12094072 on Build GRCh38
rs368956603 Chr.10: 12093810-12093810 on Build GRCh38
rs576789276 Chr.10: 12093809-12093809 on Build GRCh38
rs778336638 Chr.10: 12094155-12094155 on Build GRCh38
rs201455968 Chr.10: 12094133-12094133 on Build GRCh38
rs913738477 Chr.10: 12093792-12093792 on Build GRCh38
rs751037184 Chr.10: 12094120-12094120 on Build GRCh38
rs770009261 Chr.10: 12094167-12094167 on Build GRCh38
rs1053451589 Chr.10: 12094143-12094143 on Build GRCh38
rs909555248 Chr.10: 12094206-12094206 on Build GRCh38
rs189162856 Chr.10: 12093788-12093788 on Build GRCh38
rs767042646 Chr.10: 12094181-12094181 on Build GRCh38
rs1041924273 Chr.10: 12093851-12093851 on Build GRCh38
rs768618938 Chr.10: 12094202-12094202 on Build GRCh38
rs1014825390 Chr.10: 12093959-12093959 on Build GRCh38
rs1004814519 Chr.10: 12093958-12093958 on Build GRCh38
rs894889999 Chr.10: 12093898-12093898 on Build GRCh38
rs373471268 Chr.10: 12094023-12094023 on Build GRCh38
rs200722918 Chr.10: 12094159-12094159 on Build GRCh38

View Full Product Details