Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 16831137-16831678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUBN
Entrez Gene ID
8029
Gene Name
cubilin
Gene Aliases
IFCR, MGA1, gp280
UniGene
Hs.166206
Gene Chromosome Location
Chr.10: 16823966-17130492 on Build GRCh38
Transcripts
NM_001081.3 XM_011519709.2 XM_011519710.2 XM_011519711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
683787 c.10526C>G p.S3509C Chr.10: 16831254-16831254 on Build GRCh38
364959 c.10408_10409TG>TC p.C3470>? Chr.10: 16831371-16831372 on Build GRCh38
683786 c.10517C>T p.S3506L Chr.10: 16831263-16831263 on Build GRCh38
5603314 c.10450G>A p.E3484K Chr.10: 16831330-16831330 on Build GRCh38
3978324 c.10470G>A p.K3490K Chr.10: 16831310-16831310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929288763 Chr.10: 16831488-16831488 on Build GRCh38
rs771747676 Chr.10: 16831400-16831400 on Build GRCh38
rs376587594 Chr.10: 16831203-16831203 on Build GRCh38
rs887949730 Chr.10: 16831168-16831168 on Build GRCh38
rs774213488 Chr.10: 16831392-16831392 on Build GRCh38
rs141937843 Chr.10: 16831397-16831397 on Build GRCh38
rs749282166 Chr.10: 16831379-16831379 on Build GRCh38
rs775791705 Chr.10: 16831450-16831450 on Build GRCh38
rs964818975 Chr.10: 16831413-16831413 on Build GRCh38
rs1045430936 Chr.10: 16831580-16831580 on Build GRCh38
rs909503694 Chr.10: 16831520-16831520 on Build GRCh38
rs114249988 Chr.10: 16831468-16831468 on Build GRCh38
rs142481042 Chr.10: 16831346-16831346 on Build GRCh38
rs35038804 Chr.10: 16831620-16831621 on Build GRCh38
rs765943248 Chr.10: 16831420-16831420 on Build GRCh38
rs559936302 Chr.10: 16831602-16831602 on Build GRCh38
rs745992773 Chr.10: 16831480-16831480 on Build GRCh38
rs201426128 Chr.10: 16831318-16831318 on Build GRCh38
rs756099382 Chr.10: 16831370-16831370 on Build GRCh38
rs777566272 Chr.10: 16831455-16831455 on Build GRCh38
rs758170405 Chr.10: 16831430-16831430 on Build GRCh38
rs368486606 Chr.10: 16831411-16831411 on Build GRCh38
rs369280863 Chr.10: 16831406-16831406 on Build GRCh38
rs764524298 Chr.10: 16831317-16831317 on Build GRCh38
rs971066574 Chr.10: 16831315-16831315 on Build GRCh38
rs762706539 Chr.10: 16831258-16831258 on Build GRCh38
rs3740169 Chr.10: 16831548-16831548 on Build GRCh38
rs757611347 Chr.10: 16831319-16831319 on Build GRCh38
rs536614321 Chr.10: 16831176-16831176 on Build GRCh38
rs766959179 Chr.10: 16831283-16831283 on Build GRCh38
rs761464411 Chr.10: 16831272-16831272 on Build GRCh38
rs143300012 Chr.10: 16831287-16831287 on Build GRCh38
rs148331476 Chr.10: 16831383-16831383 on Build GRCh38
rs74787946 Chr.10: 16831175-16831175 on Build GRCh38
rs999193178 Chr.10: 16831391-16831391 on Build GRCh38
rs894261287 Chr.10: 16831223-16831223 on Build GRCh38
rs942265848 Chr.10: 16831542-16831542 on Build GRCh38
rs886046863 Chr.10: 16831314-16831314 on Build GRCh38
rs181848786 Chr.10: 16831288-16831288 on Build GRCh38
rs1055245983 Chr.10: 16831545-16831545 on Build GRCh38
rs151313566 Chr.10: 16831473-16831473 on Build GRCh38
rs765779982 Chr.10: 16831302-16831302 on Build GRCh38
rs764917718 Chr.10: 16831306-16831306 on Build GRCh38
rs779809669 Chr.10: 16831376-16831376 on Build GRCh38
rs1048449864 Chr.10: 16831510-16831510 on Build GRCh38
rs753266760 Chr.10: 16831304-16831304 on Build GRCh38
rs527476962 Chr.10: 16831605-16831605 on Build GRCh38
rs148820784 Chr.10: 16831163-16831163 on Build GRCh38
rs1055216654 Chr.10: 16831177-16831177 on Build GRCh38
rs576696171 Chr.10: 16831355-16831355 on Build GRCh38
rs775326988 Chr.10: 16831235-16831235 on Build GRCh38
rs760126310 Chr.10: 16831414-16831414 on Build GRCh38
rs773837995 Chr.10: 16831267-16831267 on Build GRCh38
rs1012641328 Chr.10: 16831571-16831571 on Build GRCh38
rs768521979 Chr.10: 16831266-16831266 on Build GRCh38

View Full Product Details